Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39097134-39097640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3H
Entrez Gene ID
164668
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3H
Gene Aliases
A3H, ARP-10, ARP10
UniGene
Hs.440515
Gene Chromosome Location
Chr.22: 39097204-39104067 on Build GRCh38
Transcripts
NM_001166002.2 NM_001166003.2 NM_001166004.2 NM_181773.4 XM_011529990.2 XM_011529991.2 XM_011529992.2

SNP Detail

SNP ID Chromosome Location
rs1046379599 Chr.22: 39097401-39097401 on Build GRCh38
rs553150913 Chr.22: 39097544-39097544 on Build GRCh38
rs981929410 Chr.22: 39097579-39097579 on Build GRCh38
rs954958924 Chr.22: 39097418-39097418 on Build GRCh38
rs934301054 Chr.22: 39097233-39097233 on Build GRCh38
rs905283568 Chr.22: 39097300-39097300 on Build GRCh38
rs924076954 Chr.22: 39097302-39097302 on Build GRCh38
rs1030563052 Chr.22: 39097541-39097541 on Build GRCh38
rs551367986 Chr.22: 39097449-39097449 on Build GRCh38
rs989530772 Chr.22: 39097562-39097562 on Build GRCh38
rs941030928 Chr.22: 39097603-39097603 on Build GRCh38
rs138782546 Chr.22: 39097537-39097537 on Build GRCh38
rs560277881 Chr.22: 39097219-39097244 on Build GRCh38
rs895726439 Chr.22: 39097247-39097247 on Build GRCh38
rs536816000 Chr.22: 39097519-39097519 on Build GRCh38
rs916378421 Chr.22: 39097377-39097377 on Build GRCh38
rs55989856 Chr.22: 39097289-39097289 on Build GRCh38
rs916658600 Chr.22: 39097563-39097563 on Build GRCh38
rs77603434 Chr.22: 39097549-39097549 on Build GRCh38
rs910343748 Chr.22: 39097182-39097182 on Build GRCh38
rs200360077 Chr.22: 39097522-39097524 on Build GRCh38
rs67577561 Chr.22: 39097592-39097592 on Build GRCh38
rs546545543 Chr.22: 39097313-39097313 on Build GRCh38
rs565075498 Chr.22: 39097609-39097609 on Build GRCh38
rs993447735 Chr.22: 39097395-39097395 on Build GRCh38
rs35338824 Chr.22: 39097156-39097156 on Build GRCh38
rs1006277033 Chr.22: 39097310-39097310 on Build GRCh38
rs534711097 Chr.22: 39097542-39097542 on Build GRCh38
rs1016363062 Chr.22: 39097202-39097202 on Build GRCh38
rs969249272 Chr.22: 39097569-39097569 on Build GRCh38
rs978556790 Chr.22: 39097462-39097462 on Build GRCh38
rs1030576058 Chr.22: 39097404-39097404 on Build GRCh38
rs56155393 Chr.22: 39097550-39097550 on Build GRCh38
rs569959243 Chr.22: 39097502-39097502 on Build GRCh38
rs954774363 Chr.22: 39097265-39097265 on Build GRCh38
rs149359564 Chr.22: 39097570-39097570 on Build GRCh38
rs947120666 Chr.22: 39097197-39097197 on Build GRCh38
rs532413658 Chr.22: 39097426-39097426 on Build GRCh38
rs868422615 Chr.22: 39097266-39097266 on Build GRCh38
rs371952738 Chr.22: 39097224-39097224 on Build GRCh38
rs540316728 Chr.22: 39097606-39097606 on Build GRCh38
rs542812158 Chr.22: 39097583-39097583 on Build GRCh38
rs968888435 Chr.22: 39097450-39097450 on Build GRCh38
rs561147397 Chr.22: 39097586-39097586 on Build GRCh38
rs1051678224 Chr.22: 39097615-39097615 on Build GRCh38
rs909507606 Chr.22: 39097598-39097598 on Build GRCh38
rs557342083 Chr.22: 39097557-39097557 on Build GRCh38
rs978845703 Chr.22: 39097222-39097222 on Build GRCh38
rs1005677161 Chr.22: 39097533-39097533 on Build GRCh38
rs543229509 Chr.22: 39097244-39097244 on Build GRCh38
rs747714349 Chr.22: 39097456-39097458 on Build GRCh38
rs60758775 Chr.22: 39097536-39097536 on Build GRCh38
rs41281251 Chr.22: 39097246-39097246 on Build GRCh38
rs60093835 Chr.22: 39097351-39097351 on Build GRCh38
rs958151153 Chr.22: 39097555-39097555 on Build GRCh38
rs982472256 Chr.22: 39097595-39097595 on Build GRCh38
rs955724373 Chr.22: 39097528-39097528 on Build GRCh38
rs1044212114 Chr.22: 39097290-39097290 on Build GRCh38
rs947959271 Chr.22: 39097396-39097396 on Build GRCh38
rs747018928 Chr.22: 39097288-39097288 on Build GRCh38
rs575727535 Chr.22: 39097190-39097190 on Build GRCh38
rs564044719 Chr.22: 39097184-39097184 on Build GRCh38

View Full Product Details