Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39099866-39100385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3H
Entrez Gene ID
164668
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3H
Gene Aliases
A3H, ARP-10, ARP10
UniGene
Hs.440515
Gene Chromosome Location
Chr.22: 39097204-39104067 on Build GRCh38
Transcripts
NM_001166002.2 NM_001166003.2 NM_001166004.2 NM_181773.4 XM_011529990.2 XM_011529991.2 XM_011529992.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4662517 c.19G>A p.E7K Chr.22: 39100297-39100297 on Build GRCh38
328621 c.45C>G p.N15K Chr.22: 39100323-39100323 on Build GRCh38
6329450 c.81G>T p.K27N Chr.22: 39100359-39100359 on Build GRCh38
5829219 c.40_42delAAC p.N15delN Chr.22: 39100318-39100320 on Build GRCh38
3759196 c.53G>T p.R18L Chr.22: 39100331-39100331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566197669 Chr.22: 39100205-39100205 on Build GRCh38
rs140936762 Chr.22: 39100318-39100320 on Build GRCh38
rs540973094 Chr.22: 39099937-39099937 on Build GRCh38
rs749450180 Chr.22: 39100233-39100233 on Build GRCh38
rs139293 Chr.22: 39100331-39100331 on Build GRCh38
rs920982883 Chr.22: 39099971-39099971 on Build GRCh38
rs189764428 Chr.22: 39100359-39100359 on Build GRCh38
rs774051298 Chr.22: 39100337-39100337 on Build GRCh38
rs1025049280 Chr.22: 39100219-39100219 on Build GRCh38
rs776130540 Chr.22: 39100253-39100253 on Build GRCh38
rs551901355 Chr.22: 39100034-39100034 on Build GRCh38
rs760113060 Chr.22: 39100297-39100297 on Build GRCh38
rs1020375404 Chr.22: 39099912-39099912 on Build GRCh38
rs139292 Chr.22: 39100323-39100325 on Build GRCh38
rs1055892265 Chr.22: 39100100-39100100 on Build GRCh38
rs749078969 Chr.22: 39100323-39100323 on Build GRCh38
rs937216265 Chr.22: 39100064-39100064 on Build GRCh38
rs746178426 Chr.22: 39100108-39100108 on Build GRCh38
rs139290 Chr.22: 39100129-39100129 on Build GRCh38
rs562547578 Chr.22: 39099917-39099917 on Build GRCh38
rs764240600 Chr.22: 39100336-39100337 on Build GRCh38
rs777073424 Chr.22: 39100242-39100247 on Build GRCh38
rs747509986 Chr.22: 39100328-39100328 on Build GRCh38
rs201177427 Chr.22: 39100322-39100324 on Build GRCh38
rs957733206 Chr.22: 39099988-39099988 on Build GRCh38
rs756279324 Chr.22: 39100229-39100229 on Build GRCh38
rs536088567 Chr.22: 39100177-39100177 on Build GRCh38
rs759128070 Chr.22: 39100341-39100341 on Build GRCh38
rs184469838 Chr.22: 39100105-39100105 on Build GRCh38
rs989551373 Chr.22: 39100003-39100003 on Build GRCh38
rs749399007 Chr.22: 39100311-39100311 on Build GRCh38
rs762492011 Chr.22: 39100332-39100332 on Build GRCh38
rs975205870 Chr.22: 39099954-39099954 on Build GRCh38
rs766070770 Chr.22: 39100281-39100281 on Build GRCh38
rs866502332 Chr.22: 39099942-39099942 on Build GRCh38
rs774804340 Chr.22: 39100240-39100240 on Build GRCh38
rs764738270 Chr.22: 39100267-39100267 on Build GRCh38
rs1027195849 Chr.22: 39099891-39099891 on Build GRCh38
rs139288 Chr.22: 39100011-39100011 on Build GRCh38
rs772425911 Chr.22: 39100249-39100249 on Build GRCh38
rs541516800 Chr.22: 39100002-39100002 on Build GRCh38
rs764174253 Chr.22: 39100360-39100360 on Build GRCh38
rs767346992 Chr.22: 39100306-39100306 on Build GRCh38
rs374766186 Chr.22: 39100327-39100327 on Build GRCh38
rs772740027 Chr.22: 39100277-39100277 on Build GRCh38
rs916867128 Chr.22: 39100081-39100081 on Build GRCh38
rs887388368 Chr.22: 39100128-39100128 on Build GRCh38
rs779316580 Chr.22: 39100318-39100318 on Build GRCh38
rs563806908 Chr.22: 39100112-39100112 on Build GRCh38
rs1019799395 Chr.22: 39100152-39100152 on Build GRCh38
rs559849715 Chr.22: 39100006-39100006 on Build GRCh38
rs372088508 Chr.22: 39100307-39100307 on Build GRCh38
rs937862707 Chr.22: 39100308-39100308 on Build GRCh38
rs754870937 Chr.22: 39100297-39100297 on Build GRCh38
rs989332734 Chr.22: 39099909-39099909 on Build GRCh38
rs767272509 Chr.22: 39100346-39100346 on Build GRCh38
rs761214813 Chr.22: 39100256-39100256 on Build GRCh38
rs958021312 Chr.22: 39100209-39100209 on Build GRCh38
rs139291 Chr.22: 39100194-39100194 on Build GRCh38
rs1048522321 Chr.22: 39100184-39100184 on Build GRCh38
rs997387325 Chr.22: 39100187-39100187 on Build GRCh38
rs965586301 Chr.22: 39100160-39100160 on Build GRCh38
rs193002279 Chr.22: 39100091-39100091 on Build GRCh38
rs931374735 Chr.22: 39100314-39100314 on Build GRCh38
rs746229527 Chr.22: 39100325-39100325 on Build GRCh38
rs777865663 Chr.22: 39100309-39100309 on Build GRCh38
rs539789572 Chr.22: 39100296-39100296 on Build GRCh38
rs991057105 Chr.22: 39100312-39100312 on Build GRCh38
rs142563386 Chr.22: 39100352-39100352 on Build GRCh38
rs576716153 Chr.22: 39099927-39099927 on Build GRCh38
rs76049870 Chr.22: 39100289-39100289 on Build GRCh38
rs771307499 Chr.22: 39100238-39100238 on Build GRCh38
rs1010218879 Chr.22: 39100270-39100270 on Build GRCh38
rs374189433 Chr.22: 39100246-39100246 on Build GRCh38
rs192445219 Chr.22: 39099898-39099898 on Build GRCh38
rs139620128 Chr.22: 39100336-39100336 on Build GRCh38
rs757446729 Chr.22: 39100037-39100037 on Build GRCh38
rs374487970 Chr.22: 39100087-39100087 on Build GRCh38
rs1004371140 Chr.22: 39099915-39099915 on Build GRCh38
rs757527498 Chr.22: 39100313-39100313 on Build GRCh38
rs139289 Chr.22: 39100080-39100080 on Build GRCh38
rs760023124 Chr.22: 39100273-39100274 on Build GRCh38
rs927802131 Chr.22: 39100088-39100088 on Build GRCh38
rs763213711 Chr.22: 39100322-39100323 on Build GRCh38
rs995559634 Chr.22: 39100109-39100109 on Build GRCh38
rs372401151 Chr.22: 39100232-39100232 on Build GRCh38
rs368714381 Chr.22: 39100278-39100278 on Build GRCh38
rs548365464 Chr.22: 39100099-39100099 on Build GRCh38

View Full Product Details