Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41515663-41516203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACO2
Entrez Gene ID
50
Gene Name
aconitase 2
Gene Aliases
ACONM, HEL-S-284, ICRD, OCA8, OPA9
UniGene
Hs.643610
Gene Chromosome Location
Chr.22: 41469125-41528989 on Build GRCh38
Transcripts
NM_001098.2 XM_017028812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6526622 c.765C>A p.L255L Chr.22: 41515847-41515847 on Build GRCh38
5393746 c.712C>T p.L238F Chr.22: 41515794-41515794 on Build GRCh38
5806208 c.835+1G>T p.? Chr.22: 41515918-41515918 on Build GRCh38
1034637 c.694G>A p.V232M Chr.22: 41515776-41515776 on Build GRCh38
5571009 c.686T>C p.V229A Chr.22: 41515768-41515768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749699669 Chr.22: 41515992-41515992 on Build GRCh38
rs141878785 Chr.22: 41515801-41515801 on Build GRCh38
rs965972541 Chr.22: 41516160-41516160 on Build GRCh38
rs779993600 Chr.22: 41515850-41515850 on Build GRCh38
rs993293125 Chr.22: 41515700-41515700 on Build GRCh38
rs371777558 Chr.22: 41515967-41515967 on Build GRCh38
rs1055958544 Chr.22: 41516047-41516048 on Build GRCh38
rs756677145 Chr.22: 41515985-41515985 on Build GRCh38
rs112533478 Chr.22: 41516041-41516041 on Build GRCh38
rs780553868 Chr.22: 41515746-41515746 on Build GRCh38
rs560262221 Chr.22: 41515886-41515886 on Build GRCh38
rs786204828 Chr.22: 41515858-41515858 on Build GRCh38
rs909127393 Chr.22: 41515735-41515735 on Build GRCh38
rs765850134 Chr.22: 41515692-41515692 on Build GRCh38
rs373316100 Chr.22: 41515940-41515940 on Build GRCh38
rs367893176 Chr.22: 41515959-41515959 on Build GRCh38
rs748474998 Chr.22: 41515874-41515874 on Build GRCh38
rs138803847 Chr.22: 41515799-41515799 on Build GRCh38
rs943054809 Chr.22: 41516171-41516171 on Build GRCh38
rs1051549990 Chr.22: 41516139-41516139 on Build GRCh38
rs199847369 Chr.22: 41515937-41515937 on Build GRCh38
rs754684779 Chr.22: 41515977-41515977 on Build GRCh38
rs771749582 Chr.22: 41515929-41515929 on Build GRCh38
rs776635095 Chr.22: 41515939-41515939 on Build GRCh38
rs115730463 Chr.22: 41515710-41515710 on Build GRCh38
rs763237550 Chr.22: 41515956-41515956 on Build GRCh38
rs748597339 Chr.22: 41515761-41515761 on Build GRCh38
rs1027444243 Chr.22: 41516116-41516116 on Build GRCh38
rs537367472 Chr.22: 41515711-41515711 on Build GRCh38
rs767849912 Chr.22: 41515969-41515969 on Build GRCh38
rs752210456 Chr.22: 41516138-41516138 on Build GRCh38
rs144602638 Chr.22: 41515861-41515861 on Build GRCh38
rs4510317 Chr.22: 41516087-41516087 on Build GRCh38
rs746435822 Chr.22: 41516029-41516029 on Build GRCh38
rs750687436 Chr.22: 41515822-41515822 on Build GRCh38
rs1014400996 Chr.22: 41516049-41516049 on Build GRCh38
rs751703387 Chr.22: 41515856-41515856 on Build GRCh38
rs111857969 Chr.22: 41515745-41515745 on Build GRCh38
rs779784760 Chr.22: 41516027-41516027 on Build GRCh38
rs751021941 Chr.22: 41515708-41515708 on Build GRCh38
rs1021918463 Chr.22: 41515802-41515802 on Build GRCh38
rs753220123 Chr.22: 41515984-41515984 on Build GRCh38
rs778164364 Chr.22: 41515916-41515916 on Build GRCh38
rs781380563 Chr.22: 41515752-41515752 on Build GRCh38
rs961784522 Chr.22: 41516090-41516090 on Build GRCh38
rs201718643 Chr.22: 41515732-41515732 on Build GRCh38
rs771553504 Chr.22: 41515787-41515787 on Build GRCh38
rs539202662 Chr.22: 41515717-41515717 on Build GRCh38
rs774028374 Chr.22: 41515775-41515775 on Build GRCh38
rs376654323 Chr.22: 41515770-41515770 on Build GRCh38
rs183827939 Chr.22: 41516134-41516134 on Build GRCh38
rs1049860 Chr.22: 41515806-41515806 on Build GRCh38
rs910295295 Chr.22: 41516143-41516143 on Build GRCh38
rs369933665 Chr.22: 41515928-41515928 on Build GRCh38
rs987955197 Chr.22: 41516178-41516178 on Build GRCh38
rs774836663 Chr.22: 41515933-41515933 on Build GRCh38
rs755420295 Chr.22: 41515857-41515857 on Build GRCh38
rs564524060 Chr.22: 41516047-41516047 on Build GRCh38
rs911834064 Chr.22: 41515892-41515892 on Build GRCh38
rs766802469 Chr.22: 41515942-41515942 on Build GRCh38
rs771906416 Chr.22: 41515830-41515830 on Build GRCh38
rs758753076 Chr.22: 41515849-41515849 on Build GRCh38
rs367627820 Chr.22: 41515731-41515731 on Build GRCh38
rs757936702 Chr.22: 41516019-41516019 on Build GRCh38
rs758983751 Chr.22: 41515786-41515786 on Build GRCh38
rs1043027906 Chr.22: 41515975-41515975 on Build GRCh38
rs375450980 Chr.22: 41515952-41515952 on Build GRCh38
rs557713361 Chr.22: 41515697-41515697 on Build GRCh38
rs765489176 Chr.22: 41515808-41515808 on Build GRCh38
rs17852363 Chr.22: 41515891-41515891 on Build GRCh38
rs764138297 Chr.22: 41515800-41515800 on Build GRCh38
rs753704051 Chr.22: 41515728-41515728 on Build GRCh38
rs769274301 Chr.22: 41515751-41515751 on Build GRCh38
rs747948591 Chr.22: 41515734-41515735 on Build GRCh38
rs960159463 Chr.22: 41515696-41515696 on Build GRCh38
rs145011805 Chr.22: 41515796-41515796 on Build GRCh38
rs747457781 Chr.22: 41515748-41515748 on Build GRCh38
rs1015509116 Chr.22: 41516060-41516060 on Build GRCh38
rs751548035 Chr.22: 41515966-41515966 on Build GRCh38
rs746549269 Chr.22: 41515936-41515936 on Build GRCh38
rs778076233 Chr.22: 41515991-41515991 on Build GRCh38
rs532870352 Chr.22: 41515949-41515949 on Build GRCh38
rs780785676 Chr.22: 41516072-41516072 on Build GRCh38
rs994985532 Chr.22: 41516112-41516112 on Build GRCh38

View Full Product Details