Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41517427-41517917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACO2
Entrez Gene ID
50
Gene Name
aconitase 2
Gene Aliases
ACONM, HEL-S-284, ICRD, OCA8, OPA9
UniGene
Hs.643610
Gene Chromosome Location
Chr.22: 41469125-41528989 on Build GRCh38
Transcripts
NM_001098.2 XM_017028812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1181609 c.865G>A p.E289K Chr.22: 41517556-41517556 on Build GRCh38
3800227 c.934C>G p.R312G Chr.22: 41517625-41517625 on Build GRCh38
1535423 c.885C>T p.S295S Chr.22: 41517576-41517576 on Build GRCh38
4642648 c.875C>T p.A292V Chr.22: 41517566-41517566 on Build GRCh38
6269133 c.915G>T p.K305N Chr.22: 41517606-41517606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979736696 Chr.22: 41517462-41517462 on Build GRCh38
rs949313271 Chr.22: 41517644-41517644 on Build GRCh38
rs372292167 Chr.22: 41517529-41517529 on Build GRCh38
rs766473354 Chr.22: 41517501-41517501 on Build GRCh38
rs184738307 Chr.22: 41517463-41517463 on Build GRCh38
rs148465407 Chr.22: 41517621-41517621 on Build GRCh38
rs762283798 Chr.22: 41517654-41517654 on Build GRCh38
rs754355417 Chr.22: 41517653-41517653 on Build GRCh38
rs776338407 Chr.22: 41517488-41517488 on Build GRCh38
rs748972301 Chr.22: 41517581-41517581 on Build GRCh38
rs891654033 Chr.22: 41517587-41517587 on Build GRCh38
rs774076940 Chr.22: 41517594-41517594 on Build GRCh38
rs918837646 Chr.22: 41517755-41517755 on Build GRCh38
rs767626111 Chr.22: 41517507-41517507 on Build GRCh38
rs536205838 Chr.22: 41517754-41517754 on Build GRCh38
rs748461464 Chr.22: 41517784-41517784 on Build GRCh38
rs994431097 Chr.22: 41517708-41517708 on Build GRCh38
rs962330728 Chr.22: 41517751-41517751 on Build GRCh38
rs17367716 Chr.22: 41517840-41517840 on Build GRCh38
rs754637681 Chr.22: 41517681-41517681 on Build GRCh38
rs964739031 Chr.22: 41517856-41517856 on Build GRCh38
rs1024515965 Chr.22: 41517623-41517623 on Build GRCh38
rs976356752 Chr.22: 41517857-41517857 on Build GRCh38
rs780818847 Chr.22: 41517566-41517566 on Build GRCh38
rs754654913 Chr.22: 41517547-41517547 on Build GRCh38
rs1026385999 Chr.22: 41517773-41517773 on Build GRCh38
rs145434182 Chr.22: 41517455-41517455 on Build GRCh38
rs576454118 Chr.22: 41517461-41517461 on Build GRCh38
rs986988866 Chr.22: 41517785-41517785 on Build GRCh38
rs373572985 Chr.22: 41517467-41517467 on Build GRCh38
rs545268377 Chr.22: 41517780-41517780 on Build GRCh38
rs764605230 Chr.22: 41517521-41517521 on Build GRCh38
rs937309980 Chr.22: 41517479-41517479 on Build GRCh38
rs560040600 Chr.22: 41517576-41517576 on Build GRCh38
rs773029843 Chr.22: 41517715-41517715 on Build GRCh38
rs760772363 Chr.22: 41517622-41517622 on Build GRCh38
rs781015104 Chr.22: 41517683-41517683 on Build GRCh38
rs774666692 Chr.22: 41517698-41517698 on Build GRCh38
rs203320 Chr.22: 41517473-41517473 on Build GRCh38
rs373585187 Chr.22: 41517489-41517489 on Build GRCh38
rs773121347 Chr.22: 41517498-41517498 on Build GRCh38
rs1039784701 Chr.22: 41517862-41517862 on Build GRCh38
rs750793467 Chr.22: 41517679-41517679 on Build GRCh38
rs572111776 Chr.22: 41517678-41517678 on Build GRCh38
rs925299703 Chr.22: 41517894-41517894 on Build GRCh38
rs757644783 Chr.22: 41517533-41517533 on Build GRCh38
rs749291290 Chr.22: 41517506-41517506 on Build GRCh38
rs759769140 Chr.22: 41517615-41517615 on Build GRCh38
rs966890415 Chr.22: 41517887-41517887 on Build GRCh38
rs756589026 Chr.22: 41517512-41517512 on Build GRCh38
rs912699488 Chr.22: 41517808-41517808 on Build GRCh38
rs61269773 Chr.22: 41517469-41517469 on Build GRCh38
rs752811289 Chr.22: 41517509-41517509 on Build GRCh38
rs775800876 Chr.22: 41517879-41517879 on Build GRCh38
rs1033990433 Chr.22: 41517704-41517704 on Build GRCh38
rs1024047591 Chr.22: 41517452-41517452 on Build GRCh38
rs746659632 Chr.22: 41517538-41517538 on Build GRCh38
rs189507216 Chr.22: 41517851-41517851 on Build GRCh38
rs1054362868 Chr.22: 41517572-41517572 on Build GRCh38
rs772325936 Chr.22: 41517618-41517618 on Build GRCh38
rs769893888 Chr.22: 41517494-41517494 on Build GRCh38
rs1049450202 Chr.22: 41517493-41517493 on Build GRCh38
rs762864872 Chr.22: 41517500-41517500 on Build GRCh38
rs201755525 Chr.22: 41517645-41517645 on Build GRCh38
rs375540731 Chr.22: 41517534-41517534 on Build GRCh38
rs923454736 Chr.22: 41517470-41517470 on Build GRCh38
rs200120553 Chr.22: 41517624-41517624 on Build GRCh38
rs777890868 Chr.22: 41517577-41517577 on Build GRCh38
rs541768137 Chr.22: 41517474-41517474 on Build GRCh38

View Full Product Details