Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41560891-41561387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSDC2
Entrez Gene ID
27254
Gene Name
cold shock domain containing C2
Gene Aliases
PIPPIN, dJ347H13.2
UniGene
Hs.310893
Gene Chromosome Location
Chr.22: 41561010-41576666 on Build GRCh38
Transcripts
NM_014460.3

SNP Detail

SNP ID Chromosome Location
rs761675733 Chr.22: 41561073-41561081 on Build GRCh38
rs934467528 Chr.22: 41561191-41561191 on Build GRCh38
rs929889532 Chr.22: 41561201-41561201 on Build GRCh38
rs1010973419 Chr.22: 41561046-41561047 on Build GRCh38
rs900608941 Chr.22: 41561330-41561330 on Build GRCh38
rs545613188 Chr.22: 41561027-41561027 on Build GRCh38
rs927073293 Chr.22: 41561162-41561162 on Build GRCh38
rs1051061190 Chr.22: 41560999-41560999 on Build GRCh38
rs571032929 Chr.22: 41561357-41561357 on Build GRCh38
rs534462001 Chr.22: 41561297-41561297 on Build GRCh38
rs561904853 Chr.22: 41561079-41561081 on Build GRCh38
rs927125987 Chr.22: 41560998-41560998 on Build GRCh38
rs550795688 Chr.22: 41561116-41561116 on Build GRCh38
rs559227215 Chr.22: 41561305-41561305 on Build GRCh38
rs560868541 Chr.22: 41561045-41561046 on Build GRCh38
rs765192525 Chr.22: 41561079-41561080 on Build GRCh38
rs890443751 Chr.22: 41561241-41561241 on Build GRCh38
rs141129549 Chr.22: 41561081-41561081 on Build GRCh38
rs562743769 Chr.22: 41561242-41561242 on Build GRCh38
rs1038628320 Chr.22: 41561172-41561172 on Build GRCh38
rs765257542 Chr.22: 41561074-41561075 on Build GRCh38
rs147660261 Chr.22: 41561075-41561081 on Build GRCh38
rs373662778 Chr.22: 41560961-41560961 on Build GRCh38
rs981392129 Chr.22: 41561046-41561053 on Build GRCh38
rs189518748 Chr.22: 41561243-41561243 on Build GRCh38
rs539790808 Chr.22: 41561026-41561026 on Build GRCh38
rs563926063 Chr.22: 41561029-41561029 on Build GRCh38
rs961130744 Chr.22: 41561067-41561081 on Build GRCh38
rs971075077 Chr.22: 41561152-41561152 on Build GRCh38
rs61037464 Chr.22: 41561082-41561083 on Build GRCh38
rs77059364 Chr.22: 41561355-41561355 on Build GRCh38
rs956494502 Chr.22: 41561139-41561139 on Build GRCh38
rs370966152 Chr.22: 41561053-41561053 on Build GRCh38
rs530049763 Chr.22: 41561138-41561138 on Build GRCh38
rs569277939 Chr.22: 41561137-41561137 on Build GRCh38
rs757119434 Chr.22: 41561072-41561073 on Build GRCh38
rs1048195679 Chr.22: 41561030-41561030 on Build GRCh38
rs899531162 Chr.22: 41560983-41560983 on Build GRCh38
rs979802091 Chr.22: 41561154-41561154 on Build GRCh38
rs905157950 Chr.22: 41561261-41561261 on Build GRCh38
rs574095187 Chr.22: 41561117-41561117 on Build GRCh38
rs41281291 Chr.22: 41560940-41560940 on Build GRCh38
rs964861860 Chr.22: 41561083-41561083 on Build GRCh38
rs567194167 Chr.22: 41561235-41561235 on Build GRCh38
rs941219549 Chr.22: 41561010-41561010 on Build GRCh38
rs941168632 Chr.22: 41561167-41561167 on Build GRCh38
rs763858929 Chr.22: 41561046-41561051 on Build GRCh38
rs1038237123 Chr.22: 41561025-41561025 on Build GRCh38
rs558076293 Chr.22: 41561031-41561031 on Build GRCh38
rs1022242476 Chr.22: 41561046-41561049 on Build GRCh38
rs1044156157 Chr.22: 41560958-41560958 on Build GRCh38
rs982965834 Chr.22: 41561122-41561122 on Build GRCh38
rs916968419 Chr.22: 41560924-41560924 on Build GRCh38
rs973852919 Chr.22: 41561142-41561142 on Build GRCh38
rs988902767 Chr.22: 41560913-41560913 on Build GRCh38
rs890496131 Chr.22: 41561033-41561033 on Build GRCh38
rs1035286142 Chr.22: 41561059-41561081 on Build GRCh38
rs13055812 Chr.22: 41561055-41561055 on Build GRCh38
rs533343515 Chr.22: 41561271-41561271 on Build GRCh38
rs891059692 Chr.22: 41561175-41561175 on Build GRCh38
rs918471676 Chr.22: 41561193-41561193 on Build GRCh38
rs988463177 Chr.22: 41561145-41561145 on Build GRCh38
rs370154480 Chr.22: 41561081-41561082 on Build GRCh38
rs201425136 Chr.22: 41561045-41561045 on Build GRCh38
rs916891593 Chr.22: 41561146-41561146 on Build GRCh38
rs573310247 Chr.22: 41561071-41561081 on Build GRCh38
rs908735146 Chr.22: 41561136-41561136 on Build GRCh38
rs1038025941 Chr.22: 41560963-41560963 on Build GRCh38
rs1000327788 Chr.22: 41561353-41561353 on Build GRCh38
rs776680683 Chr.22: 41561073-41561079 on Build GRCh38
rs527533480 Chr.22: 41560992-41560992 on Build GRCh38
rs763515158 Chr.22: 41561214-41561214 on Build GRCh38
rs760741127 Chr.22: 41561051-41561052 on Build GRCh38
rs1035656869 Chr.22: 41561304-41561304 on Build GRCh38
rs12484315 Chr.22: 41561212-41561212 on Build GRCh38

View Full Product Details