Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46685428-46685938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERK
Entrez Gene ID
64781
Gene Name
ceramide kinase
Gene Aliases
LK4, dA59H18.2, dA59H18.3, hCERK
UniGene
Hs.200668
Gene Chromosome Location
Chr.22: 46684410-46738255 on Build GRCh38
Transcripts
NM_022766.5 XM_017028909.1

SNP Detail

SNP ID Chromosome Location
rs1015460660 Chr.22: 46685806-46685806 on Build GRCh38
rs370657795 Chr.22: 46685737-46685737 on Build GRCh38
rs1050878631 Chr.22: 46685571-46685571 on Build GRCh38
rs938121321 Chr.22: 46685566-46685566 on Build GRCh38
rs553528779 Chr.22: 46685493-46685493 on Build GRCh38
rs376156919 Chr.22: 46685578-46685578 on Build GRCh38
rs73471135 Chr.22: 46685454-46685454 on Build GRCh38
rs73471136 Chr.22: 46685855-46685855 on Build GRCh38
rs936886954 Chr.22: 46685893-46685893 on Build GRCh38
rs62232569 Chr.22: 46685833-46685833 on Build GRCh38
rs925445119 Chr.22: 46685870-46685870 on Build GRCh38
rs904141060 Chr.22: 46685651-46685651 on Build GRCh38
rs555052121 Chr.22: 46685623-46685623 on Build GRCh38
rs1034119228 Chr.22: 46685776-46685776 on Build GRCh38
rs953754857 Chr.22: 46685825-46685825 on Build GRCh38
rs5769077 Chr.22: 46685516-46685516 on Build GRCh38
rs570722162 Chr.22: 46685498-46685498 on Build GRCh38
rs945550671 Chr.22: 46685590-46685590 on Build GRCh38
rs1004723058 Chr.22: 46685692-46685694 on Build GRCh38
rs1042650025 Chr.22: 46685600-46685600 on Build GRCh38
rs1021234324 Chr.22: 46685464-46685464 on Build GRCh38
rs1024238634 Chr.22: 46685544-46685544 on Build GRCh38
rs1042499409 Chr.22: 46685597-46685597 on Build GRCh38
rs532837533 Chr.22: 46685710-46685710 on Build GRCh38
rs994888185 Chr.22: 46685667-46685670 on Build GRCh38
rs891263510 Chr.22: 46685587-46685587 on Build GRCh38
rs764203999 Chr.22: 46685909-46685909 on Build GRCh38
rs191819073 Chr.22: 46685670-46685670 on Build GRCh38
rs989559367 Chr.22: 46685575-46685575 on Build GRCh38
rs900148259 Chr.22: 46685639-46685639 on Build GRCh38
rs536604020 Chr.22: 46685892-46685892 on Build GRCh38
rs560311249 Chr.22: 46685683-46685683 on Build GRCh38
rs527442854 Chr.22: 46685869-46685869 on Build GRCh38
rs571847533 Chr.22: 46685809-46685809 on Build GRCh38
rs56161409 Chr.22: 46685895-46685895 on Build GRCh38
rs574704462 Chr.22: 46685625-46685625 on Build GRCh38
rs544274493 Chr.22: 46685562-46685562 on Build GRCh38
rs908494396 Chr.22: 46685535-46685535 on Build GRCh38
rs982641460 Chr.22: 46685526-46685526 on Build GRCh38
rs1039139608 Chr.22: 46685638-46685638 on Build GRCh38
rs531761922 Chr.22: 46685738-46685738 on Build GRCh38
rs903838718 Chr.22: 46685479-46685479 on Build GRCh38
rs575849991 Chr.22: 46685548-46685548 on Build GRCh38
rs145632572 Chr.22: 46685511-46685520 on Build GRCh38
rs801721 Chr.22: 46685819-46685819 on Build GRCh38
rs1027421529 Chr.22: 46685687-46685687 on Build GRCh38
rs550432091 Chr.22: 46685760-46685760 on Build GRCh38
rs974183058 Chr.22: 46685547-46685547 on Build GRCh38
rs746662382 Chr.22: 46685773-46685773 on Build GRCh38
rs912344433 Chr.22: 46685840-46685840 on Build GRCh38
rs945560899 Chr.22: 46685596-46685596 on Build GRCh38
rs753337097 Chr.22: 46685510-46685511 on Build GRCh38
rs886156800 Chr.22: 46685691-46685691 on Build GRCh38
rs974025577 Chr.22: 46685818-46685818 on Build GRCh38
rs115734174 Chr.22: 46685539-46685539 on Build GRCh38
rs149250882 Chr.22: 46685716-46685716 on Build GRCh38
rs1024527483 Chr.22: 46685519-46685519 on Build GRCh38
rs971335798 Chr.22: 46685745-46685745 on Build GRCh38
rs1034058599 Chr.22: 46685703-46685703 on Build GRCh38
rs932359158 Chr.22: 46685565-46685565 on Build GRCh38
rs183424616 Chr.22: 46685899-46685899 on Build GRCh38
rs548146796 Chr.22: 46685525-46685525 on Build GRCh38
rs1033762172 Chr.22: 46685497-46685497 on Build GRCh38
rs920939394 Chr.22: 46685550-46685550 on Build GRCh38
rs539315648 Chr.22: 46685905-46685905 on Build GRCh38

View Full Product Details