Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94897201-94897742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNN3
Entrez Gene ID
1266
Gene Name
calponin 3
Gene Aliases
-
UniGene
Hs.483454
Gene Chromosome Location
Chr.1: 94896949-94927223 on Build GRCh38
Transcripts
NM_001286055.1 NM_001286056.1 NM_001839.4 XM_017000245.1

SNP Detail

SNP ID Chromosome Location
rs192297808 Chr.1: 94897702-94897702 on Build GRCh38
rs927039265 Chr.1: 94897611-94897611 on Build GRCh38
rs531084514 Chr.1: 94897449-94897449 on Build GRCh38
rs531558706 Chr.1: 94897427-94897427 on Build GRCh38
rs1020296745 Chr.1: 94897630-94897630 on Build GRCh38
rs571604039 Chr.1: 94897453-94897453 on Build GRCh38
rs1035950873 Chr.1: 94897426-94897426 on Build GRCh38
rs986329984 Chr.1: 94897346-94897346 on Build GRCh38
rs58217295 Chr.1: 94897393-94897394 on Build GRCh38
rs3789700 Chr.1: 94897498-94897498 on Build GRCh38
rs3182732 Chr.1: 94897350-94897350 on Build GRCh38
rs3789699 Chr.1: 94897445-94897445 on Build GRCh38
rs146582464 Chr.1: 94897482-94897483 on Build GRCh38
rs377001315 Chr.1: 94897371-94897372 on Build GRCh38
rs1006194015 Chr.1: 94897387-94897387 on Build GRCh38
rs998313879 Chr.1: 94897475-94897475 on Build GRCh38
rs375439592 Chr.1: 94897708-94897708 on Build GRCh38
rs1002423581 Chr.1: 94897552-94897552 on Build GRCh38
rs888713806 Chr.1: 94897370-94897370 on Build GRCh38
rs896370487 Chr.1: 94897599-94897599 on Build GRCh38
rs745741865 Chr.1: 94897373-94897376 on Build GRCh38
rs549074175 Chr.1: 94897416-94897416 on Build GRCh38
rs769477904 Chr.1: 94897372-94897376 on Build GRCh38
rs551288924 Chr.1: 94897517-94897517 on Build GRCh38
rs779933325 Chr.1: 94897374-94897376 on Build GRCh38
rs56226800 Chr.1: 94897391-94897394 on Build GRCh38
rs1000274083 Chr.1: 94897685-94897685 on Build GRCh38
rs1056409779 Chr.1: 94897594-94897594 on Build GRCh38
rs944388072 Chr.1: 94897371-94897371 on Build GRCh38
rs1006853142 Chr.1: 94897515-94897515 on Build GRCh38
rs960046754 Chr.1: 94897254-94897254 on Build GRCh38
rs1014236247 Chr.1: 94897280-94897280 on Build GRCh38
rs770375059 Chr.1: 94897310-94897312 on Build GRCh38
rs11540277 Chr.1: 94897322-94897322 on Build GRCh38
rs774950822 Chr.1: 94897267-94897267 on Build GRCh38
rs1014267359 Chr.1: 94897608-94897608 on Build GRCh38
rs912120713 Chr.1: 94897365-94897365 on Build GRCh38
rs913100911 Chr.1: 94897587-94897587 on Build GRCh38
rs979381205 Chr.1: 94897293-94897293 on Build GRCh38
rs1048777595 Chr.1: 94897632-94897632 on Build GRCh38
rs941650125 Chr.1: 94897686-94897686 on Build GRCh38
rs967449864 Chr.1: 94897292-94897292 on Build GRCh38
rs930408175 Chr.1: 94897447-94897447 on Build GRCh38
rs748610679 Chr.1: 94897719-94897719 on Build GRCh38
rs989123671 Chr.1: 94897571-94897571 on Build GRCh38
rs79385426 Chr.1: 94897370-94897375 on Build GRCh38
rs1033620439 Chr.1: 94897689-94897689 on Build GRCh38
rs188421793 Chr.1: 94897343-94897343 on Build GRCh38
rs954087790 Chr.1: 94897486-94897486 on Build GRCh38
rs547326662 Chr.1: 94897633-94897633 on Build GRCh38
rs1020388511 Chr.1: 94897285-94897285 on Build GRCh38
rs879041563 Chr.1: 94897372-94897374 on Build GRCh38
rs757600317 Chr.1: 94897606-94897606 on Build GRCh38
rs111824275 Chr.1: 94897326-94897326 on Build GRCh38
rs562537097 Chr.1: 94897423-94897423 on Build GRCh38
rs971636516 Chr.1: 94897607-94897607 on Build GRCh38
rs879075272 Chr.1: 94897374-94897374 on Build GRCh38

View Full Product Details