Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94897581-94898098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNN3
Entrez Gene ID
1266
Gene Name
calponin 3
Gene Aliases
-
UniGene
Hs.483454
Gene Chromosome Location
Chr.1: 94896949-94927223 on Build GRCh38
Transcripts
NM_001286055.1 NM_001286056.1 NM_001839.4 XM_017000245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1320827 c.978C>T p.G326G Chr.1: 94897754-94897754 on Build GRCh38
6064482 c.955G>C p.D319H Chr.1: 94897777-94897777 on Build GRCh38
1320826 c.926A>G p.H309R Chr.1: 94897806-94897806 on Build GRCh38
3866295 c.722C>T p.S241L Chr.1: 94898010-94898010 on Build GRCh38
4673119 c.894T>C p.S298S Chr.1: 94897838-94897838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192297808 Chr.1: 94897702-94897702 on Build GRCh38
rs927039265 Chr.1: 94897611-94897611 on Build GRCh38
rs138353466 Chr.1: 94897902-94897902 on Build GRCh38
rs1020296745 Chr.1: 94897630-94897630 on Build GRCh38
rs145115019 Chr.1: 94898009-94898009 on Build GRCh38
rs141237478 Chr.1: 94897863-94897863 on Build GRCh38
rs776310681 Chr.1: 94897763-94897763 on Build GRCh38
rs775431251 Chr.1: 94897801-94897801 on Build GRCh38
rs971305604 Chr.1: 94897946-94897946 on Build GRCh38
rs761661107 Chr.1: 94898027-94898027 on Build GRCh38
rs752685578 Chr.1: 94898072-94898072 on Build GRCh38
rs779415213 Chr.1: 94897987-94897987 on Build GRCh38
rs924800875 Chr.1: 94898021-94898021 on Build GRCh38
rs375439592 Chr.1: 94897708-94897708 on Build GRCh38
rs776859156 Chr.1: 94898044-94898044 on Build GRCh38
rs780781651 Chr.1: 94897906-94897906 on Build GRCh38
rs776824889 Chr.1: 94898025-94898025 on Build GRCh38
rs371131326 Chr.1: 94897854-94897854 on Build GRCh38
rs556260625 Chr.1: 94897769-94897769 on Build GRCh38
rs769288428 Chr.1: 94897963-94897963 on Build GRCh38
rs144459710 Chr.1: 94897818-94897818 on Build GRCh38
rs772000644 Chr.1: 94898038-94898038 on Build GRCh38
rs774452812 Chr.1: 94898013-94898013 on Build GRCh38
rs1000274083 Chr.1: 94897685-94897685 on Build GRCh38
rs774171877 Chr.1: 94897781-94897781 on Build GRCh38
rs774258664 Chr.1: 94897811-94897811 on Build GRCh38
rs536217522 Chr.1: 94897762-94897762 on Build GRCh38
rs116004008 Chr.1: 94897803-94897803 on Build GRCh38
rs763937439 Chr.1: 94897871-94897871 on Build GRCh38
rs768694788 Chr.1: 94898010-94898010 on Build GRCh38
rs889879185 Chr.1: 94897808-94897808 on Build GRCh38
rs147978281 Chr.1: 94897894-94897894 on Build GRCh38
rs10803 Chr.1: 94897723-94897723 on Build GRCh38
rs374373076 Chr.1: 94897883-94897883 on Build GRCh38
rs368719683 Chr.1: 94897853-94897853 on Build GRCh38
rs751895133 Chr.1: 94897896-94897896 on Build GRCh38
rs1014267359 Chr.1: 94897608-94897608 on Build GRCh38
rs1048777595 Chr.1: 94897632-94897632 on Build GRCh38
rs765565843 Chr.1: 94898071-94898071 on Build GRCh38
rs941650125 Chr.1: 94897686-94897686 on Build GRCh38
rs745510441 Chr.1: 94897911-94897911 on Build GRCh38
rs1020625813 Chr.1: 94897923-94897923 on Build GRCh38
rs766660179 Chr.1: 94897845-94897845 on Build GRCh38
rs747438073 Chr.1: 94897733-94897733 on Build GRCh38
rs748610679 Chr.1: 94897719-94897719 on Build GRCh38
rs759667748 Chr.1: 94898045-94898045 on Build GRCh38
rs937836322 Chr.1: 94897924-94897924 on Build GRCh38
rs1033620439 Chr.1: 94897689-94897689 on Build GRCh38
rs761716570 Chr.1: 94897802-94897802 on Build GRCh38
rs547326662 Chr.1: 94897633-94897633 on Build GRCh38
rs897359252 Chr.1: 94897736-94897736 on Build GRCh38
rs757600317 Chr.1: 94897606-94897606 on Build GRCh38
rs756638260 Chr.1: 94897938-94897938 on Build GRCh38
rs769146696 Chr.1: 94897811-94897812 on Build GRCh38
rs971636516 Chr.1: 94897607-94897607 on Build GRCh38
rs769217653 Chr.1: 94897782-94897782 on Build GRCh38
rs758733559 Chr.1: 94897728-94897728 on Build GRCh38

View Full Product Details