Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32370123-32370629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM8
Entrez Gene ID
152189
Gene Name
CKLF like MARVEL transmembrane domain containing 8
Gene Aliases
CKLFSF8, CKLFSF8-V2
UniGene
Hs.154986
Gene Chromosome Location
Chr.3: 32238679-32370325 on Build GRCh38
Transcripts
NM_001320308.1 NM_178868.4 XM_011533416.2 XM_017005779.1 XM_017005780.1

SNP Detail

SNP ID Chromosome Location
rs1000428842 Chr.3: 32370478-32370478 on Build GRCh38
rs116568275 Chr.3: 32370590-32370590 on Build GRCh38
rs1051890246 Chr.3: 32370432-32370432 on Build GRCh38
rs1033539420 Chr.3: 32370571-32370571 on Build GRCh38
rs9852142 Chr.3: 32370465-32370465 on Build GRCh38
rs971978217 Chr.3: 32370398-32370398 on Build GRCh38
rs950361695 Chr.3: 32370427-32370427 on Build GRCh38
rs80316770 Chr.3: 32370366-32370366 on Build GRCh38
rs973355974 Chr.3: 32370595-32370595 on Build GRCh38
rs943516579 Chr.3: 32370458-32370458 on Build GRCh38
rs550327632 Chr.3: 32370459-32370459 on Build GRCh38
rs200973281 Chr.3: 32370395-32370395 on Build GRCh38
rs759111217 Chr.3: 32370470-32370470 on Build GRCh38
rs146687397 Chr.3: 32370492-32370492 on Build GRCh38
rs17029321 Chr.3: 32370204-32370204 on Build GRCh38
rs1053861109 Chr.3: 32370479-32370479 on Build GRCh38
rs144680656 Chr.3: 32370372-32370372 on Build GRCh38
rs548304871 Chr.3: 32370543-32370543 on Build GRCh38
rs532210490 Chr.3: 32370440-32370440 on Build GRCh38
rs981115118 Chr.3: 32370402-32370402 on Build GRCh38
rs949826024 Chr.3: 32370162-32370162 on Build GRCh38
rs919027835 Chr.3: 32370406-32370406 on Build GRCh38
rs540561115 Chr.3: 32370381-32370381 on Build GRCh38
rs998263449 Chr.3: 32370252-32370252 on Build GRCh38
rs970554373 Chr.3: 32370247-32370247 on Build GRCh38
rs918887292 Chr.3: 32370168-32370168 on Build GRCh38
rs189947296 Chr.3: 32370556-32370556 on Build GRCh38
rs1051152120 Chr.3: 32370481-32370481 on Build GRCh38
rs1038849339 Chr.3: 32370244-32370244 on Build GRCh38
rs11710558 Chr.3: 32370430-32370430 on Build GRCh38
rs903160828 Chr.3: 32370289-32370289 on Build GRCh38
rs943312540 Chr.3: 32370235-32370235 on Build GRCh38
rs539895750 Chr.3: 32370319-32370319 on Build GRCh38
rs1009657433 Chr.3: 32370551-32370551 on Build GRCh38
rs943299089 Chr.3: 32370449-32370449 on Build GRCh38
rs184250215 Chr.3: 32370165-32370165 on Build GRCh38
rs998962290 Chr.3: 32370504-32370504 on Build GRCh38
rs928978994 Chr.3: 32370193-32370193 on Build GRCh38
rs1051189713 Chr.3: 32370269-32370269 on Build GRCh38
rs1002006807 Chr.3: 32370570-32370570 on Build GRCh38
rs980604740 Chr.3: 32370297-32370297 on Build GRCh38
rs760206845 Chr.3: 32370497-32370497 on Build GRCh38
rs765029874 Chr.3: 32370561-32370561 on Build GRCh38
rs987948678 Chr.3: 32370197-32370197 on Build GRCh38
rs770661240 Chr.3: 32370445-32370445 on Build GRCh38
rs11549648 Chr.3: 32370164-32370164 on Build GRCh38

View Full Product Details