Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32391580-32392080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM7
Entrez Gene ID
112616
Gene Name
CKLF like MARVEL transmembrane domain containing 7
Gene Aliases
CKLFSF7
UniGene
Hs.440494
Gene Chromosome Location
Chr.3: 32391671-32455528 on Build GRCh38
Transcripts
NM_138410.3 NM_181472.2 XM_011533319.2 XM_017005646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4953303 c.64G>A p.A22T Chr.3: 32391970-32391970 on Build GRCh38
5775940 c.81C>T p.P27P Chr.3: 32391987-32391987 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs11717165 Chr.3: 32391967-32391967 on Build GRCh38
rs149855043 Chr.3: 32391850-32391850 on Build GRCh38
rs916399022 Chr.3: 32391665-32391665 on Build GRCh38
rs901824399 Chr.3: 32391931-32391931 on Build GRCh38
rs959496779 Chr.3: 32391818-32391827 on Build GRCh38
rs969796545 Chr.3: 32391746-32391746 on Build GRCh38
rs1047990387 Chr.3: 32391641-32391641 on Build GRCh38
rs1011619423 Chr.3: 32391961-32391961 on Build GRCh38
rs963267058 Chr.3: 32392020-32392020 on Build GRCh38
rs529663350 Chr.3: 32392036-32392036 on Build GRCh38
rs1048890290 Chr.3: 32391877-32391877 on Build GRCh38
rs977650302 Chr.3: 32391839-32391839 on Build GRCh38
rs928332027 Chr.3: 32391753-32391753 on Build GRCh38
rs1049000700 Chr.3: 32391687-32391687 on Build GRCh38
rs901855948 Chr.3: 32391793-32391793 on Build GRCh38
rs991909712 Chr.3: 32391855-32391855 on Build GRCh38
rs574900000 Chr.3: 32391692-32391692 on Build GRCh38
rs960677086 Chr.3: 32391843-32391843 on Build GRCh38
rs891343328 Chr.3: 32391699-32391699 on Build GRCh38
rs572660378 Chr.3: 32391861-32391861 on Build GRCh38
rs921786394 Chr.3: 32392060-32392060 on Build GRCh38
rs932751968 Chr.3: 32391847-32391847 on Build GRCh38
rs778730266 Chr.3: 32391744-32391744 on Build GRCh38
rs546173159 Chr.3: 32391875-32391875 on Build GRCh38
rs1008676182 Chr.3: 32391851-32391851 on Build GRCh38
rs980139393 Chr.3: 32391747-32391747 on Build GRCh38
rs967563908 Chr.3: 32391835-32391835 on Build GRCh38
rs1021713839 Chr.3: 32391965-32391965 on Build GRCh38
rs956181779 Chr.3: 32391729-32391729 on Build GRCh38
rs1029513274 Chr.3: 32391801-32391801 on Build GRCh38
rs889238747 Chr.3: 32391957-32391957 on Build GRCh38
rs1011715335 Chr.3: 32391896-32391896 on Build GRCh38
rs939079335 Chr.3: 32391614-32391614 on Build GRCh38
rs909055982 Chr.3: 32391894-32391894 on Build GRCh38
rs564751013 Chr.3: 32391930-32391930 on Build GRCh38
rs755300397 Chr.3: 32391640-32391640 on Build GRCh38
rs999046235 Chr.3: 32391971-32391971 on Build GRCh38
rs780949461 Chr.3: 32392047-32392047 on Build GRCh38
rs947897701 Chr.3: 32391685-32391685 on Build GRCh38
rs11915090 Chr.3: 32391681-32391681 on Build GRCh38
rs907810460 Chr.3: 32391797-32391797 on Build GRCh38
rs1050523163 Chr.3: 32391949-32391949 on Build GRCh38
rs754680313 Chr.3: 32391984-32391984 on Build GRCh38
rs1036332039 Chr.3: 32391774-32391774 on Build GRCh38
rs923604249 Chr.3: 32391612-32391612 on Build GRCh38
rs1011787517 Chr.3: 32391740-32391740 on Build GRCh38
rs1032363665 Chr.3: 32391726-32391727 on Build GRCh38
rs953557624 Chr.3: 32391818-32391818 on Build GRCh38
rs960395041 Chr.3: 32392004-32392004 on Build GRCh38
rs887735143 Chr.3: 32391713-32391713 on Build GRCh38
rs562211318 Chr.3: 32391996-32391996 on Build GRCh38
rs941891687 Chr.3: 32391652-32391652 on Build GRCh38
rs997584716 Chr.3: 32391800-32391800 on Build GRCh38
rs200617949 Chr.3: 32391681-32391681 on Build GRCh38
rs997951524 Chr.3: 32391937-32391937 on Build GRCh38
rs3732846 Chr.3: 32391841-32391841 on Build GRCh38
rs940561578 Chr.3: 32391899-32391899 on Build GRCh38
rs1011732275 Chr.3: 32391828-32391828 on Build GRCh38
rs550295009 Chr.3: 32391973-32391973 on Build GRCh38
rs907717208 Chr.3: 32391648-32391648 on Build GRCh38
rs938618098 Chr.3: 32391619-32391619 on Build GRCh38
rs1021830005 Chr.3: 32391832-32391832 on Build GRCh38
rs1056636334 Chr.3: 32391623-32391623 on Build GRCh38

View Full Product Details