Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 36715002-36715499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK3
Entrez Gene ID
85443
Gene Name
doublecortin like kinase 3
Gene Aliases
CLR, DCAMKL3, DCDC3C, DCK3
UniGene
Hs.631907
Gene Chromosome Location
Chr.3: 36712422-36764206 on Build GRCh38
Transcripts
NM_033403.1 XM_011534167.2 XM_011534168.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1165941 c.1870C>T p.R624* Chr.3: 36715405-36715405 on Build GRCh38
5974741 c.1839C>T p.I613I Chr.3: 36715436-36715436 on Build GRCh38
5908273 c.1834T>A p.W612R Chr.3: 36715441-36715441 on Build GRCh38
4676539 c.1860T>C p.N620N Chr.3: 36715415-36715415 on Build GRCh38
6640907 c.1802G>A p.R601H Chr.3: 36715473-36715473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879463903 Chr.3: 36715242-36715242 on Build GRCh38
rs759518365 Chr.3: 36715062-36715062 on Build GRCh38
rs199761100 Chr.3: 36715330-36715330 on Build GRCh38
rs560482238 Chr.3: 36715378-36715378 on Build GRCh38
rs201391885 Chr.3: 36715371-36715371 on Build GRCh38
rs200230590 Chr.3: 36715335-36715336 on Build GRCh38
rs555976451 Chr.3: 36715200-36715200 on Build GRCh38
rs768586504 Chr.3: 36715389-36715389 on Build GRCh38
rs1047626438 Chr.3: 36715032-36715032 on Build GRCh38
rs867181277 Chr.3: 36715402-36715402 on Build GRCh38
rs906604705 Chr.3: 36715424-36715424 on Build GRCh38
rs749324049 Chr.3: 36715474-36715474 on Build GRCh38
rs533805237 Chr.3: 36715179-36715179 on Build GRCh38
rs200497284 Chr.3: 36715372-36715372 on Build GRCh38
rs767530225 Chr.3: 36715322-36715322 on Build GRCh38
rs951861808 Chr.3: 36715248-36715248 on Build GRCh38
rs775636532 Chr.3: 36715382-36715382 on Build GRCh38
rs565197644 Chr.3: 36715379-36715379 on Build GRCh38
rs958599616 Chr.3: 36715170-36715170 on Build GRCh38
rs372446395 Chr.3: 36715436-36715436 on Build GRCh38
rs138931496 Chr.3: 36715386-36715386 on Build GRCh38
rs750905282 Chr.3: 36715339-36715339 on Build GRCh38
rs201728869 Chr.3: 36715404-36715404 on Build GRCh38
rs368451641 Chr.3: 36715336-36715336 on Build GRCh38
rs200646257 Chr.3: 36715315-36715315 on Build GRCh38
rs1045653494 Chr.3: 36715320-36715320 on Build GRCh38
rs920375926 Chr.3: 36715220-36715220 on Build GRCh38
rs35704209 Chr.3: 36715376-36715376 on Build GRCh38
rs753143970 Chr.3: 36715433-36715433 on Build GRCh38
rs567500122 Chr.3: 36715201-36715201 on Build GRCh38
rs553268158 Chr.3: 36715244-36715244 on Build GRCh38
rs138738623 Chr.3: 36715250-36715250 on Build GRCh38
rs766029704 Chr.3: 36715385-36715385 on Build GRCh38
rs907819579 Chr.3: 36715036-36715036 on Build GRCh38
rs373746140 Chr.3: 36715388-36715388 on Build GRCh38
rs544783490 Chr.3: 36715055-36715055 on Build GRCh38
rs746565966 Chr.3: 36715171-36715172 on Build GRCh38
rs535957790 Chr.3: 36715123-36715123 on Build GRCh38
rs369306232 Chr.3: 36715435-36715435 on Build GRCh38
rs1008488735 Chr.3: 36715193-36715193 on Build GRCh38
rs1021423860 Chr.3: 36715107-36715107 on Build GRCh38
rs780591358 Chr.3: 36715365-36715365 on Build GRCh38
rs568945787 Chr.3: 36715117-36715117 on Build GRCh38
rs893279564 Chr.3: 36715102-36715102 on Build GRCh38
rs970840066 Chr.3: 36715152-36715152 on Build GRCh38
rs150186698 Chr.3: 36715092-36715092 on Build GRCh38
rs376002629 Chr.3: 36715415-36715415 on Build GRCh38
rs571294917 Chr.3: 36715045-36715045 on Build GRCh38
rs568759319 Chr.3: 36715411-36715411 on Build GRCh38
rs997087702 Chr.3: 36715048-36715048 on Build GRCh38
rs949970606 Chr.3: 36715303-36715303 on Build GRCh38
rs769774395 Chr.3: 36715302-36715302 on Build GRCh38
rs529825317 Chr.3: 36715420-36715420 on Build GRCh38
rs1010436655 Chr.3: 36715153-36715153 on Build GRCh38
rs758805251 Chr.3: 36715350-36715350 on Build GRCh38
rs200071698 Chr.3: 36715324-36715324 on Build GRCh38
rs368184483 Chr.3: 36715319-36715319 on Build GRCh38
rs149109120 Chr.3: 36715300-36715300 on Build GRCh38
rs752594417 Chr.3: 36715156-36715156 on Build GRCh38
rs893953124 Chr.3: 36715074-36715074 on Build GRCh38
rs756736128 Chr.3: 36715334-36715334 on Build GRCh38
rs535020225 Chr.3: 36715206-36715206 on Build GRCh38
rs1018986811 Chr.3: 36715202-36715202 on Build GRCh38
rs964429777 Chr.3: 36715205-36715205 on Build GRCh38
rs1054563305 Chr.3: 36715098-36715098 on Build GRCh38
rs1033716192 Chr.3: 36715160-36715160 on Build GRCh38
rs762587300 Chr.3: 36715403-36715403 on Build GRCh38
rs763835075 Chr.3: 36715332-36715332 on Build GRCh38
rs781330977 Chr.3: 36715367-36715367 on Build GRCh38
rs749881938 Chr.3: 36715400-36715400 on Build GRCh38
rs764985283 Chr.3: 36715398-36715398 on Build GRCh38
rs545388536 Chr.3: 36715272-36715272 on Build GRCh38
rs919857904 Chr.3: 36715217-36715217 on Build GRCh38
rs369590553 Chr.3: 36715366-36715366 on Build GRCh38
rs753547299 Chr.3: 36715333-36715333 on Build GRCh38
rs550263579 Chr.3: 36715095-36715095 on Build GRCh38
rs199999022 Chr.3: 36715393-36715393 on Build GRCh38
rs745309900 Chr.3: 36715405-36715405 on Build GRCh38
rs991429671 Chr.3: 36715213-36715213 on Build GRCh38

View Full Product Details