Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 36717871-36718371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK3
Entrez Gene ID
85443
Gene Name
doublecortin like kinase 3
Gene Aliases
CLR, DCAMKL3, DCDC3C, DCK3
UniGene
Hs.631907
Gene Chromosome Location
Chr.3: 36712422-36764206 on Build GRCh38
Transcripts
NM_033403.1 XM_011534167.2 XM_011534168.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6640904 c.1740C>A p.D580E Chr.3: 36718023-36718023 on Build GRCh38
4614917 c.1730delC p.P577fs*14 Chr.3: 36718033-36718033 on Build GRCh38
3915795 c.1748C>T p.S583F Chr.3: 36718015-36718015 on Build GRCh38
3915796 c.1684G>A p.E562K Chr.3: 36718079-36718079 on Build GRCh38
5420988 c.1683C>T p.D561D Chr.3: 36718080-36718080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004014654 Chr.3: 36718102-36718102 on Build GRCh38
rs567135963 Chr.3: 36718187-36718187 on Build GRCh38
rs762888851 Chr.3: 36718208-36718208 on Build GRCh38
rs370718609 Chr.3: 36718080-36718080 on Build GRCh38
rs570867955 Chr.3: 36718253-36718253 on Build GRCh38
rs922399826 Chr.3: 36717955-36717955 on Build GRCh38
rs771625779 Chr.3: 36718143-36718143 on Build GRCh38
rs977456558 Chr.3: 36717893-36717893 on Build GRCh38
rs923815529 Chr.3: 36717907-36717907 on Build GRCh38
rs761277661 Chr.3: 36718172-36718172 on Build GRCh38
rs116583441 Chr.3: 36718273-36718273 on Build GRCh38
rs567892553 Chr.3: 36718305-36718305 on Build GRCh38
rs1036960969 Chr.3: 36718092-36718092 on Build GRCh38
rs757523514 Chr.3: 36718056-36718056 on Build GRCh38
rs545357359 Chr.3: 36717901-36717901 on Build GRCh38
rs747827180 Chr.3: 36717969-36717969 on Build GRCh38
rs183276893 Chr.3: 36718314-36718314 on Build GRCh38
rs745542516 Chr.3: 36718227-36718227 on Build GRCh38
rs114769020 Chr.3: 36717959-36717959 on Build GRCh38
rs750016832 Chr.3: 36718231-36718231 on Build GRCh38
rs777628053 Chr.3: 36718220-36718220 on Build GRCh38
rs765607463 Chr.3: 36718066-36718066 on Build GRCh38
rs770555851 Chr.3: 36718129-36718129 on Build GRCh38
rs374711533 Chr.3: 36718103-36718103 on Build GRCh38
rs187053045 Chr.3: 36718127-36718127 on Build GRCh38
rs1022094172 Chr.3: 36718110-36718110 on Build GRCh38
rs745309621 Chr.3: 36718142-36718142 on Build GRCh38
rs991979780 Chr.3: 36717918-36717918 on Build GRCh38
rs999787143 Chr.3: 36718210-36718210 on Build GRCh38
rs530447954 Chr.3: 36718035-36718035 on Build GRCh38
rs756054216 Chr.3: 36718217-36718217 on Build GRCh38
rs754075081 Chr.3: 36718040-36718040 on Build GRCh38
rs972696748 Chr.3: 36718325-36718325 on Build GRCh38
rs370384908 Chr.3: 36718216-36718216 on Build GRCh38
rs370527915 Chr.3: 36718173-36718174 on Build GRCh38
rs759104295 Chr.3: 36718012-36718012 on Build GRCh38
rs762997349 Chr.3: 36717953-36717953 on Build GRCh38
rs999258794 Chr.3: 36718162-36718162 on Build GRCh38
rs373164557 Chr.3: 36718034-36718034 on Build GRCh38
rs764542912 Chr.3: 36718038-36718038 on Build GRCh38
rs761691293 Chr.3: 36718108-36718108 on Build GRCh38
rs13070703 Chr.3: 36718067-36718067 on Build GRCh38
rs200976050 Chr.3: 36718213-36718213 on Build GRCh38
rs770608885 Chr.3: 36717978-36717978 on Build GRCh38
rs186123765 Chr.3: 36717914-36717914 on Build GRCh38
rs769641958 Chr.3: 36717974-36717974 on Build GRCh38
rs754948651 Chr.3: 36717964-36717964 on Build GRCh38
rs775812169 Chr.3: 36718166-36718166 on Build GRCh38
rs774056976 Chr.3: 36718134-36718134 on Build GRCh38
rs908955150 Chr.3: 36718278-36718278 on Build GRCh38
rs538085193 Chr.3: 36718065-36718065 on Build GRCh38
rs976531531 Chr.3: 36717933-36717933 on Build GRCh38
rs947887942 Chr.3: 36717941-36717941 on Build GRCh38
rs765556595 Chr.3: 36718200-36718200 on Build GRCh38
rs79431295 Chr.3: 36718313-36718313 on Build GRCh38
rs758622367 Chr.3: 36718074-36718074 on Build GRCh38
rs956591786 Chr.3: 36718214-36718214 on Build GRCh38
rs774668478 Chr.3: 36717951-36717951 on Build GRCh38
rs371145113 Chr.3: 36717997-36717997 on Build GRCh38
rs552499575 Chr.3: 36718222-36718222 on Build GRCh38
rs781253529 Chr.3: 36717967-36717967 on Build GRCh38
rs777116260 Chr.3: 36718182-36718182 on Build GRCh38
rs764551561 Chr.3: 36718181-36718181 on Build GRCh38
rs755822985 Chr.3: 36718093-36718093 on Build GRCh38
rs750683514 Chr.3: 36718201-36718201 on Build GRCh38
rs182724793 Chr.3: 36717975-36717975 on Build GRCh38
rs867509081 Chr.3: 36718079-36718079 on Build GRCh38
rs909843324 Chr.3: 36718023-36718023 on Build GRCh38
rs767905416 Chr.3: 36718022-36718022 on Build GRCh38
rs190591044 Chr.3: 36717926-36717926 on Build GRCh38
rs191352502 Chr.3: 36718285-36718285 on Build GRCh38

View Full Product Details