Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 36737109-36737612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK3
Entrez Gene ID
85443
Gene Name
doublecortin like kinase 3
Gene Aliases
CLR, DCAMKL3, DCDC3C, DCK3
UniGene
Hs.631907
Gene Chromosome Location
Chr.3: 36712422-36764206 on Build GRCh38
Transcripts
NM_033403.1 XM_011534167.2 XM_011534168.2 XM_011534169.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3915797 c.1438C>T p.P480S Chr.3: 36737222-36737222 on Build GRCh38
7363855 c.1126C>G p.R376G Chr.3: 36737534-36737534 on Build GRCh38
3915798 c.1347C>T p.F449F Chr.3: 36737313-36737313 on Build GRCh38
7363852 c.1305G>T p.Q435H Chr.3: 36737355-36737355 on Build GRCh38
1131473 c.1263C>T p.Y421Y Chr.3: 36737397-36737397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750717043 Chr.3: 36737524-36737524 on Build GRCh38
rs775252078 Chr.3: 36737586-36737586 on Build GRCh38
rs1000608140 Chr.3: 36737264-36737264 on Build GRCh38
rs750172914 Chr.3: 36737185-36737185 on Build GRCh38
rs760755792 Chr.3: 36737289-36737289 on Build GRCh38
rs938211918 Chr.3: 36737369-36737369 on Build GRCh38
rs963904703 Chr.3: 36737196-36737196 on Build GRCh38
rs775498135 Chr.3: 36737285-36737285 on Build GRCh38
rs749809209 Chr.3: 36737372-36737372 on Build GRCh38
rs765209447 Chr.3: 36737181-36737181 on Build GRCh38
rs780812718 Chr.3: 36737231-36737231 on Build GRCh38
rs755754129 Chr.3: 36737361-36737361 on Build GRCh38
rs569208624 Chr.3: 36737531-36737531 on Build GRCh38
rs373605259 Chr.3: 36737396-36737396 on Build GRCh38
rs773962632 Chr.3: 36737434-36737434 on Build GRCh38
rs551077216 Chr.3: 36737385-36737385 on Build GRCh38
rs371444783 Chr.3: 36737213-36737213 on Build GRCh38
rs775612515 Chr.3: 36737532-36737532 on Build GRCh38
rs189132452 Chr.3: 36737337-36737337 on Build GRCh38
rs760240037 Chr.3: 36737481-36737481 on Build GRCh38
rs926280843 Chr.3: 36737365-36737365 on Build GRCh38
rs1037991536 Chr.3: 36737383-36737383 on Build GRCh38
rs747084479 Chr.3: 36737397-36737397 on Build GRCh38
rs757610264 Chr.3: 36737379-36737379 on Build GRCh38
rs528794239 Chr.3: 36737310-36737310 on Build GRCh38
rs895719609 Chr.3: 36737135-36737135 on Build GRCh38
rs774499578 Chr.3: 36737575-36737575 on Build GRCh38
rs768777296 Chr.3: 36737406-36737406 on Build GRCh38
rs778148473 Chr.3: 36737246-36737246 on Build GRCh38
rs539549840 Chr.3: 36737408-36737408 on Build GRCh38
rs570611854 Chr.3: 36737382-36737382 on Build GRCh38
rs868849468 Chr.3: 36737570-36737570 on Build GRCh38
rs34416671 Chr.3: 36737581-36737581 on Build GRCh38
rs377602149 Chr.3: 36737492-36737492 on Build GRCh38
rs746104762 Chr.3: 36737478-36737478 on Build GRCh38
rs762763566 Chr.3: 36737411-36737411 on Build GRCh38
rs913671079 Chr.3: 36737304-36737304 on Build GRCh38
rs376770547 Chr.3: 36737258-36737258 on Build GRCh38
rs753419825 Chr.3: 36737496-36737496 on Build GRCh38
rs370155703 Chr.3: 36737495-36737495 on Build GRCh38
rs368932640 Chr.3: 36737179-36737179 on Build GRCh38
rs765088898 Chr.3: 36737301-36737301 on Build GRCh38
rs975303042 Chr.3: 36737202-36737202 on Build GRCh38
rs759411546 Chr.3: 36737471-36737471 on Build GRCh38
rs116865058 Chr.3: 36737220-36737220 on Build GRCh38
rs748276161 Chr.3: 36737552-36737552 on Build GRCh38
rs770926877 Chr.3: 36737418-36737418 on Build GRCh38
rs779487369 Chr.3: 36737512-36737512 on Build GRCh38
rs751404842 Chr.3: 36737332-36737332 on Build GRCh38
rs779106841 Chr.3: 36737384-36737384 on Build GRCh38
rs373705108 Chr.3: 36737553-36737553 on Build GRCh38
rs202244952 Chr.3: 36737323-36737323 on Build GRCh38
rs780963223 Chr.3: 36737483-36737484 on Build GRCh38
rs200297338 Chr.3: 36737346-36737346 on Build GRCh38
rs747503381 Chr.3: 36737253-36737253 on Build GRCh38
rs929082284 Chr.3: 36737533-36737533 on Build GRCh38
rs778835744 Chr.3: 36737534-36737534 on Build GRCh38
rs758826501 Chr.3: 36737528-36737528 on Build GRCh38
rs373553939 Chr.3: 36737234-36737234 on Build GRCh38
rs865969776 Chr.3: 36737327-36737327 on Build GRCh38
rs561669121 Chr.3: 36737265-36737265 on Build GRCh38
rs369906307 Chr.3: 36737221-36737221 on Build GRCh38
rs759393087 Chr.3: 36737579-36737579 on Build GRCh38
rs757654846 Chr.3: 36737500-36737500 on Build GRCh38
rs369687833 Chr.3: 36737360-36737360 on Build GRCh38
rs761708545 Chr.3: 36737175-36737175 on Build GRCh38
rs756634489 Chr.3: 36737237-36737237 on Build GRCh38
rs758129013 Chr.3: 36737198-36737198 on Build GRCh38
rs773087239 Chr.3: 36737302-36737302 on Build GRCh38
rs1017982376 Chr.3: 36737523-36737523 on Build GRCh38
rs1009001120 Chr.3: 36737489-36737489 on Build GRCh38
rs185492799 Chr.3: 36737275-36737275 on Build GRCh38
rs772221898 Chr.3: 36737395-36737395 on Build GRCh38
rs553144855 Chr.3: 36737468-36737468 on Build GRCh38
rs56028200 Chr.3: 36737540-36737540 on Build GRCh38
rs778330901 Chr.3: 36737366-36737366 on Build GRCh38
rs772311550 Chr.3: 36737267-36737267 on Build GRCh38

View Full Product Details