Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 36985932-36986478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPM2AIP1
Entrez Gene ID
9852
Gene Name
EPM2A interacting protein 1
Gene Aliases
-
UniGene
Hs.28020
Gene Chromosome Location
Chr.3: 36985866-36993304 on Build GRCh38
Transcripts
NM_014805.3

SNP Detail

SNP ID Chromosome Location
rs931717162 Chr.3: 36986008-36986008 on Build GRCh38
rs980423748 Chr.3: 36986190-36986190 on Build GRCh38
rs551885474 Chr.3: 36985981-36985981 on Build GRCh38
rs1043169878 Chr.3: 36986318-36986318 on Build GRCh38
rs1055095 Chr.3: 36986442-36986442 on Build GRCh38
rs894331404 Chr.3: 36986253-36986253 on Build GRCh38
rs938625004 Chr.3: 36986088-36986088 on Build GRCh38
rs942267731 Chr.3: 36986301-36986301 on Build GRCh38
rs188481028 Chr.3: 36986167-36986167 on Build GRCh38
rs998865305 Chr.3: 36986327-36986327 on Build GRCh38
rs903144702 Chr.3: 36986323-36986323 on Build GRCh38
rs1028426318 Chr.3: 36986380-36986380 on Build GRCh38
rs908958683 Chr.3: 36986393-36986393 on Build GRCh38
rs879857949 Chr.3: 36986125-36986125 on Build GRCh38
rs567577220 Chr.3: 36986158-36986158 on Build GRCh38
rs555653342 Chr.3: 36986153-36986153 on Build GRCh38
rs1051791031 Chr.3: 36986430-36986430 on Build GRCh38
rs975544870 Chr.3: 36985982-36985982 on Build GRCh38
rs1006843990 Chr.3: 36986023-36986023 on Build GRCh38
rs755514690 Chr.3: 36986171-36986171 on Build GRCh38
rs552890719 Chr.3: 36986176-36986176 on Build GRCh38
rs947718116 Chr.3: 36986189-36986189 on Build GRCh38
rs1036475356 Chr.3: 36986061-36986061 on Build GRCh38
rs1011638471 Chr.3: 36986298-36986298 on Build GRCh38
rs967954221 Chr.3: 36986118-36986118 on Build GRCh38
rs553797838 Chr.3: 36986056-36986056 on Build GRCh38
rs997081148 Chr.3: 36986338-36986338 on Build GRCh38
rs917622463 Chr.3: 36986183-36986183 on Build GRCh38
rs569916535 Chr.3: 36985986-36985986 on Build GRCh38
rs897980756 Chr.3: 36986386-36986386 on Build GRCh38
rs150267463 Chr.3: 36986115-36986115 on Build GRCh38
rs765782836 Chr.3: 36986276-36986276 on Build GRCh38
rs371152727 Chr.3: 36986048-36986049 on Build GRCh38
rs962134356 Chr.3: 36986416-36986416 on Build GRCh38
rs909473246 Chr.3: 36986239-36986239 on Build GRCh38
rs765903760 Chr.3: 36986082-36986082 on Build GRCh38
rs570197667 Chr.3: 36986427-36986427 on Build GRCh38
rs889174278 Chr.3: 36986435-36986435 on Build GRCh38
rs965647673 Chr.3: 36986333-36986333 on Build GRCh38
rs867085536 Chr.3: 36986446-36986446 on Build GRCh38
rs952896596 Chr.3: 36986384-36986384 on Build GRCh38
rs907107440 Chr.3: 36986073-36986073 on Build GRCh38
rs1036518374 Chr.3: 36986330-36986330 on Build GRCh38
rs984525524 Chr.3: 36986028-36986028 on Build GRCh38
rs138907552 Chr.3: 36986428-36986428 on Build GRCh38
rs908855071 Chr.3: 36986042-36986042 on Build GRCh38

View Full Product Details