Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 36991858-36992368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPM2AIP1
Entrez Gene ID
9852
Gene Name
EPM2A interacting protein 1
Gene Aliases
-
UniGene
Hs.28020
Gene Chromosome Location
Chr.3: 36985866-36993304 on Build GRCh38
Transcripts
NM_014805.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7160736 c.1060C>G p.L354V Chr.3: 36992018-36992018 on Build GRCh38
1309063 c.905C>T p.S302F Chr.3: 36992173-36992173 on Build GRCh38
1617519 c.1162A>G p.M388V Chr.3: 36991916-36991916 on Build GRCh38
4681393 c.937G>A p.V313I Chr.3: 36992141-36992141 on Build GRCh38
1422562 c.943C>T p.R315* Chr.3: 36992135-36992135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745563339 Chr.3: 36992155-36992155 on Build GRCh38
rs4647201 Chr.3: 36992000-36992000 on Build GRCh38
rs781762112 Chr.3: 36992313-36992313 on Build GRCh38
rs1043950209 Chr.3: 36992076-36992076 on Build GRCh38
rs752692722 Chr.3: 36991995-36991995 on Build GRCh38
rs367573129 Chr.3: 36992232-36992232 on Build GRCh38
rs771097590 Chr.3: 36992329-36992329 on Build GRCh38
rs777283517 Chr.3: 36992211-36992211 on Build GRCh38
rs774557411 Chr.3: 36992330-36992330 on Build GRCh38
rs754083403 Chr.3: 36991954-36991954 on Build GRCh38
rs771175042 Chr.3: 36992097-36992097 on Build GRCh38
rs759553941 Chr.3: 36992334-36992334 on Build GRCh38
rs770166494 Chr.3: 36992323-36992323 on Build GRCh38
rs779886091 Chr.3: 36992298-36992298 on Build GRCh38
rs748674005 Chr.3: 36992321-36992321 on Build GRCh38
rs4647202 Chr.3: 36992081-36992081 on Build GRCh38
rs933664832 Chr.3: 36992026-36992026 on Build GRCh38
rs750770992 Chr.3: 36991953-36991953 on Build GRCh38
rs765005616 Chr.3: 36992271-36992271 on Build GRCh38
rs773481069 Chr.3: 36992226-36992226 on Build GRCh38
rs781130377 Chr.3: 36991977-36991977 on Build GRCh38
rs375379887 Chr.3: 36992221-36992221 on Build GRCh38
rs773323023 Chr.3: 36992089-36992089 on Build GRCh38
rs757940682 Chr.3: 36992291-36992291 on Build GRCh38
rs765401720 Chr.3: 36991960-36991960 on Build GRCh38
rs769121875 Chr.3: 36992029-36992029 on Build GRCh38
rs775865024 Chr.3: 36991931-36991931 on Build GRCh38
rs779761478 Chr.3: 36992178-36992178 on Build GRCh38
rs756881536 Chr.3: 36992009-36992009 on Build GRCh38
rs755900593 Chr.3: 36991999-36991999 on Build GRCh38
rs757061677 Chr.3: 36992269-36992269 on Build GRCh38
rs762116131 Chr.3: 36992071-36992071 on Build GRCh38
rs746542345 Chr.3: 36992202-36992202 on Build GRCh38
rs753586982 Chr.3: 36992115-36992115 on Build GRCh38
rs776803768 Chr.3: 36992228-36992228 on Build GRCh38
rs983941435 Chr.3: 36991950-36991950 on Build GRCh38
rs776125272 Chr.3: 36992024-36992024 on Build GRCh38
rs750059401 Chr.3: 36992276-36992276 on Build GRCh38
rs769246599 Chr.3: 36992206-36992206 on Build GRCh38
rs761061915 Chr.3: 36991934-36991934 on Build GRCh38
rs914740335 Chr.3: 36992154-36992154 on Build GRCh38
rs766420979 Chr.3: 36992234-36992234 on Build GRCh38
rs757920762 Chr.3: 36992171-36992171 on Build GRCh38
rs765434479 Chr.3: 36992083-36992083 on Build GRCh38
rs760982762 Chr.3: 36992019-36992019 on Build GRCh38
rs572137294 Chr.3: 36991903-36991903 on Build GRCh38
rs757007062 Chr.3: 36992131-36992131 on Build GRCh38
rs546128727 Chr.3: 36991919-36991919 on Build GRCh38
rs752602885 Chr.3: 36992092-36992092 on Build GRCh38
rs766437860 Chr.3: 36992091-36992091 on Build GRCh38
rs554186217 Chr.3: 36992162-36992162 on Build GRCh38
rs946938072 Chr.3: 36992046-36992046 on Build GRCh38
rs747584787 Chr.3: 36992027-36992027 on Build GRCh38
rs890066895 Chr.3: 36992035-36992035 on Build GRCh38
rs778457036 Chr.3: 36992016-36992016 on Build GRCh38
rs185831966 Chr.3: 36992326-36992326 on Build GRCh38
rs770313148 Chr.3: 36991912-36991912 on Build GRCh38
rs967801643 Chr.3: 36992156-36992156 on Build GRCh38
rs762176158 Chr.3: 36991955-36991955 on Build GRCh38
rs764609675 Chr.3: 36991951-36991951 on Build GRCh38
rs923574927 Chr.3: 36992253-36992253 on Build GRCh38
rs371339694 Chr.3: 36992266-36992266 on Build GRCh38
rs4647200 Chr.3: 36991971-36991971 on Build GRCh38
rs772590334 Chr.3: 36991921-36991921 on Build GRCh38
rs748490958 Chr.3: 36992216-36992216 on Build GRCh38
rs763078547 Chr.3: 36992090-36992090 on Build GRCh38
rs777089380 Chr.3: 36992066-36992066 on Build GRCh38
rs543497247 Chr.3: 36992110-36992110 on Build GRCh38
rs774363662 Chr.3: 36992247-36992247 on Build GRCh38
rs763948684 Chr.3: 36992263-36992263 on Build GRCh38
rs528861282 Chr.3: 36992300-36992300 on Build GRCh38
rs750476497 Chr.3: 36991961-36991961 on Build GRCh38
rs754549421 Chr.3: 36992307-36992307 on Build GRCh38
rs759639726 Chr.3: 36992249-36992249 on Build GRCh38
rs778569984 Chr.3: 36992147-36992147 on Build GRCh38
rs756091009 Chr.3: 36992109-36992109 on Build GRCh38

View Full Product Details