Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38131839-38132349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAA1
Entrez Gene ID
30
Gene Name
acetyl-CoA acyltransferase 1
Gene Aliases
ACAA, PTHIO, THIO
UniGene
Hs.643487
Gene Chromosome Location
Chr.3: 38122710-38137242 on Build GRCh38
Transcripts
NM_001130410.1 NM_001607.3 NR_024024.1 XM_006713122.1 XM_006713123.2 XM_011533650.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7277476 c.331C>A p.P111T Chr.3: 38131998-38131998 on Build GRCh38
1181476 c.371C>T p.S124L Chr.3: 38131958-38131958 on Build GRCh38
5864421 c.350C>T p.S117F Chr.3: 38131979-38131979 on Build GRCh38
6591443 c.402A>C p.A134A Chr.3: 38131927-38131927 on Build GRCh38
6591442 c.339T>C p.T113T Chr.3: 38131990-38131990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769168524 Chr.3: 38131888-38131888 on Build GRCh38
rs747650775 Chr.3: 38131883-38131883 on Build GRCh38
rs549863064 Chr.3: 38132203-38132203 on Build GRCh38
rs956843885 Chr.3: 38132302-38132302 on Build GRCh38
rs1057408 Chr.3: 38131960-38131960 on Build GRCh38
rs751417560 Chr.3: 38132021-38132021 on Build GRCh38
rs201538385 Chr.3: 38131902-38131902 on Build GRCh38
rs765154469 Chr.3: 38132151-38132151 on Build GRCh38
rs767214055 Chr.3: 38132036-38132036 on Build GRCh38
rs1006095060 Chr.3: 38131864-38131864 on Build GRCh38
rs150756727 Chr.3: 38131954-38131954 on Build GRCh38
rs372816021 Chr.3: 38132042-38132042 on Build GRCh38
rs1045057194 Chr.3: 38132074-38132074 on Build GRCh38
rs775839933 Chr.3: 38131990-38131990 on Build GRCh38
rs375526157 Chr.3: 38131929-38131929 on Build GRCh38
rs942650320 Chr.3: 38132275-38132275 on Build GRCh38
rs374663027 Chr.3: 38131919-38131919 on Build GRCh38
rs968441969 Chr.3: 38132227-38132227 on Build GRCh38
rs765146534 Chr.3: 38131986-38131987 on Build GRCh38
rs758262605 Chr.3: 38131931-38131931 on Build GRCh38
rs764064870 Chr.3: 38131925-38131925 on Build GRCh38
rs751301935 Chr.3: 38131946-38131946 on Build GRCh38
rs369207321 Chr.3: 38132113-38132113 on Build GRCh38
rs935472133 Chr.3: 38132180-38132180 on Build GRCh38
rs924079349 Chr.3: 38131994-38131994 on Build GRCh38
rs527288624 Chr.3: 38132073-38132073 on Build GRCh38
rs866657453 Chr.3: 38131866-38131866 on Build GRCh38
rs143507571 Chr.3: 38132002-38132002 on Build GRCh38
rs544641735 Chr.3: 38132273-38132273 on Build GRCh38
rs776296974 Chr.3: 38131887-38131888 on Build GRCh38
rs562195228 Chr.3: 38132128-38132128 on Build GRCh38
rs925388903 Chr.3: 38131862-38131862 on Build GRCh38
rs1014755952 Chr.3: 38131865-38131865 on Build GRCh38
rs771428138 Chr.3: 38131894-38131894 on Build GRCh38
rs898298981 Chr.3: 38131970-38131970 on Build GRCh38
rs767738958 Chr.3: 38131913-38131913 on Build GRCh38
rs772301831 Chr.3: 38131979-38131979 on Build GRCh38
rs746285369 Chr.3: 38131969-38131969 on Build GRCh38
rs1057408 Chr.3: 38131960-38131960 on Build GRCh38
rs60935636 Chr.3: 38132322-38132322 on Build GRCh38
rs752402239 Chr.3: 38132039-38132039 on Build GRCh38
rs942057794 Chr.3: 38132121-38132121 on Build GRCh38
rs538629220 Chr.3: 38132033-38132033 on Build GRCh38
rs752282975 Chr.3: 38131953-38131953 on Build GRCh38
rs773916988 Chr.3: 38131889-38131889 on Build GRCh38
rs923345395 Chr.3: 38132241-38132241 on Build GRCh38
rs1038664629 Chr.3: 38132316-38132316 on Build GRCh38
rs997455215 Chr.3: 38132181-38132181 on Build GRCh38
rs547119058 Chr.3: 38132093-38132093 on Build GRCh38
rs776632031 Chr.3: 38131997-38131997 on Build GRCh38
rs577888328 Chr.3: 38132102-38132102 on Build GRCh38
rs775542395 Chr.3: 38131914-38131914 on Build GRCh38
rs887893000 Chr.3: 38132091-38132091 on Build GRCh38
rs768261472 Chr.3: 38131883-38131884 on Build GRCh38
rs780586143 Chr.3: 38131877-38131877 on Build GRCh38
rs761614794 Chr.3: 38131923-38131924 on Build GRCh38
rs145935424 Chr.3: 38131965-38131965 on Build GRCh38
rs754614303 Chr.3: 38131947-38131947 on Build GRCh38
rs750371562 Chr.3: 38132036-38132036 on Build GRCh38
rs960365926 Chr.3: 38132110-38132110 on Build GRCh38
rs766261453 Chr.3: 38132009-38132009 on Build GRCh38
rs77229058 Chr.3: 38131893-38131893 on Build GRCh38
rs760346891 Chr.3: 38131879-38131880 on Build GRCh38
rs1050402807 Chr.3: 38132271-38132271 on Build GRCh38
rs539131243 Chr.3: 38132164-38132164 on Build GRCh38
rs149465180 Chr.3: 38131957-38131957 on Build GRCh38
rs889025465 Chr.3: 38132199-38132199 on Build GRCh38
rs184653956 Chr.3: 38132047-38132047 on Build GRCh38
rs889700753 Chr.3: 38131899-38131899 on Build GRCh38
rs768725966 Chr.3: 38131996-38131996 on Build GRCh38
rs925495415 Chr.3: 38132172-38132172 on Build GRCh38
rs780950573 Chr.3: 38131948-38131948 on Build GRCh38
rs1021288809 Chr.3: 38132218-38132218 on Build GRCh38
rs569850110 Chr.3: 38132220-38132220 on Build GRCh38
rs766171603 Chr.3: 38131935-38131935 on Build GRCh38
rs953132629 Chr.3: 38131938-38131938 on Build GRCh38
rs753361109 Chr.3: 38132050-38132050 on Build GRCh38
rs1021816538 Chr.3: 38131951-38131951 on Build GRCh38
rs563704732 Chr.3: 38131993-38131993 on Build GRCh38
rs2239621 Chr.3: 38132242-38132242 on Build GRCh38
rs369897061 Chr.3: 38131958-38131958 on Build GRCh38
rs1030691676 Chr.3: 38132194-38132194 on Build GRCh38

View Full Product Details