Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42265983-42266448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCK
Entrez Gene ID
885
Gene Name
cholecystokinin
Gene Aliases
-
UniGene
Hs.458426
Gene Chromosome Location
Chr.3: 42257824-42266195 on Build GRCh38
Transcripts
NM_000729.5

SNP Detail

SNP ID Chromosome Location
rs62259576 Chr.3: 42266332-42266332 on Build GRCh38
rs765176131 Chr.3: 42266303-42266303 on Build GRCh38
rs536986539 Chr.3: 42266178-42266178 on Build GRCh38
rs1024806874 Chr.3: 42266380-42266380 on Build GRCh38
rs983072079 Chr.3: 42266141-42266141 on Build GRCh38
rs541027801 Chr.3: 42266426-42266426 on Build GRCh38
rs989751062 Chr.3: 42266264-42266264 on Build GRCh38
rs994823475 Chr.3: 42266135-42266135 on Build GRCh38
rs1042910824 Chr.3: 42266064-42266064 on Build GRCh38
rs899427600 Chr.3: 42266244-42266244 on Build GRCh38
rs534292811 Chr.3: 42266052-42266052 on Build GRCh38
rs886434684 Chr.3: 42266420-42266420 on Build GRCh38
rs112600163 Chr.3: 42266164-42266164 on Build GRCh38
rs1033512291 Chr.3: 42266022-42266022 on Build GRCh38
rs890637235 Chr.3: 42266080-42266080 on Build GRCh38
rs894453515 Chr.3: 42266361-42266361 on Build GRCh38
rs958794831 Chr.3: 42266024-42266024 on Build GRCh38
rs931724345 Chr.3: 42266236-42266236 on Build GRCh38
rs369284586 Chr.3: 42266194-42266194 on Build GRCh38
rs1015846632 Chr.3: 42266157-42266157 on Build GRCh38
rs986273496 Chr.3: 42266246-42266246 on Build GRCh38
rs957297330 Chr.3: 42266254-42266254 on Build GRCh38
rs1009513273 Chr.3: 42266091-42266091 on Build GRCh38
rs747912043 Chr.3: 42266033-42266033 on Build GRCh38
rs13069836 Chr.3: 42266417-42266417 on Build GRCh38
rs944872995 Chr.3: 42266366-42266366 on Build GRCh38
rs912955427 Chr.3: 42266021-42266021 on Build GRCh38
rs900777485 Chr.3: 42266131-42266131 on Build GRCh38
rs1041622196 Chr.3: 42266318-42266318 on Build GRCh38
rs1024846050 Chr.3: 42266196-42266196 on Build GRCh38
rs971254695 Chr.3: 42266240-42266240 on Build GRCh38
rs1000116399 Chr.3: 42266000-42266000 on Build GRCh38
rs188347021 Chr.3: 42266410-42266410 on Build GRCh38
rs1013025567 Chr.3: 42266375-42266375 on Build GRCh38
rs903066782 Chr.3: 42266331-42266331 on Build GRCh38
rs1013514354 Chr.3: 42266050-42266050 on Build GRCh38
rs542913323 Chr.3: 42266007-42266007 on Build GRCh38
rs945818442 Chr.3: 42266028-42266028 on Build GRCh38
rs557735742 Chr.3: 42266087-42266087 on Build GRCh38
rs114501851 Chr.3: 42266099-42266099 on Build GRCh38
rs944533239 Chr.3: 42266317-42266317 on Build GRCh38
rs140722016 Chr.3: 42266341-42266346 on Build GRCh38
rs912901493 Chr.3: 42266270-42266270 on Build GRCh38
rs556733642 Chr.3: 42266287-42266287 on Build GRCh38
rs528663787 Chr.3: 42266190-42266190 on Build GRCh38
rs866598283 Chr.3: 42266235-42266235 on Build GRCh38
rs866423981 Chr.3: 42266212-42266212 on Build GRCh38
rs1036456189 Chr.3: 42266243-42266243 on Build GRCh38
rs927201359 Chr.3: 42266250-42266250 on Build GRCh38
rs912339781 Chr.3: 42266347-42266347 on Build GRCh38
rs542292179 Chr.3: 42266344-42266344 on Build GRCh38
rs555848217 Chr.3: 42266245-42266245 on Build GRCh38
rs1027668164 Chr.3: 42266136-42266136 on Build GRCh38

View Full Product Details