Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 43702137-43702641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD5
Entrez Gene ID
51099
Gene Name
abhydrolase domain containing 5
Gene Aliases
CDS, CGI58, IECN2, NCIE2
UniGene
Hs.19385
Gene Chromosome Location
Chr.3: 43690883-43722725 on Build GRCh38
Transcripts
NM_016006.4 XM_011533779.2 XM_011533780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4408237 c.211A>C p.N71H Chr.3: 43702292-43702292 on Build GRCh38
4414650 c.480T>C p.A160A Chr.3: 43702561-43702561 on Build GRCh38
7242733 c.312C>G p.V104V Chr.3: 43702393-43702393 on Build GRCh38
4492689 c.401C>T p.S134F Chr.3: 43702482-43702482 on Build GRCh38
7353718 c.473T>A p.L158* Chr.3: 43702554-43702554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536642005 Chr.3: 43702405-43702405 on Build GRCh38
rs148750021 Chr.3: 43702381-43702381 on Build GRCh38
rs769135188 Chr.3: 43702295-43702295 on Build GRCh38
rs141535425 Chr.3: 43702521-43702521 on Build GRCh38
rs962581009 Chr.3: 43702492-43702492 on Build GRCh38
rs773516287 Chr.3: 43702574-43702574 on Build GRCh38
rs1037633003 Chr.3: 43702461-43702461 on Build GRCh38
rs772479012 Chr.3: 43702318-43702318 on Build GRCh38
rs573378765 Chr.3: 43702433-43702433 on Build GRCh38
rs767783380 Chr.3: 43702203-43702203 on Build GRCh38
rs779669249 Chr.3: 43702276-43702276 on Build GRCh38
rs145548259 Chr.3: 43702326-43702326 on Build GRCh38
rs752495601 Chr.3: 43702513-43702513 on Build GRCh38
rs773922601 Chr.3: 43702596-43702596 on Build GRCh38
rs377223020 Chr.3: 43702570-43702570 on Build GRCh38
rs1041234550 Chr.3: 43702389-43702389 on Build GRCh38
rs749810227 Chr.3: 43702590-43702590 on Build GRCh38
rs537368660 Chr.3: 43702339-43702339 on Build GRCh38
rs373933908 Chr.3: 43702529-43702529 on Build GRCh38
rs749426567 Chr.3: 43702193-43702193 on Build GRCh38
rs755220242 Chr.3: 43702558-43702558 on Build GRCh38
rs776529778 Chr.3: 43702181-43702181 on Build GRCh38
rs201137660 Chr.3: 43702303-43702303 on Build GRCh38
rs780866502 Chr.3: 43702409-43702409 on Build GRCh38
rs1042260491 Chr.3: 43702343-43702343 on Build GRCh38
rs764214615 Chr.3: 43702432-43702433 on Build GRCh38
rs373148191 Chr.3: 43702477-43702477 on Build GRCh38
rs758868179 Chr.3: 43702262-43702262 on Build GRCh38
rs377718797 Chr.3: 43702309-43702309 on Build GRCh38
rs375199350 Chr.3: 43702173-43702173 on Build GRCh38
rs200927822 Chr.3: 43702282-43702282 on Build GRCh38
rs751016701 Chr.3: 43702507-43702507 on Build GRCh38
rs752473449 Chr.3: 43702421-43702421 on Build GRCh38
rs777005988 Chr.3: 43702341-43702341 on Build GRCh38
rs371138384 Chr.3: 43702443-43702443 on Build GRCh38
rs147218153 Chr.3: 43702302-43702302 on Build GRCh38
rs753853386 Chr.3: 43702525-43702525 on Build GRCh38
rs2302349 Chr.3: 43702296-43702296 on Build GRCh38
rs141184431 Chr.3: 43702226-43702226 on Build GRCh38
rs775175189 Chr.3: 43702613-43702613 on Build GRCh38
rs201554978 Chr.3: 43702240-43702240 on Build GRCh38
rs115209685 Chr.3: 43702283-43702283 on Build GRCh38
rs758048276 Chr.3: 43702274-43702274 on Build GRCh38
rs777375929 Chr.3: 43702467-43702467 on Build GRCh38
rs776862217 Chr.3: 43702456-43702456 on Build GRCh38
rs1038323436 Chr.3: 43702280-43702280 on Build GRCh38
rs757742884 Chr.3: 43702248-43702248 on Build GRCh38
rs776061830 Chr.3: 43702212-43702212 on Build GRCh38
rs749965827 Chr.3: 43702363-43702363 on Build GRCh38
rs573986214 Chr.3: 43702371-43702371 on Build GRCh38
rs771365110 Chr.3: 43702595-43702595 on Build GRCh38
rs146635810 Chr.3: 43702453-43702453 on Build GRCh38
rs1050448357 Chr.3: 43702291-43702291 on Build GRCh38
rs566224501 Chr.3: 43702233-43702233 on Build GRCh38
rs745991593 Chr.3: 43702431-43702431 on Build GRCh38
rs781461249 Chr.3: 43702569-43702569 on Build GRCh38
rs773740366 Chr.3: 43702479-43702479 on Build GRCh38
rs777126097 Chr.3: 43702453-43702455 on Build GRCh38
rs777749420 Chr.3: 43702426-43702426 on Build GRCh38
rs371050752 Chr.3: 43702166-43702166 on Build GRCh38
rs765897182 Chr.3: 43702484-43702484 on Build GRCh38
rs777609553 Chr.3: 43702523-43702523 on Build GRCh38
rs761158107 Chr.3: 43702334-43702334 on Build GRCh38
rs747255945 Chr.3: 43702435-43702435 on Build GRCh38
rs369683357 Chr.3: 43702347-43702347 on Build GRCh38
rs376683599 Chr.3: 43702528-43702528 on Build GRCh38
rs969130040 Chr.3: 43702583-43702583 on Build GRCh38
rs781001493 Chr.3: 43702287-43702287 on Build GRCh38
rs764742894 Chr.3: 43702232-43702232 on Build GRCh38
rs767121885 Chr.3: 43702604-43702604 on Build GRCh38
rs749889127 Chr.3: 43702270-43702270 on Build GRCh38
rs978995454 Chr.3: 43702357-43702357 on Build GRCh38
rs1000849111 Chr.3: 43702395-43702395 on Build GRCh38
rs762239030 Chr.3: 43702344-43702344 on Build GRCh38
rs767197048 Chr.3: 43702511-43702511 on Build GRCh38
rs200595589 Chr.3: 43702480-43702480 on Build GRCh38
rs979899167 Chr.3: 43702199-43702199 on Build GRCh38
rs911219163 Chr.3: 43702609-43702609 on Build GRCh38
rs759694389 Chr.3: 43702202-43702202 on Build GRCh38
rs557609500 Chr.3: 43702348-43702348 on Build GRCh38
rs747171625 Chr.3: 43702188-43702189 on Build GRCh38
rs754576816 Chr.3: 43702408-43702408 on Build GRCh38
rs759036867 Chr.3: 43702603-43702603 on Build GRCh38
rs374480855 Chr.3: 43702508-43702508 on Build GRCh38
rs770227805 Chr.3: 43702571-43702571 on Build GRCh38
rs549346750 Chr.3: 43702295-43702295 on Build GRCh38
rs774444076 Chr.3: 43702198-43702198 on Build GRCh38
rs1024005356 Chr.3: 43702449-43702449 on Build GRCh38
rs748850518 Chr.3: 43702522-43702522 on Build GRCh38
rs762212861 Chr.3: 43702562-43702563 on Build GRCh38
rs141383973 Chr.3: 43702530-43702530 on Build GRCh38
rs28939077 Chr.3: 43702470-43702470 on Build GRCh38
rs367883757 Chr.3: 43702197-43702197 on Build GRCh38
rs764673952 Chr.3: 43702252-43702252 on Build GRCh38
rs780238239 Chr.3: 43702434-43702434 on Build GRCh38
rs148743497 Chr.3: 43702422-43702422 on Build GRCh38
rs1033263791 Chr.3: 43702417-43702417 on Build GRCh38
rs747018565 Chr.3: 43702555-43702555 on Build GRCh38

View Full Product Details