Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 43720074-43720576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD5
Entrez Gene ID
51099
Gene Name
abhydrolase domain containing 5
Gene Aliases
CDS, CGI58, IECN2, NCIE2
UniGene
Hs.19385
Gene Chromosome Location
Chr.3: 43690883-43722725 on Build GRCh38
Transcripts
NM_016006.4 XM_011533779.2 XM_011533780.1

SNP Detail

SNP ID Chromosome Location
rs139091067 Chr.3: 43720261-43720261 on Build GRCh38
rs767379534 Chr.3: 43720496-43720496 on Build GRCh38
rs1046062726 Chr.3: 43720529-43720529 on Build GRCh38
rs149517586 Chr.3: 43720325-43720325 on Build GRCh38
rs576143123 Chr.3: 43720330-43720330 on Build GRCh38
rs541779619 Chr.3: 43720343-43720343 on Build GRCh38
rs534344658 Chr.3: 43720148-43720148 on Build GRCh38
rs12488350 Chr.3: 43720366-43720366 on Build GRCh38
rs746473026 Chr.3: 43720122-43720122 on Build GRCh38
rs1004642575 Chr.3: 43720339-43720339 on Build GRCh38
rs12491747 Chr.3: 43720145-43720145 on Build GRCh38
rs886058494 Chr.3: 43720392-43720392 on Build GRCh38
rs117110703 Chr.3: 43720194-43720194 on Build GRCh38
rs571429944 Chr.3: 43720391-43720391 on Build GRCh38
rs1052624004 Chr.3: 43720128-43720128 on Build GRCh38
rs12488392 Chr.3: 43720395-43720395 on Build GRCh38
rs945467834 Chr.3: 43720188-43720188 on Build GRCh38
rs893940566 Chr.3: 43720144-43720144 on Build GRCh38
rs949663639 Chr.3: 43720521-43720521 on Build GRCh38
rs144021874 Chr.3: 43720489-43720489 on Build GRCh38
rs942417643 Chr.3: 43720500-43720500 on Build GRCh38
rs927932687 Chr.3: 43720367-43720367 on Build GRCh38
rs969858616 Chr.3: 43720344-43720344 on Build GRCh38
rs1016075585 Chr.3: 43720364-43720364 on Build GRCh38
rs34011416 Chr.3: 43720290-43720291 on Build GRCh38
rs1025550986 Chr.3: 43720522-43720522 on Build GRCh38
rs948113438 Chr.3: 43720115-43720115 on Build GRCh38
rs1019686280 Chr.3: 43720240-43720240 on Build GRCh38
rs954173528 Chr.3: 43720135-43720135 on Build GRCh38
rs895426323 Chr.3: 43720305-43720305 on Build GRCh38
rs563435148 Chr.3: 43720100-43720100 on Build GRCh38
rs912646161 Chr.3: 43720160-43720160 on Build GRCh38
rs1051017019 Chr.3: 43720499-43720499 on Build GRCh38
rs1046826697 Chr.3: 43720121-43720121 on Build GRCh38
rs886150025 Chr.3: 43720333-43720333 on Build GRCh38
rs886119105 Chr.3: 43720535-43720535 on Build GRCh38
rs901101532 Chr.3: 43720260-43720260 on Build GRCh38
rs113347419 Chr.3: 43720289-43720289 on Build GRCh38
rs954690260 Chr.3: 43720417-43720417 on Build GRCh38
rs533137321 Chr.3: 43720485-43720485 on Build GRCh38
rs368470132 Chr.3: 43720165-43720165 on Build GRCh38
rs765007173 Chr.3: 43720226-43720226 on Build GRCh38
rs878912236 Chr.3: 43720552-43720552 on Build GRCh38
rs996053690 Chr.3: 43720382-43720382 on Build GRCh38
rs34048061 Chr.3: 43720288-43720288 on Build GRCh38
rs886058495 Chr.3: 43720427-43720427 on Build GRCh38
rs564255323 Chr.3: 43720475-43720475 on Build GRCh38
rs1019583326 Chr.3: 43720427-43720427 on Build GRCh38
rs916814705 Chr.3: 43720501-43720501 on Build GRCh38
rs900806449 Chr.3: 43720508-43720508 on Build GRCh38
rs541306296 Chr.3: 43720466-43720466 on Build GRCh38
rs534505586 Chr.3: 43720207-43720207 on Build GRCh38
rs746232107 Chr.3: 43720123-43720123 on Build GRCh38
rs1013660068 Chr.3: 43720313-43720313 on Build GRCh38
rs936819857 Chr.3: 43720488-43720488 on Build GRCh38
rs987168595 Chr.3: 43720419-43720419 on Build GRCh38
rs966739512 Chr.3: 43720449-43720449 on Build GRCh38
rs886058496 Chr.3: 43720466-43720466 on Build GRCh38

View Full Product Details