Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 43720690-43721200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD5
Entrez Gene ID
51099
Gene Name
abhydrolase domain containing 5
Gene Aliases
CDS, CGI58, IECN2, NCIE2
UniGene
Hs.19385
Gene Chromosome Location
Chr.3: 43690883-43722725 on Build GRCh38
Transcripts
NM_016006.4 XM_011533779.2 XM_011533780.1

SNP Detail

SNP ID Chromosome Location
rs535550939 Chr.3: 43720871-43720871 on Build GRCh38
rs1041571107 Chr.3: 43721082-43721082 on Build GRCh38
rs1012685351 Chr.3: 43721157-43721157 on Build GRCh38
rs1009602610 Chr.3: 43720893-43720894 on Build GRCh38
rs539609211 Chr.3: 43720835-43720835 on Build GRCh38
rs916762006 Chr.3: 43720738-43720738 on Build GRCh38
rs893824718 Chr.3: 43721156-43721156 on Build GRCh38
rs75590382 Chr.3: 43721040-43721040 on Build GRCh38
rs563472893 Chr.3: 43721094-43721094 on Build GRCh38
rs753966054 Chr.3: 43721019-43721019 on Build GRCh38
rs375122733 Chr.3: 43720839-43720839 on Build GRCh38
rs956759601 Chr.3: 43720888-43720890 on Build GRCh38
rs903068215 Chr.3: 43721085-43721085 on Build GRCh38
rs543597397 Chr.3: 43721064-43721064 on Build GRCh38
rs886058500 Chr.3: 43721074-43721074 on Build GRCh38
rs973032054 Chr.3: 43721032-43721032 on Build GRCh38
rs1022278027 Chr.3: 43720929-43720929 on Build GRCh38
rs17075919 Chr.3: 43720856-43720856 on Build GRCh38
rs892568627 Chr.3: 43721101-43721101 on Build GRCh38
rs1008706767 Chr.3: 43720859-43720859 on Build GRCh38
rs571081349 Chr.3: 43720733-43720733 on Build GRCh38
rs76200032 Chr.3: 43720995-43720995 on Build GRCh38
rs942465762 Chr.3: 43720715-43720715 on Build GRCh38
rs903140341 Chr.3: 43720892-43720892 on Build GRCh38
rs982311926 Chr.3: 43721021-43721021 on Build GRCh38
rs890315770 Chr.3: 43720857-43720857 on Build GRCh38
rs1045421655 Chr.3: 43721067-43721067 on Build GRCh38
rs982378887 Chr.3: 43720777-43720777 on Build GRCh38
rs968310240 Chr.3: 43720932-43720932 on Build GRCh38
rs970906942 Chr.3: 43721003-43721003 on Build GRCh38
rs1037412061 Chr.3: 43720817-43720817 on Build GRCh38
rs555017968 Chr.3: 43720936-43720939 on Build GRCh38
rs1054166837 Chr.3: 43721129-43721129 on Build GRCh38
rs772371727 Chr.3: 43721140-43721140 on Build GRCh38
rs999478963 Chr.3: 43720898-43720898 on Build GRCh38
rs1022305857 Chr.3: 43721173-43721173 on Build GRCh38
rs1050167074 Chr.3: 43721056-43721056 on Build GRCh38
rs940331510 Chr.3: 43720809-43720809 on Build GRCh38
rs991354894 Chr.3: 43720723-43720723 on Build GRCh38
rs1003813049 Chr.3: 43721069-43721069 on Build GRCh38
rs886058499 Chr.3: 43720924-43720925 on Build GRCh38
rs886058498 Chr.3: 43720836-43720836 on Build GRCh38
rs555617596 Chr.3: 43720924-43720924 on Build GRCh38
rs911656338 Chr.3: 43721068-43721068 on Build GRCh38
rs529144741 Chr.3: 43721113-43721113 on Build GRCh38
rs756061412 Chr.3: 43720987-43720987 on Build GRCh38
rs779175770 Chr.3: 43721052-43721052 on Build GRCh38
rs192172748 Chr.3: 43721022-43721022 on Build GRCh38
rs962293407 Chr.3: 43721024-43721024 on Build GRCh38
rs572387719 Chr.3: 43720977-43720977 on Build GRCh38
rs1024200110 Chr.3: 43721002-43721002 on Build GRCh38
rs922346803 Chr.3: 43720740-43720740 on Build GRCh38
rs949639116 Chr.3: 43720757-43720757 on Build GRCh38
rs908153833 Chr.3: 43720799-43720799 on Build GRCh38
rs186484499 Chr.3: 43720771-43720771 on Build GRCh38
rs556346785 Chr.3: 43720838-43720838 on Build GRCh38
rs746117389 Chr.3: 43721072-43721072 on Build GRCh38
rs183518530 Chr.3: 43720716-43720716 on Build GRCh38
rs767567435 Chr.3: 43720913-43720913 on Build GRCh38
rs966642202 Chr.3: 43720991-43720991 on Build GRCh38
rs1032306562 Chr.3: 43720959-43720959 on Build GRCh38
rs931759021 Chr.3: 43720852-43720852 on Build GRCh38

View Full Product Details