Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46670105-46670599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALS2CL
Entrez Gene ID
259173
Gene Name
ALS2 C-terminal like
Gene Aliases
RN49018
UniGene
Hs.517937
Gene Chromosome Location
Chr.3: 46668995-46693713 on Build GRCh38
Transcripts
NM_001190707.1 NM_147129.4 NR_033815.2 NR_135622.1 XM_005265025.1

SNP Detail

SNP ID Chromosome Location
rs111523463 Chr.3: 46670444-46670444 on Build GRCh38
rs954841854 Chr.3: 46670328-46670328 on Build GRCh38
rs916772893 Chr.3: 46670331-46670331 on Build GRCh38
rs986329190 Chr.3: 46670335-46670335 on Build GRCh38
rs759729308 Chr.3: 46670330-46670331 on Build GRCh38
rs764114223 Chr.3: 46670241-46670241 on Build GRCh38
rs73831184 Chr.3: 46670413-46670413 on Build GRCh38
rs929887582 Chr.3: 46670171-46670171 on Build GRCh38
rs562147863 Chr.3: 46670130-46670130 on Build GRCh38
rs1047438706 Chr.3: 46670385-46670385 on Build GRCh38
rs532050382 Chr.3: 46670371-46670371 on Build GRCh38
rs910634387 Chr.3: 46670345-46670345 on Build GRCh38
rs1042700684 Chr.3: 46670454-46670454 on Build GRCh38
rs1034042952 Chr.3: 46670255-46670255 on Build GRCh38
rs952833229 Chr.3: 46670327-46670327 on Build GRCh38
rs971312426 Chr.3: 46670478-46670478 on Build GRCh38
rs188002432 Chr.3: 46670550-46670550 on Build GRCh38
rs138128338 Chr.3: 46670304-46670304 on Build GRCh38
rs371518703 Chr.3: 46670448-46670448 on Build GRCh38
rs77957046 Chr.3: 46670281-46670281 on Build GRCh38
rs947142668 Chr.3: 46670383-46670383 on Build GRCh38
rs781122264 Chr.3: 46670534-46670534 on Build GRCh38
rs919258040 Chr.3: 46670362-46670362 on Build GRCh38
rs967623119 Chr.3: 46670282-46670282 on Build GRCh38
rs554205740 Chr.3: 46670422-46670422 on Build GRCh38
rs1011743098 Chr.3: 46670453-46670453 on Build GRCh38
rs11711100 Chr.3: 46670317-46670317 on Build GRCh38
rs1049482144 Chr.3: 46670405-46670405 on Build GRCh38
rs577289561 Chr.3: 46670134-46670134 on Build GRCh38
rs192476211 Chr.3: 46670278-46670278 on Build GRCh38
rs555703248 Chr.3: 46670556-46670556 on Build GRCh38
rs1046858105 Chr.3: 46670224-46670224 on Build GRCh38
rs75741978 Chr.3: 46670425-46670425 on Build GRCh38
rs3210183 Chr.3: 46670360-46670360 on Build GRCh38
rs533786852 Chr.3: 46670473-46670473 on Build GRCh38
rs185669632 Chr.3: 46670464-46670464 on Build GRCh38
rs754769205 Chr.3: 46670525-46670525 on Build GRCh38
rs1023647226 Chr.3: 46670467-46670467 on Build GRCh38
rs141719804 Chr.3: 46670346-46670346 on Build GRCh38
rs934323842 Chr.3: 46670359-46670359 on Build GRCh38
rs550173306 Chr.3: 46670382-46670382 on Build GRCh38
rs949558602 Chr.3: 46670332-46670332 on Build GRCh38
rs999527037 Chr.3: 46670296-46670296 on Build GRCh38
rs954977565 Chr.3: 46670528-46670528 on Build GRCh38
rs894258017 Chr.3: 46670268-46670268 on Build GRCh38
rs898410821 Chr.3: 46670169-46670169 on Build GRCh38
rs571684322 Chr.3: 46670423-46670423 on Build GRCh38
rs559211982 Chr.3: 46670526-46670526 on Build GRCh38
rs548133077 Chr.3: 46670554-46670554 on Build GRCh38
rs994036332 Chr.3: 46670445-46670445 on Build GRCh38
rs555546222 Chr.3: 46670496-46670496 on Build GRCh38
rs776275083 Chr.3: 46670231-46670231 on Build GRCh38
rs907057302 Chr.3: 46670233-46670233 on Build GRCh38
rs544716689 Chr.3: 46670222-46670222 on Build GRCh38
rs566657883 Chr.3: 46670465-46670465 on Build GRCh38
rs924103560 Chr.3: 46670575-46670575 on Build GRCh38

View Full Product Details