Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46670490-46670983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALS2CL
Entrez Gene ID
259173
Gene Name
ALS2 C-terminal like
Gene Aliases
RN49018
UniGene
Hs.517937
Gene Chromosome Location
Chr.3: 46668995-46693713 on Build GRCh38
Transcripts
NM_001190707.1 NM_147129.4 NR_033815.2 NR_135622.1 XM_005265025.1

SNP Detail

SNP ID Chromosome Location
rs1017791152 Chr.3: 46670578-46670578 on Build GRCh38
rs749916076 Chr.3: 46670949-46670949 on Build GRCh38
rs995038852 Chr.3: 46670895-46670895 on Build GRCh38
rs756823469 Chr.3: 46670939-46670939 on Build GRCh38
rs565258302 Chr.3: 46670858-46670858 on Build GRCh38
rs542809971 Chr.3: 46670628-46670628 on Build GRCh38
rs938638790 Chr.3: 46670722-46670722 on Build GRCh38
rs532011356 Chr.3: 46670748-46670748 on Build GRCh38
rs1007824476 Chr.3: 46670813-46670813 on Build GRCh38
rs902962450 Chr.3: 46670758-46670758 on Build GRCh38
rs755744810 Chr.3: 46670935-46670935 on Build GRCh38
rs763928849 Chr.3: 46670952-46670952 on Build GRCh38
rs950756213 Chr.3: 46670643-46670643 on Build GRCh38
rs757945773 Chr.3: 46670961-46670961 on Build GRCh38
rs188002432 Chr.3: 46670550-46670550 on Build GRCh38
rs1012168988 Chr.3: 46670863-46670863 on Build GRCh38
rs369238932 Chr.3: 46670621-46670621 on Build GRCh38
rs1040917147 Chr.3: 46670773-46670773 on Build GRCh38
rs777115725 Chr.3: 46670715-46670715 on Build GRCh38
rs1017755272 Chr.3: 46670854-46670854 on Build GRCh38
rs565999255 Chr.3: 46670936-46670936 on Build GRCh38
rs192093319 Chr.3: 46670793-46670793 on Build GRCh38
rs528416242 Chr.3: 46670808-46670808 on Build GRCh38
rs781122264 Chr.3: 46670534-46670534 on Build GRCh38
rs184390150 Chr.3: 46670908-46670908 on Build GRCh38
rs1031191078 Chr.3: 46670947-46670947 on Build GRCh38
rs1043181845 Chr.3: 46670756-46670756 on Build GRCh38
rs889503014 Chr.3: 46670721-46670721 on Build GRCh38
rs776606672 Chr.3: 46670800-46670813 on Build GRCh38
rs555703248 Chr.3: 46670556-46670556 on Build GRCh38
rs532598902 Chr.3: 46670859-46670859 on Build GRCh38
rs561554867 Chr.3: 46670739-46670739 on Build GRCh38
rs577250101 Chr.3: 46670577-46670577 on Build GRCh38
rs754769205 Chr.3: 46670525-46670525 on Build GRCh38
rs545064803 Chr.3: 46670584-46670584 on Build GRCh38
rs947006914 Chr.3: 46670624-46670624 on Build GRCh38
rs1026802187 Chr.3: 46670927-46670927 on Build GRCh38
rs752595591 Chr.3: 46670581-46670581 on Build GRCh38
rs954977565 Chr.3: 46670528-46670528 on Build GRCh38
rs113245918 Chr.3: 46670620-46670620 on Build GRCh38
rs888462966 Chr.3: 46670847-46670847 on Build GRCh38
rs998921488 Chr.3: 46670791-46670791 on Build GRCh38
rs963425224 Chr.3: 46670579-46670579 on Build GRCh38
rs755609955 Chr.3: 46670662-46670662 on Build GRCh38
rs559211982 Chr.3: 46670526-46670526 on Build GRCh38
rs548133077 Chr.3: 46670554-46670554 on Build GRCh38
rs748854273 Chr.3: 46670740-46670740 on Build GRCh38
rs899345291 Chr.3: 46670879-46670879 on Build GRCh38
rs758079880 Chr.3: 46670946-46670946 on Build GRCh38
rs924103560 Chr.3: 46670575-46670575 on Build GRCh38
rs111751718 Chr.3: 46670706-46670706 on Build GRCh38
rs928546792 Chr.3: 46670701-46670701 on Build GRCh38

View Full Product Details