Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46670875-46671367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALS2CL
Entrez Gene ID
259173
Gene Name
ALS2 C-terminal like
Gene Aliases
RN49018
UniGene
Hs.517937
Gene Chromosome Location
Chr.3: 46668995-46693713 on Build GRCh38
Transcripts
NM_001190707.1 NM_147129.4 NR_033815.2 NR_135622.1 XM_005265025.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3068279 c.2791G>A p.E931K Chr.3: 46671055-46671055 on Build GRCh38
3594560 c.2799C>T p.I933I Chr.3: 46671047-46671047 on Build GRCh38
1741746 c.2846C>T p.S949F Chr.3: 46671000-46671000 on Build GRCh38
4540507 c.2851G>T p.E951* Chr.3: 46670995-46670995 on Build GRCh38
6164994 c.2809G>A p.D937N Chr.3: 46671037-46671037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747138367 Chr.3: 46671115-46671115 on Build GRCh38
rs915852238 Chr.3: 46671035-46671035 on Build GRCh38
rs572021791 Chr.3: 46671158-46671158 on Build GRCh38
rs995404651 Chr.3: 46671182-46671182 on Build GRCh38
rs570904633 Chr.3: 46671045-46671045 on Build GRCh38
rs749916076 Chr.3: 46670949-46670949 on Build GRCh38
rs138204155 Chr.3: 46671144-46671144 on Build GRCh38
rs748006382 Chr.3: 46670995-46670995 on Build GRCh38
rs768652836 Chr.3: 46670977-46670977 on Build GRCh38
rs149150920 Chr.3: 46671024-46671024 on Build GRCh38
rs756823469 Chr.3: 46670939-46670939 on Build GRCh38
rs1022467201 Chr.3: 46671238-46671238 on Build GRCh38
rs959348660 Chr.3: 46671258-46671258 on Build GRCh38
rs779271304 Chr.3: 46671099-46671099 on Build GRCh38
rs780782567 Chr.3: 46670982-46670982 on Build GRCh38
rs542725664 Chr.3: 46671176-46671176 on Build GRCh38
rs781210496 Chr.3: 46671090-46671090 on Build GRCh38
rs368965614 Chr.3: 46671087-46671087 on Build GRCh38
rs755744810 Chr.3: 46670935-46670935 on Build GRCh38
rs906507983 Chr.3: 46671175-46671175 on Build GRCh38
rs149595583 Chr.3: 46671244-46671244 on Build GRCh38
rs763928849 Chr.3: 46670952-46670952 on Build GRCh38
rs1041175065 Chr.3: 46671060-46671060 on Build GRCh38
rs757945773 Chr.3: 46670961-46670961 on Build GRCh38
rs956865882 Chr.3: 46671218-46671218 on Build GRCh38
rs370191447 Chr.3: 46671093-46671093 on Build GRCh38
rs575936933 Chr.3: 46671269-46671270 on Build GRCh38
rs775675461 Chr.3: 46671113-46671113 on Build GRCh38
rs1026522067 Chr.3: 46671189-46671189 on Build GRCh38
rs772346681 Chr.3: 46671256-46671256 on Build GRCh38
rs532849213 Chr.3: 46671277-46671277 on Build GRCh38
rs144181784 Chr.3: 46671021-46671021 on Build GRCh38
rs565999255 Chr.3: 46670936-46670936 on Build GRCh38
rs189148441 Chr.3: 46670971-46670971 on Build GRCh38
rs772432807 Chr.3: 46671109-46671109 on Build GRCh38
rs537730786 Chr.3: 46671022-46671022 on Build GRCh38
rs1031191078 Chr.3: 46670947-46670947 on Build GRCh38
rs184390150 Chr.3: 46670908-46670908 on Build GRCh38
rs151283213 Chr.3: 46671058-46671058 on Build GRCh38
rs565620006 Chr.3: 46671260-46671260 on Build GRCh38
rs990810393 Chr.3: 46671314-46671314 on Build GRCh38
rs781189708 Chr.3: 46671247-46671248 on Build GRCh38
rs918447223 Chr.3: 46671065-46671065 on Build GRCh38
rs200979337 Chr.3: 46671094-46671094 on Build GRCh38
rs375535702 Chr.3: 46671292-46671292 on Build GRCh38
rs576737754 Chr.3: 46671208-46671208 on Build GRCh38
rs555185785 Chr.3: 46671203-46671203 on Build GRCh38
rs943576389 Chr.3: 46671131-46671131 on Build GRCh38
rs764697064 Chr.3: 46671032-46671032 on Build GRCh38
rs746178342 Chr.3: 46671016-46671016 on Build GRCh38
rs374861021 Chr.3: 46671056-46671056 on Build GRCh38
rs910914654 Chr.3: 46671276-46671276 on Build GRCh38
rs774761469 Chr.3: 46671003-46671003 on Build GRCh38
rs1026802187 Chr.3: 46670927-46670927 on Build GRCh38
rs1011037108 Chr.3: 46671234-46671234 on Build GRCh38
rs747006949 Chr.3: 46670974-46670974 on Build GRCh38
rs77367607 Chr.3: 46671000-46671000 on Build GRCh38
rs373709504 Chr.3: 46671209-46671209 on Build GRCh38
rs567139051 Chr.3: 46671004-46671004 on Build GRCh38
rs965394811 Chr.3: 46671280-46671280 on Build GRCh38
rs751083820 Chr.3: 46671059-46671059 on Build GRCh38
rs746145601 Chr.3: 46671103-46671103 on Build GRCh38
rs1034962087 Chr.3: 46671248-46671248 on Build GRCh38
rs758079880 Chr.3: 46670946-46670946 on Build GRCh38

View Full Product Details