Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46672003-46672510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALS2CL
Entrez Gene ID
259173
Gene Name
ALS2 C-terminal like
Gene Aliases
RN49018
UniGene
Hs.517937
Gene Chromosome Location
Chr.3: 46668995-46693713 on Build GRCh38
Transcripts
NM_001190707.1 NM_147129.4 NR_033815.2 NR_135622.1 XM_005265025.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3068288 c.2541G>A p.T847T Chr.3: 46672027-46672027 on Build GRCh38
1283388 c.2532C>A p.I844I Chr.3: 46672142-46672142 on Build GRCh38
6026804 c.2484G>T p.L828L Chr.3: 46672190-46672190 on Build GRCh38
6301288 c.2552G>A p.R851Q Chr.3: 46672016-46672016 on Build GRCh38
1423429 c.2515G>A p.E839K Chr.3: 46672159-46672159 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs887502221 Chr.3: 46672440-46672440 on Build GRCh38
rs377194325 Chr.3: 46672127-46672127 on Build GRCh38
rs891677215 Chr.3: 46672090-46672090 on Build GRCh38
rs775618276 Chr.3: 46672159-46672159 on Build GRCh38
rs749122253 Chr.3: 46672031-46672031 on Build GRCh38
rs768096819 Chr.3: 46672226-46672226 on Build GRCh38
rs548179214 Chr.3: 46672317-46672317 on Build GRCh38
rs1047967609 Chr.3: 46672308-46672308 on Build GRCh38
rs761147004 Chr.3: 46672240-46672240 on Build GRCh38
rs3935248 Chr.3: 46672375-46672375 on Build GRCh38
rs569682911 Chr.3: 46672361-46672361 on Build GRCh38
rs916856327 Chr.3: 46672210-46672210 on Build GRCh38
rs773117961 Chr.3: 46672129-46672129 on Build GRCh38
rs1002379361 Chr.3: 46672400-46672400 on Build GRCh38
rs367731987 Chr.3: 46672266-46672266 on Build GRCh38
rs527408088 Chr.3: 46672052-46672052 on Build GRCh38
rs752056950 Chr.3: 46672193-46672194 on Build GRCh38
rs778811928 Chr.3: 46672188-46672188 on Build GRCh38
rs564912464 Chr.3: 46672121-46672121 on Build GRCh38
rs541779411 Chr.3: 46672064-46672064 on Build GRCh38
rs374281760 Chr.3: 46672204-46672204 on Build GRCh38
rs923454090 Chr.3: 46672288-46672288 on Build GRCh38
rs766928282 Chr.3: 46672191-46672192 on Build GRCh38
rs773525547 Chr.3: 46672205-46672205 on Build GRCh38
rs903962159 Chr.3: 46672065-46672065 on Build GRCh38
rs1027905265 Chr.3: 46672406-46672406 on Build GRCh38
rs368143166 Chr.3: 46672089-46672089 on Build GRCh38
rs374276986 Chr.3: 46672082-46672082 on Build GRCh38
rs752951452 Chr.3: 46672235-46672235 on Build GRCh38
rs377241827 Chr.3: 46672160-46672160 on Build GRCh38
rs763620570 Chr.3: 46672246-46672246 on Build GRCh38
rs761942059 Chr.3: 46672071-46672071 on Build GRCh38
rs775372245 Chr.3: 46672122-46672122 on Build GRCh38
rs761878946 Chr.3: 46672125-46672125 on Build GRCh38
rs61277784 Chr.3: 46672274-46672274 on Build GRCh38
rs370602500 Chr.3: 46672027-46672027 on Build GRCh38
rs774088054 Chr.3: 46672051-46672051 on Build GRCh38
rs763176763 Chr.3: 46672135-46672135 on Build GRCh38
rs367618629 Chr.3: 46672108-46672108 on Build GRCh38
rs935335792 Chr.3: 46672079-46672079 on Build GRCh38
rs944582565 Chr.3: 46672328-46672328 on Build GRCh38
rs766963598 Chr.3: 46672139-46672139 on Build GRCh38
rs755804098 Chr.3: 46672105-46672105 on Build GRCh38
rs980041213 Chr.3: 46672255-46672255 on Build GRCh38
rs148591844 Chr.3: 46672190-46672190 on Build GRCh38
rs764276046 Chr.3: 46672057-46672057 on Build GRCh38
rs996022484 Chr.3: 46672397-46672397 on Build GRCh38
rs758793004 Chr.3: 46672083-46672083 on Build GRCh38
rs375379223 Chr.3: 46672407-46672407 on Build GRCh38
rs369596510 Chr.3: 46672184-46672184 on Build GRCh38
rs763121973 Chr.3: 46672209-46672209 on Build GRCh38
rs548058222 Chr.3: 46672143-46672143 on Build GRCh38
rs779678498 Chr.3: 46672194-46672194 on Build GRCh38
rs774585446 Chr.3: 46672218-46672218 on Build GRCh38
rs570950182 Chr.3: 46672481-46672481 on Build GRCh38
rs888066686 Chr.3: 46672329-46672329 on Build GRCh38
rs886780848 Chr.3: 46672074-46672074 on Build GRCh38
rs181077701 Chr.3: 46672435-46672435 on Build GRCh38
rs773486618 Chr.3: 46672187-46672188 on Build GRCh38
rs201683237 Chr.3: 46672197-46672197 on Build GRCh38
rs891144100 Chr.3: 46672286-46672286 on Build GRCh38
rs35302659 Chr.3: 46672255-46672256 on Build GRCh38
rs760053713 Chr.3: 46672219-46672219 on Build GRCh38
rs929440757 Chr.3: 46672289-46672289 on Build GRCh38
rs778867387 Chr.3: 46672115-46672115 on Build GRCh38
rs144505150 Chr.3: 46672028-46672028 on Build GRCh38
rs371657552 Chr.3: 46672097-46672097 on Build GRCh38
rs770856299 Chr.3: 46672045-46672045 on Build GRCh38
rs375923505 Chr.3: 46672104-46672104 on Build GRCh38
rs935414956 Chr.3: 46672273-46672273 on Build GRCh38
rs771191067 Chr.3: 46672213-46672213 on Build GRCh38
rs372807128 Chr.3: 46672109-46672109 on Build GRCh38
rs960832514 Chr.3: 46672446-46672446 on Build GRCh38
rs113488576 Chr.3: 46672049-46672050 on Build GRCh38
rs139684532 Chr.3: 46672185-46672185 on Build GRCh38
rs147524245 Chr.3: 46672091-46672091 on Build GRCh38
rs200361913 Chr.3: 46672077-46672077 on Build GRCh38
rs755435445 Chr.3: 46672092-46672092 on Build GRCh38
rs545930278 Chr.3: 46672110-46672110 on Build GRCh38
rs759965651 Chr.3: 46672140-46672140 on Build GRCh38
rs763353135 Chr.3: 46672189-46672190 on Build GRCh38

View Full Product Details