Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48565018-48565513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL7A1
Entrez Gene ID
1294
Gene Name
collagen type VII alpha 1
Gene Aliases
EBD1, EBDCT, EBR1, NDNC8
UniGene
Hs.476218
Gene Chromosome Location
Chr.3: 48564073-48595302 on Build GRCh38
Transcripts
NM_000094.3 XM_011533337.1 XM_017005688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7362339 c.8467A>G p.T2823A Chr.3: 48565470-48565470 on Build GRCh38
112098 c.8565_8566insCC p.S2856fs*43 Chr.3: 48565163-48565164 on Build GRCh38
5615294 c.8512C>A p.H2838N Chr.3: 48565425-48565425 on Build GRCh38
7416788 c.8471C>T p.A2824V Chr.3: 48565466-48565466 on Build GRCh38
6010969 c.8605C>T p.P2869S Chr.3: 48565124-48565124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746146532 Chr.3: 48565118-48565118 on Build GRCh38
rs201339290 Chr.3: 48565435-48565435 on Build GRCh38
rs762451231 Chr.3: 48565361-48565361 on Build GRCh38
rs372154665 Chr.3: 48565307-48565307 on Build GRCh38
rs879016311 Chr.3: 48565388-48565388 on Build GRCh38
rs758649802 Chr.3: 48565198-48565198 on Build GRCh38
rs781077978 Chr.3: 48565246-48565246 on Build GRCh38
rs772690795 Chr.3: 48565247-48565247 on Build GRCh38
rs149393855 Chr.3: 48565413-48565413 on Build GRCh38
rs749395019 Chr.3: 48565098-48565098 on Build GRCh38
rs374396716 Chr.3: 48565240-48565241 on Build GRCh38
rs775951487 Chr.3: 48565368-48565368 on Build GRCh38
rs550169353 Chr.3: 48565297-48565297 on Build GRCh38
rs566181351 Chr.3: 48565400-48565413 on Build GRCh38
rs35631233 Chr.3: 48565091-48565092 on Build GRCh38
rs770632676 Chr.3: 48565362-48565362 on Build GRCh38
rs913050647 Chr.3: 48565056-48565056 on Build GRCh38
rs751914144 Chr.3: 48565085-48565085 on Build GRCh38
rs771747896 Chr.3: 48565119-48565119 on Build GRCh38
rs748854889 Chr.3: 48565432-48565432 on Build GRCh38
rs975284002 Chr.3: 48565257-48565257 on Build GRCh38
rs908283742 Chr.3: 48565330-48565330 on Build GRCh38
rs939696846 Chr.3: 48565331-48565331 on Build GRCh38
rs148159956 Chr.3: 48565429-48565429 on Build GRCh38
rs921061602 Chr.3: 48565298-48565298 on Build GRCh38
rs750604521 Chr.3: 48565193-48565193 on Build GRCh38
rs776644467 Chr.3: 48565210-48565212 on Build GRCh38
rs762728307 Chr.3: 48565165-48565165 on Build GRCh38
rs762512119 Chr.3: 48565322-48565322 on Build GRCh38
rs879469736 Chr.3: 48565462-48565462 on Build GRCh38
rs138595277 Chr.3: 48565090-48565091 on Build GRCh38
rs372166543 Chr.3: 48565160-48565160 on Build GRCh38
rs531585457 Chr.3: 48565243-48565243 on Build GRCh38
rs777583032 Chr.3: 48565076-48565076 on Build GRCh38
rs781022857 Chr.3: 48565423-48565423 on Build GRCh38
rs140041143 Chr.3: 48565190-48565190 on Build GRCh38
rs780284810 Chr.3: 48565401-48565401 on Build GRCh38
rs146892299 Chr.3: 48565321-48565321 on Build GRCh38
rs950877774 Chr.3: 48565248-48565256 on Build GRCh38
rs761813842 Chr.3: 48565373-48565375 on Build GRCh38
rs768592959 Chr.3: 48565151-48565151 on Build GRCh38
rs751768659 Chr.3: 48565480-48565480 on Build GRCh38
rs753118545 Chr.3: 48565180-48565180 on Build GRCh38
rs139308059 Chr.3: 48565464-48565464 on Build GRCh38
rs148454724 Chr.3: 48565161-48565161 on Build GRCh38
rs755260852 Chr.3: 48565490-48565490 on Build GRCh38
rs1036649782 Chr.3: 48565078-48565078 on Build GRCh38
rs747790197 Chr.3: 48565189-48565189 on Build GRCh38
rs368947460 Chr.3: 48565146-48565146 on Build GRCh38
rs532051347 Chr.3: 48565077-48565077 on Build GRCh38
rs754379615 Chr.3: 48565383-48565383 on Build GRCh38
rs764599739 Chr.3: 48565375-48565375 on Build GRCh38
rs779889929 Chr.3: 48565134-48565134 on Build GRCh38
rs542807792 Chr.3: 48565402-48565402 on Build GRCh38
rs116591500 Chr.3: 48565465-48565465 on Build GRCh38
rs750529756 Chr.3: 48565398-48565398 on Build GRCh38
rs1001325189 Chr.3: 48565094-48565094 on Build GRCh38
rs41290684 Chr.3: 48565086-48565086 on Build GRCh38
rs1045246092 Chr.3: 48565378-48565378 on Build GRCh38
rs748032086 Chr.3: 48565425-48565425 on Build GRCh38
rs138002006 Chr.3: 48565199-48565199 on Build GRCh38
rs371458442 Chr.3: 48565369-48565369 on Build GRCh38
rs777935673 Chr.3: 48565096-48565096 on Build GRCh38
rs772717870 Chr.3: 48565163-48565163 on Build GRCh38
rs757458214 Chr.3: 48565106-48565106 on Build GRCh38
rs765795502 Chr.3: 48565441-48565441 on Build GRCh38
rs189731761 Chr.3: 48565391-48565391 on Build GRCh38
rs905322767 Chr.3: 48565384-48565384 on Build GRCh38
rs1003879036 Chr.3: 48565051-48565051 on Build GRCh38
rs536894832 Chr.3: 48565434-48565434 on Build GRCh38
rs753245446 Chr.3: 48565092-48565092 on Build GRCh38
rs746827485 Chr.3: 48565239-48565239 on Build GRCh38
rs1039134351 Chr.3: 48565179-48565179 on Build GRCh38
rs756085968 Chr.3: 48565427-48565427 on Build GRCh38
rs767773721 Chr.3: 48565167-48565167 on Build GRCh38
rs762167271 Chr.3: 48565440-48565440 on Build GRCh38
rs761395853 Chr.3: 48565059-48565059 on Build GRCh38
rs769174648 Chr.3: 48565359-48565359 on Build GRCh38
rs138432078 Chr.3: 48565089-48565090 on Build GRCh38
rs886734235 Chr.3: 48565042-48565042 on Build GRCh38
rs576703460 Chr.3: 48565437-48565437 on Build GRCh38
rs756755395 Chr.3: 48565093-48565093 on Build GRCh38
rs143054969 Chr.3: 48565453-48565453 on Build GRCh38
rs896727342 Chr.3: 48565083-48565083 on Build GRCh38
rs772849671 Chr.3: 48565360-48565360 on Build GRCh38
rs139014122 Chr.3: 48565433-48565433 on Build GRCh38
rs778969774 Chr.3: 48565117-48565117 on Build GRCh38
rs767974409 Chr.3: 48565087-48565087 on Build GRCh38
rs765162830 Chr.3: 48565392-48565392 on Build GRCh38
rs387906604 Chr.3: 48565458-48565458 on Build GRCh38
rs79473918 Chr.3: 48565188-48565188 on Build GRCh38
rs7624006 Chr.3: 48565090-48565090 on Build GRCh38
rs963083485 Chr.3: 48565351-48565351 on Build GRCh38
rs539095375 Chr.3: 48565308-48565308 on Build GRCh38

View Full Product Details