Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48565168-48565663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL7A1
Entrez Gene ID
1294
Gene Name
collagen type VII alpha 1
Gene Aliases
EBD1, EBDCT, EBR1, NDNC8
UniGene
Hs.476218
Gene Chromosome Location
Chr.3: 48564073-48595302 on Build GRCh38
Transcripts
NM_000094.3 XM_011533337.1 XM_017005688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7362339 c.8467A>G p.T2823A Chr.3: 48565470-48565470 on Build GRCh38
5615294 c.8512C>A p.H2838N Chr.3: 48565425-48565425 on Build GRCh38
7416788 c.8471C>T p.A2824V Chr.3: 48565466-48565466 on Build GRCh38
4824457 c.8518G>C p.E2840Q Chr.3: 48565419-48565419 on Build GRCh38
3070534 c.8459C>A p.A2820D Chr.3: 48565478-48565478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762451231 Chr.3: 48565361-48565361 on Build GRCh38
rs201339290 Chr.3: 48565435-48565435 on Build GRCh38
rs777800464 Chr.3: 48565523-48565523 on Build GRCh38
rs372154665 Chr.3: 48565307-48565307 on Build GRCh38
rs879016311 Chr.3: 48565388-48565388 on Build GRCh38
rs758649802 Chr.3: 48565198-48565198 on Build GRCh38
rs781077978 Chr.3: 48565246-48565246 on Build GRCh38
rs772690795 Chr.3: 48565247-48565247 on Build GRCh38
rs149393855 Chr.3: 48565413-48565413 on Build GRCh38
rs770094470 Chr.3: 48565604-48565604 on Build GRCh38
rs749229597 Chr.3: 48565539-48565539 on Build GRCh38
rs374396716 Chr.3: 48565240-48565241 on Build GRCh38
rs965327916 Chr.3: 48565558-48565558 on Build GRCh38
rs775951487 Chr.3: 48565368-48565368 on Build GRCh38
rs1016529136 Chr.3: 48565595-48565595 on Build GRCh38
rs773588700 Chr.3: 48565506-48565507 on Build GRCh38
rs550169353 Chr.3: 48565297-48565297 on Build GRCh38
rs566181351 Chr.3: 48565400-48565413 on Build GRCh38
rs767236747 Chr.3: 48565496-48565496 on Build GRCh38
rs770632676 Chr.3: 48565362-48565362 on Build GRCh38
rs368639366 Chr.3: 48565601-48565601 on Build GRCh38
rs771863426 Chr.3: 48565588-48565588 on Build GRCh38
rs778629867 Chr.3: 48565542-48565542 on Build GRCh38
rs748854889 Chr.3: 48565432-48565432 on Build GRCh38
rs908283742 Chr.3: 48565330-48565330 on Build GRCh38
rs778507083 Chr.3: 48565638-48565638 on Build GRCh38
rs939696846 Chr.3: 48565331-48565331 on Build GRCh38
rs975284002 Chr.3: 48565257-48565257 on Build GRCh38
rs759699315 Chr.3: 48565616-48565616 on Build GRCh38
rs745444203 Chr.3: 48565587-48565587 on Build GRCh38
rs200575328 Chr.3: 48565628-48565628 on Build GRCh38
rs148159956 Chr.3: 48565429-48565429 on Build GRCh38
rs748576670 Chr.3: 48565602-48565602 on Build GRCh38
rs921061602 Chr.3: 48565298-48565298 on Build GRCh38
rs776644467 Chr.3: 48565210-48565212 on Build GRCh38
rs750604521 Chr.3: 48565193-48565193 on Build GRCh38
rs879469736 Chr.3: 48565462-48565462 on Build GRCh38
rs762512119 Chr.3: 48565322-48565322 on Build GRCh38
rs34010829 Chr.3: 48565613-48565614 on Build GRCh38
rs531585457 Chr.3: 48565243-48565243 on Build GRCh38
rs752397643 Chr.3: 48565620-48565620 on Build GRCh38
rs756810451 Chr.3: 48565541-48565541 on Build GRCh38
rs781022857 Chr.3: 48565423-48565423 on Build GRCh38
rs780284810 Chr.3: 48565401-48565401 on Build GRCh38
rs373849523 Chr.3: 48565512-48565512 on Build GRCh38
rs146892299 Chr.3: 48565321-48565321 on Build GRCh38
rs950877774 Chr.3: 48565248-48565256 on Build GRCh38
rs1051795889 Chr.3: 48565556-48565556 on Build GRCh38
rs774420894 Chr.3: 48565615-48565615 on Build GRCh38
rs761813842 Chr.3: 48565373-48565375 on Build GRCh38
rs751768659 Chr.3: 48565480-48565480 on Build GRCh38
rs375356289 Chr.3: 48565563-48565563 on Build GRCh38
rs139308059 Chr.3: 48565464-48565464 on Build GRCh38
rs755260852 Chr.3: 48565490-48565490 on Build GRCh38
rs752553107 Chr.3: 48565506-48565506 on Build GRCh38
rs754379615 Chr.3: 48565383-48565383 on Build GRCh38
rs764599739 Chr.3: 48565375-48565375 on Build GRCh38
rs116591500 Chr.3: 48565465-48565465 on Build GRCh38
rs1001774733 Chr.3: 48565635-48565635 on Build GRCh38
rs542807792 Chr.3: 48565402-48565402 on Build GRCh38
rs750529756 Chr.3: 48565398-48565398 on Build GRCh38
rs113593855 Chr.3: 48565634-48565634 on Build GRCh38
rs1045246092 Chr.3: 48565378-48565378 on Build GRCh38
rs755960601 Chr.3: 48565509-48565509 on Build GRCh38
rs759833929 Chr.3: 48565578-48565578 on Build GRCh38
rs748032086 Chr.3: 48565425-48565425 on Build GRCh38
rs371458442 Chr.3: 48565369-48565369 on Build GRCh38
rs138002006 Chr.3: 48565199-48565199 on Build GRCh38
rs143457874 Chr.3: 48565636-48565636 on Build GRCh38
rs111639610 Chr.3: 48565633-48565633 on Build GRCh38
rs765795502 Chr.3: 48565441-48565441 on Build GRCh38
rs189731761 Chr.3: 48565391-48565391 on Build GRCh38
rs905322767 Chr.3: 48565384-48565384 on Build GRCh38
rs376929936 Chr.3: 48565609-48565609 on Build GRCh38
rs536894832 Chr.3: 48565434-48565434 on Build GRCh38
rs746827485 Chr.3: 48565239-48565239 on Build GRCh38
rs766445126 Chr.3: 48565612-48565612 on Build GRCh38
rs762167271 Chr.3: 48565440-48565440 on Build GRCh38
rs756085968 Chr.3: 48565427-48565427 on Build GRCh38
rs755912924 Chr.3: 48565622-48565622 on Build GRCh38
rs113223137 Chr.3: 48565497-48565497 on Build GRCh38
rs769174648 Chr.3: 48565359-48565359 on Build GRCh38
rs576703460 Chr.3: 48565437-48565437 on Build GRCh38
rs143054969 Chr.3: 48565453-48565453 on Build GRCh38
rs776810293 Chr.3: 48565598-48565598 on Build GRCh38
rs772849671 Chr.3: 48565360-48565360 on Build GRCh38
rs139014122 Chr.3: 48565433-48565433 on Build GRCh38
rs765162830 Chr.3: 48565392-48565392 on Build GRCh38
rs387906604 Chr.3: 48565458-48565458 on Build GRCh38
rs373216427 Chr.3: 48565611-48565611 on Build GRCh38
rs963083485 Chr.3: 48565351-48565351 on Build GRCh38
rs539095375 Chr.3: 48565308-48565308 on Build GRCh38

View Full Product Details