Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48641761-48642259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR3
Entrez Gene ID
1951
Gene Name
cadherin EGF LAG seven-pass G-type receptor 3
Gene Aliases
ADGRC3, CDHF11, EGFL1, FMI1, HFMI1, MEGF2, RESDA1
UniGene
Hs.631926
Gene Chromosome Location
Chr.3: 48636463-48662915 on Build GRCh38
Transcripts
NM_001407.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5726685 c.8775G>A p.R2925R Chr.3: 48641915-48641915 on Build GRCh38
7181500 c.8718C>G p.L2906L Chr.3: 48641957-48641957 on Build GRCh38
7181501 c.8733C>G p.L2911L Chr.3: 48641957-48641957 on Build GRCh38
4671473 c.8756C>T p.T2919M Chr.3: 48641919-48641919 on Build GRCh38
1045903 c.8739C>T p.S2913S Chr.3: 48641936-48641936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887480961 Chr.3: 48642187-48642187 on Build GRCh38
rs540289509 Chr.3: 48642127-48642127 on Build GRCh38
rs747576733 Chr.3: 48641923-48641923 on Build GRCh38
rs969788358 Chr.3: 48642233-48642233 on Build GRCh38
rs148633163 Chr.3: 48641900-48641900 on Build GRCh38
rs1022760552 Chr.3: 48642231-48642231 on Build GRCh38
rs955432428 Chr.3: 48641782-48641782 on Build GRCh38
rs747849249 Chr.3: 48641822-48641822 on Build GRCh38
rs775249241 Chr.3: 48642096-48642096 on Build GRCh38
rs1025985400 Chr.3: 48641984-48641984 on Build GRCh38
rs767449268 Chr.3: 48641977-48641977 on Build GRCh38
rs943577063 Chr.3: 48642166-48642166 on Build GRCh38
rs773333376 Chr.3: 48641935-48641935 on Build GRCh38
rs770006980 Chr.3: 48641932-48641932 on Build GRCh38
rs919654140 Chr.3: 48641861-48641861 on Build GRCh38
rs544468713 Chr.3: 48642181-48642181 on Build GRCh38
rs916806994 Chr.3: 48642226-48642226 on Build GRCh38
rs780740673 Chr.3: 48641910-48641910 on Build GRCh38
rs989094339 Chr.3: 48642104-48642104 on Build GRCh38
rs760805913 Chr.3: 48641859-48641859 on Build GRCh38
rs746225137 Chr.3: 48641829-48641829 on Build GRCh38
rs370620381 Chr.3: 48642015-48642015 on Build GRCh38
rs573761768 Chr.3: 48642157-48642157 on Build GRCh38
rs761994257 Chr.3: 48641931-48641931 on Build GRCh38
rs951042380 Chr.3: 48641902-48641902 on Build GRCh38
rs768975822 Chr.3: 48642216-48642216 on Build GRCh38
rs994124401 Chr.3: 48641927-48641927 on Build GRCh38
rs759768540 Chr.3: 48641858-48641858 on Build GRCh38
rs528538834 Chr.3: 48641841-48641841 on Build GRCh38
rs948162683 Chr.3: 48641936-48641936 on Build GRCh38
rs751487887 Chr.3: 48641955-48641955 on Build GRCh38
rs752345322 Chr.3: 48641916-48641916 on Build GRCh38
rs199889115 Chr.3: 48641903-48641903 on Build GRCh38
rs928571435 Chr.3: 48642077-48642077 on Build GRCh38
rs903917167 Chr.3: 48641972-48641972 on Build GRCh38
rs753482650 Chr.3: 48642007-48642007 on Build GRCh38
rs376729798 Chr.3: 48642139-48642139 on Build GRCh38
rs113642286 Chr.3: 48642095-48642095 on Build GRCh38
rs780586975 Chr.3: 48641922-48641922 on Build GRCh38
rs539973694 Chr.3: 48642126-48642126 on Build GRCh38
rs776977294 Chr.3: 48641890-48641890 on Build GRCh38
rs758776366 Chr.3: 48642008-48642008 on Build GRCh38
rs1043807001 Chr.3: 48641961-48641961 on Build GRCh38
rs758145394 Chr.3: 48641901-48641901 on Build GRCh38
rs982822845 Chr.3: 48641915-48641915 on Build GRCh38
rs992351104 Chr.3: 48642191-48642191 on Build GRCh38
rs142164680 Chr.3: 48642111-48642111 on Build GRCh38
rs969903251 Chr.3: 48642060-48642060 on Build GRCh38
rs765087604 Chr.3: 48641946-48641946 on Build GRCh38
rs982508502 Chr.3: 48642134-48642134 on Build GRCh38
rs946822971 Chr.3: 48641830-48641830 on Build GRCh38
rs951779973 Chr.3: 48641986-48641986 on Build GRCh38
rs1040509277 Chr.3: 48642175-48642175 on Build GRCh38
rs764317806 Chr.3: 48641866-48641866 on Build GRCh38
rs752147044 Chr.3: 48641978-48641978 on Build GRCh38
rs961016669 Chr.3: 48642184-48642184 on Build GRCh38
rs777442730 Chr.3: 48641998-48641998 on Build GRCh38
rs535700877 Chr.3: 48641968-48641968 on Build GRCh38
rs755602790 Chr.3: 48641981-48641981 on Build GRCh38
rs774508555 Chr.3: 48641826-48641826 on Build GRCh38
rs557207583 Chr.3: 48642056-48642056 on Build GRCh38
rs762781631 Chr.3: 48641940-48641940 on Build GRCh38
rs774848061 Chr.3: 48641828-48641828 on Build GRCh38
rs986651339 Chr.3: 48641786-48641786 on Build GRCh38
rs868600719 Chr.3: 48642182-48642182 on Build GRCh38
rs762192150 Chr.3: 48641896-48641896 on Build GRCh38
rs114112415 Chr.3: 48642147-48642147 on Build GRCh38
rs1052823287 Chr.3: 48641999-48641999 on Build GRCh38
rs754466127 Chr.3: 48641908-48641908 on Build GRCh38
rs1018146525 Chr.3: 48642082-48642082 on Build GRCh38
rs751541203 Chr.3: 48641906-48641906 on Build GRCh38
rs780347312 Chr.3: 48642049-48642049 on Build GRCh38
rs981271210 Chr.3: 48642076-48642076 on Build GRCh38
rs187876703 Chr.3: 48641919-48641919 on Build GRCh38
rs1018115123 Chr.3: 48641796-48641796 on Build GRCh38
rs746195414 Chr.3: 48641917-48641917 on Build GRCh38
rs569254108 Chr.3: 48642088-48642088 on Build GRCh38
rs200119182 Chr.3: 48641842-48641842 on Build GRCh38
rs768850213 Chr.3: 48641930-48641930 on Build GRCh38
rs771320228 Chr.3: 48641803-48641803 on Build GRCh38

View Full Product Details