Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48918524-48919034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARIH2
Entrez Gene ID
10425
Gene Name
ariadne RBR E3 ubiquitin protein ligase 2
Gene Aliases
ARI2, TRIAD1
UniGene
Hs.633601
Gene Chromosome Location
Chr.3: 48918820-48985541 on Build GRCh38
Transcripts
NM_001317333.1 NM_001317334.1 NM_006321.3 XM_011533265.1 XM_011533266.1 XM_011533267.1 XM_011533268.1 XM_011533270.1 XM_011533271.1 XM_011533273.1 XM_011533275.2 XM_017005531.1 XM_017005532.1 XM_017005534.1 XM_017005536.1 XM_017005537.1 XM_017005538.1 XM_017005539.1 XM_017005540.1 XM_017005541.1 NM_001123040.1

Gene Symbol
ARIH2OS
Entrez Gene ID
646450
Gene Name
ariadne homolog 2 opposite strand
Gene Aliases
C3orf71
UniGene
Hs.720727
Gene Chromosome Location
Chr.3: 48917788-48919385 on Build GRCh38
Transcripts
NM_001317333.1 NM_001317334.1 NM_006321.3 XM_011533265.1 XM_011533266.1 XM_011533267.1 XM_011533268.1 XM_011533270.1 XM_011533271.1 XM_011533273.1 XM_011533275.2 XM_017005531.1 XM_017005532.1 XM_017005534.1 XM_017005536.1 XM_017005537.1 XM_017005538.1 XM_017005539.1 XM_017005540.1 XM_017005541.1 NM_001123040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7178431 c.259G>A p.A87T Chr.3: 48918891-48918891 on Build GRCh38
3993136 c.492A>T p.P164P Chr.3: 48918658-48918658 on Build GRCh38
419960 c.256G>A p.D86N Chr.3: 48918894-48918894 on Build GRCh38
7122078 c.458C>G p.A153G Chr.3: 48918692-48918692 on Build GRCh38
3781649 c.406G>A p.A136T Chr.3: 48918744-48918744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759731586 Chr.3: 48918785-48918785 on Build GRCh38
rs745759736 Chr.3: 48918804-48918804 on Build GRCh38
rs777002472 Chr.3: 48918695-48918695 on Build GRCh38
rs763370255 Chr.3: 48918775-48918775 on Build GRCh38
rs758444385 Chr.3: 48918642-48918642 on Build GRCh38
rs751587028 Chr.3: 48918729-48918729 on Build GRCh38
rs774523567 Chr.3: 48918879-48918879 on Build GRCh38
rs570433496 Chr.3: 48918810-48918810 on Build GRCh38
rs770729764 Chr.3: 48918993-48918993 on Build GRCh38
rs747511473 Chr.3: 48918921-48918921 on Build GRCh38
rs900885911 Chr.3: 48918829-48918829 on Build GRCh38
rs770834980 Chr.3: 48918765-48918765 on Build GRCh38
rs777434968 Chr.3: 48918790-48918790 on Build GRCh38
rs758393486 Chr.3: 48918853-48918854 on Build GRCh38
rs764889998 Chr.3: 48918797-48918800 on Build GRCh38
rs747564744 Chr.3: 48918967-48918967 on Build GRCh38
rs950218329 Chr.3: 48918565-48918565 on Build GRCh38
rs150858838 Chr.3: 48918824-48918824 on Build GRCh38
rs754586401 Chr.3: 48918904-48918904 on Build GRCh38
rs761009053 Chr.3: 48918849-48918849 on Build GRCh38
rs778958161 Chr.3: 48918797-48918797 on Build GRCh38
rs770200020 Chr.3: 48918774-48918774 on Build GRCh38
rs957495001 Chr.3: 48918975-48918975 on Build GRCh38
rs369648980 Chr.3: 48918853-48918853 on Build GRCh38
rs749245700 Chr.3: 48918684-48918684 on Build GRCh38
rs997989727 Chr.3: 48918677-48918677 on Build GRCh38
rs747249161 Chr.3: 48918651-48918651 on Build GRCh38
rs766160744 Chr.3: 48918929-48918929 on Build GRCh38
rs762094897 Chr.3: 48918924-48918924 on Build GRCh38
rs755181299 Chr.3: 48918730-48918730 on Build GRCh38
rs781359004 Chr.3: 48918968-48918968 on Build GRCh38
rs369081967 Chr.3: 48918776-48918776 on Build GRCh38
rs759215580 Chr.3: 48918586-48918586 on Build GRCh38
rs754135090 Chr.3: 48918614-48918614 on Build GRCh38
rs763428699 Chr.3: 48918825-48918825 on Build GRCh38
rs764456047 Chr.3: 48918593-48918593 on Build GRCh38
rs376707348 Chr.3: 48918656-48918656 on Build GRCh38
rs768838738 Chr.3: 48918923-48918923 on Build GRCh38
rs376925252 Chr.3: 48918726-48918726 on Build GRCh38
rs891896270 Chr.3: 48918949-48918949 on Build GRCh38
rs760110889 Chr.3: 48918838-48918838 on Build GRCh38
rs752109280 Chr.3: 48918947-48918947 on Build GRCh38
rs1024737553 Chr.3: 48918942-48918942 on Build GRCh38
rs751471779 Chr.3: 48918789-48918789 on Build GRCh38
rs780557806 Chr.3: 48918913-48918913 on Build GRCh38
rs769818359 Chr.3: 48918554-48918554 on Build GRCh38
rs755655776 Chr.3: 48918914-48918914 on Build GRCh38
rs990333468 Chr.3: 48918976-48918976 on Build GRCh38
rs781618107 Chr.3: 48918732-48918732 on Build GRCh38
rs764048126 Chr.3: 48918889-48918889 on Build GRCh38
rs754796075 Chr.3: 48919009-48919010 on Build GRCh38
rs755166299 Chr.3: 48918653-48918653 on Build GRCh38
rs777698715 Chr.3: 48918762-48918762 on Build GRCh38
rs774069479 Chr.3: 48918998-48918998 on Build GRCh38
rs945036358 Chr.3: 48918823-48918823 on Build GRCh38
rs780552957 Chr.3: 48918961-48918961 on Build GRCh38
rs748917418 Chr.3: 48918957-48918957 on Build GRCh38
rs746822855 Chr.3: 48918861-48918861 on Build GRCh38
rs946298428 Chr.3: 48918997-48918997 on Build GRCh38
rs746478215 Chr.3: 48918864-48918864 on Build GRCh38
rs910951047 Chr.3: 48918595-48918595 on Build GRCh38
rs73082317 Chr.3: 48918616-48918616 on Build GRCh38
rs758271050 Chr.3: 48918862-48918862 on Build GRCh38
rs372287013 Chr.3: 48918926-48918926 on Build GRCh38
rs779539730 Chr.3: 48918863-48918863 on Build GRCh38
rs747753591 Chr.3: 48918874-48918874 on Build GRCh38
rs771547046 Chr.3: 48918767-48918767 on Build GRCh38
rs756667444 Chr.3: 48918836-48918836 on Build GRCh38
rs762420972 Chr.3: 48918572-48918572 on Build GRCh38
rs750356582 Chr.3: 48918803-48918803 on Build GRCh38
rs758360856 Chr.3: 48918902-48918902 on Build GRCh38
rs780175864 Chr.3: 48918649-48918649 on Build GRCh38
rs768134987 Chr.3: 48918865-48918865 on Build GRCh38
rs779452205 Chr.3: 48918915-48918915 on Build GRCh38
rs913497148 Chr.3: 48918821-48918821 on Build GRCh38
rs769692768 Chr.3: 48918669-48918669 on Build GRCh38
rs780942346 Chr.3: 48918869-48918869 on Build GRCh38
rs767308189 Chr.3: 48919010-48919010 on Build GRCh38
rs756348549 Chr.3: 48918788-48918788 on Build GRCh38
rs765334565 Chr.3: 48918925-48918925 on Build GRCh38
rs1053443789 Chr.3: 48918627-48918627 on Build GRCh38
rs772544492 Chr.3: 48918920-48918920 on Build GRCh38
rs372170656 Chr.3: 48918570-48918570 on Build GRCh38
rs900935371 Chr.3: 48918663-48918663 on Build GRCh38
rs201154437 Chr.3: 48918860-48918860 on Build GRCh38
rs973159054 Chr.3: 48918773-48918773 on Build GRCh38
rs536644096 Chr.3: 48918987-48918987 on Build GRCh38
rs754818427 Chr.3: 48918933-48918933 on Build GRCh38
rs893517847 Chr.3: 48918644-48918644 on Build GRCh38
rs371737087 Chr.3: 48918781-48918781 on Build GRCh38
rs745333408 Chr.3: 48918766-48918766 on Build GRCh38
rs369699183 Chr.3: 48918878-48918878 on Build GRCh38
rs746383210 Chr.3: 48918880-48918880 on Build GRCh38
rs369121404 Chr.3: 48918805-48918805 on Build GRCh38
rs750147597 Chr.3: 48918899-48918899 on Build GRCh38
rs201866859 Chr.3: 48918678-48918678 on Build GRCh38
rs781759206 Chr.3: 48918827-48918827 on Build GRCh38
rs767703547 Chr.3: 48918786-48918786 on Build GRCh38
rs756042992 Chr.3: 48918748-48918748 on Build GRCh38
rs746188675 Chr.3: 48918919-48918919 on Build GRCh38
rs773854719 Chr.3: 48918688-48918688 on Build GRCh38
rs763150173 Chr.3: 48918989-48918989 on Build GRCh38
rs757115126 Chr.3: 48918856-48918856 on Build GRCh38
rs772667338 Chr.3: 48918881-48918881 on Build GRCh38
rs750415243 Chr.3: 48918861-48918861 on Build GRCh38
rs546855001 Chr.3: 48918852-48918852 on Build GRCh38
rs765885209 Chr.3: 48918575-48918575 on Build GRCh38
rs751559608 Chr.3: 48918931-48918931 on Build GRCh38
rs779523412 Chr.3: 48918806-48918806 on Build GRCh38
rs775734914 Chr.3: 48918882-48918882 on Build GRCh38
rs752760678 Chr.3: 48918846-48918846 on Build GRCh38
rs761926279 Chr.3: 48918894-48918894 on Build GRCh38
rs748125823 Chr.3: 48918970-48918970 on Build GRCh38
rs775162444 Chr.3: 48918768-48918768 on Build GRCh38
rs762779199 Chr.3: 48918928-48918928 on Build GRCh38
rs770407318 Chr.3: 48918687-48918687 on Build GRCh38
rs765553268 Chr.3: 48918696-48918696 on Build GRCh38
rs769762472 Chr.3: 48919007-48919007 on Build GRCh38
rs769838601 Chr.3: 48918973-48918973 on Build GRCh38
rs751992576 Chr.3: 48918782-48918782 on Build GRCh38
rs767060066 Chr.3: 48918943-48918943 on Build GRCh38
rs971891801 Chr.3: 48918780-48918780 on Build GRCh38
rs750906766 Chr.3: 48918640-48918640 on Build GRCh38
rs779654273 Chr.3: 48918861-48918862 on Build GRCh38
rs749198952 Chr.3: 48918764-48918764 on Build GRCh38
rs965156948 Chr.3: 48918844-48918844 on Build GRCh38
rs372570123 Chr.3: 48918953-48918953 on Build GRCh38
rs757792584 Chr.3: 48918615-48918615 on Build GRCh38
rs879431590 Chr.3: 48918984-48918984 on Build GRCh38
rs368509662 Chr.3: 48918693-48918693 on Build GRCh38
rs376377066 Chr.3: 48918794-48918794 on Build GRCh38
rs1040704821 Chr.3: 48918655-48918655 on Build GRCh38
rs748351511 Chr.3: 48918742-48918742 on Build GRCh38
rs781168619 Chr.3: 48918654-48918654 on Build GRCh38
rs377462237 Chr.3: 48918830-48918830 on Build GRCh38
rs759550722 Chr.3: 48919005-48919005 on Build GRCh38
rs906077266 Chr.3: 48918589-48918589 on Build GRCh38
rs200347138 Chr.3: 48918692-48918692 on Build GRCh38
rs751239325 Chr.3: 48918585-48918585 on Build GRCh38
rs760312536 Chr.3: 48918769-48918769 on Build GRCh38
rs550168944 Chr.3: 48918954-48918954 on Build GRCh38
rs776977816 Chr.3: 48918892-48918892 on Build GRCh38
rs773326381 Chr.3: 48918682-48918682 on Build GRCh38
rs918289311 Chr.3: 48918690-48918690 on Build GRCh38
rs1035781743 Chr.3: 48918793-48918793 on Build GRCh38
rs372379639 Chr.3: 48918712-48918712 on Build GRCh38
rs765089859 Chr.3: 48918895-48918895 on Build GRCh38

View Full Product Details