Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49419178-49419679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMT
Entrez Gene ID
275
Gene Name
aminomethyltransferase
Gene Aliases
GCE, GCST, GCVT, NKH
UniGene
Hs.102
Gene Chromosome Location
Chr.3: 49416778-49422678 on Build GRCh38
Transcripts
NM_000481.3 NM_001164710.1 NM_001164711.1 NM_001164712.1 NR_028435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4888870 c.551G>T p.G184V Chr.3: 49419405-49419405 on Build GRCh38
6230531 c.657C>T p.R219R Chr.3: 49419299-49419299 on Build GRCh38
6599475 c.658G>A p.V220M Chr.3: 49419298-49419298 on Build GRCh38
1046151 c.587C>T p.A196V Chr.3: 49419369-49419369 on Build GRCh38
5548957 c.559G>A p.A187T Chr.3: 49419397-49419397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778728514 Chr.3: 49419386-49419386 on Build GRCh38
rs140250976 Chr.3: 49419322-49419322 on Build GRCh38
rs116192290 Chr.3: 49419325-49419325 on Build GRCh38
rs754332895 Chr.3: 49419231-49419231 on Build GRCh38
rs138276307 Chr.3: 49419214-49419214 on Build GRCh38
rs750787542 Chr.3: 49419250-49419250 on Build GRCh38
rs201141125 Chr.3: 49419321-49419321 on Build GRCh38
rs937463808 Chr.3: 49419411-49419411 on Build GRCh38
rs562695274 Chr.3: 49419291-49419291 on Build GRCh38
rs775019756 Chr.3: 49419414-49419414 on Build GRCh38
rs116637858 Chr.3: 49419538-49419538 on Build GRCh38
rs762408187 Chr.3: 49419504-49419504 on Build GRCh38
rs755978333 Chr.3: 49419329-49419329 on Build GRCh38
rs1030529954 Chr.3: 49419545-49419545 on Build GRCh38
rs780474627 Chr.3: 49419506-49419507 on Build GRCh38
rs770272854 Chr.3: 49419425-49419425 on Build GRCh38
rs929879659 Chr.3: 49419369-49419369 on Build GRCh38
rs760119523 Chr.3: 49419416-49419416 on Build GRCh38
rs781466698 Chr.3: 49419292-49419292 on Build GRCh38
rs747298727 Chr.3: 49419289-49419289 on Build GRCh38
rs751804881 Chr.3: 49419349-49419349 on Build GRCh38
rs762345907 Chr.3: 49419323-49419323 on Build GRCh38
rs766893584 Chr.3: 49419540-49419540 on Build GRCh38
rs763599694 Chr.3: 49419447-49419447 on Build GRCh38
rs1020520820 Chr.3: 49419254-49419254 on Build GRCh38
rs758602446 Chr.3: 49419251-49419251 on Build GRCh38
rs773791854 Chr.3: 49419218-49419218 on Build GRCh38
rs1011549926 Chr.3: 49419526-49419526 on Build GRCh38
rs766904475 Chr.3: 49419343-49419343 on Build GRCh38
rs962354821 Chr.3: 49419653-49419653 on Build GRCh38
rs202043041 Chr.3: 49419300-49419300 on Build GRCh38
rs773590861 Chr.3: 49419431-49419431 on Build GRCh38
rs770700817 Chr.3: 49419385-49419385 on Build GRCh38
rs1000277859 Chr.3: 49419602-49419602 on Build GRCh38
rs541594122 Chr.3: 49419258-49419258 on Build GRCh38
rs201085273 Chr.3: 49419215-49419215 on Build GRCh38
rs767083373 Chr.3: 49419230-49419230 on Build GRCh38
rs1041995667 Chr.3: 49419537-49419537 on Build GRCh38
rs62261214 Chr.3: 49419565-49419565 on Build GRCh38
rs150469803 Chr.3: 49419213-49419213 on Build GRCh38
rs534625484 Chr.3: 49419604-49419604 on Build GRCh38
rs121964986 Chr.3: 49419382-49419382 on Build GRCh38
rs568027863 Chr.3: 49419621-49419621 on Build GRCh38
rs976237396 Chr.3: 49419299-49419299 on Build GRCh38
rs766816589 Chr.3: 49419454-49419454 on Build GRCh38
rs749047993 Chr.3: 49419306-49419306 on Build GRCh38
rs199638653 Chr.3: 49419298-49419298 on Build GRCh38
rs1020178363 Chr.3: 49419569-49419569 on Build GRCh38
rs143984123 Chr.3: 49419610-49419610 on Build GRCh38
rs1037672208 Chr.3: 49419483-49419483 on Build GRCh38
rs386833689 Chr.3: 49419282-49419282 on Build GRCh38
rs973966471 Chr.3: 49419344-49419344 on Build GRCh38
rs200550585 Chr.3: 49419367-49419367 on Build GRCh38
rs773990204 Chr.3: 49419308-49419308 on Build GRCh38
rs749940667 Chr.3: 49419495-49419495 on Build GRCh38
rs1030915796 Chr.3: 49419313-49419313 on Build GRCh38
rs186390975 Chr.3: 49419233-49419233 on Build GRCh38
rs551893496 Chr.3: 49419478-49419478 on Build GRCh38
rs953654435 Chr.3: 49419547-49419547 on Build GRCh38
rs765606234 Chr.3: 49419245-49419245 on Build GRCh38
rs937481466 Chr.3: 49419520-49419520 on Build GRCh38
rs372496075 Chr.3: 49419373-49419373 on Build GRCh38
rs755288353 Chr.3: 49419355-49419355 on Build GRCh38
rs752740839 Chr.3: 49419372-49419372 on Build GRCh38
rs903069991 Chr.3: 49419576-49419576 on Build GRCh38
rs771745221 Chr.3: 49419404-49419404 on Build GRCh38
rs144790394 Chr.3: 49419401-49419401 on Build GRCh38
rs749313171 Chr.3: 49419377-49419377 on Build GRCh38
rs546353720 Chr.3: 49419606-49419606 on Build GRCh38
rs963795935 Chr.3: 49419318-49419318 on Build GRCh38
rs933458798 Chr.3: 49419352-49419352 on Build GRCh38
rs372864975 Chr.3: 49419199-49419199 on Build GRCh38
rs759245586 Chr.3: 49419314-49419314 on Build GRCh38
rs767552814 Chr.3: 49419199-49419199 on Build GRCh38
rs377552553 Chr.3: 49419374-49419374 on Build GRCh38
rs576365400 Chr.3: 49419200-49419200 on Build GRCh38
rs769605378 Chr.3: 49419202-49419202 on Build GRCh38
rs770502702 Chr.3: 49419307-49419307 on Build GRCh38
rs759050945 Chr.3: 49419228-49419228 on Build GRCh38
rs752108129 Chr.3: 49419615-49419615 on Build GRCh38
rs150079386 Chr.3: 49419301-49419301 on Build GRCh38
rs370287514 Chr.3: 49419331-49419331 on Build GRCh38
rs1006652556 Chr.3: 49419613-49419613 on Build GRCh38
rs758841205 Chr.3: 49419340-49419340 on Build GRCh38

View Full Product Details