Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49422243-49422732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMT
Entrez Gene ID
275
Gene Name
aminomethyltransferase
Gene Aliases
GCE, GCST, GCVT, NKH
UniGene
Hs.102
Gene Chromosome Location
Chr.3: 49416778-49422678 on Build GRCh38
Transcripts
NM_000481.3 NM_001164710.1 NM_001164711.1 NM_001164712.1 NR_028435.1 NM_032316.3

Gene Symbol
NICN1
Entrez Gene ID
84276
Gene Name
nicolin 1
Gene Aliases
-
UniGene
Hs.191460
Gene Chromosome Location
Chr.3: 49422333-49429324 on Build GRCh38
Transcripts
NM_000481.3 NM_001164710.1 NM_001164711.1 NM_001164712.1 NR_028435.1 NM_032316.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1423947 c.59C>T p.P20L Chr.3: 49422392-49422392 on Build GRCh38
4118498 c.29G>A p.R10H Chr.3: 49422422-49422422 on Build GRCh38
6599474 c.91-10C>T p.? Chr.3: 49422281-49422281 on Build GRCh38
5399595 c.67T>A p.C23S Chr.3: 49422384-49422384 on Build GRCh38
4118497 c.53T>C p.F18S Chr.3: 49422398-49422398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770510324 Chr.3: 49422391-49422392 on Build GRCh38
rs573551723 Chr.3: 49422656-49422656 on Build GRCh38
rs377597372 Chr.3: 49422318-49422318 on Build GRCh38
rs761235679 Chr.3: 49422280-49422280 on Build GRCh38
rs773139530 Chr.3: 49422575-49422586 on Build GRCh38
rs148166670 Chr.3: 49422463-49422463 on Build GRCh38
rs192377809 Chr.3: 49422550-49422550 on Build GRCh38
rs749087037 Chr.3: 49422392-49422392 on Build GRCh38
rs763223038 Chr.3: 49422367-49422367 on Build GRCh38
rs555476786 Chr.3: 49422652-49422652 on Build GRCh38
rs751869343 Chr.3: 49422378-49422378 on Build GRCh38
rs375632843 Chr.3: 49422279-49422279 on Build GRCh38
rs944024742 Chr.3: 49422611-49422611 on Build GRCh38
rs759187553 Chr.3: 49422573-49422576 on Build GRCh38
rs746730300 Chr.3: 49422547-49422547 on Build GRCh38
rs201779973 Chr.3: 49422411-49422411 on Build GRCh38
rs771599621 Chr.3: 49422554-49422554 on Build GRCh38
rs759639271 Chr.3: 49422495-49422495 on Build GRCh38
rs202052678 Chr.3: 49422317-49422317 on Build GRCh38
rs1008774394 Chr.3: 49422647-49422647 on Build GRCh38
rs765489264 Chr.3: 49422572-49422572 on Build GRCh38
rs745598809 Chr.3: 49422402-49422402 on Build GRCh38
rs1023508368 Chr.3: 49422665-49422665 on Build GRCh38
rs886058684 Chr.3: 49422566-49422567 on Build GRCh38
rs750260933 Chr.3: 49422533-49422533 on Build GRCh38
rs757568116 Chr.3: 49422534-49422534 on Build GRCh38
rs768697561 Chr.3: 49422587-49422587 on Build GRCh38
rs755011710 Chr.3: 49422379-49422379 on Build GRCh38
rs765668315 Chr.3: 49422348-49422348 on Build GRCh38
rs386833686 Chr.3: 49422392-49422392 on Build GRCh38
rs974641286 Chr.3: 49422309-49422309 on Build GRCh38
rs923667012 Chr.3: 49422299-49422299 on Build GRCh38
rs746258766 Chr.3: 49422560-49422560 on Build GRCh38
rs1048942270 Chr.3: 49422645-49422645 on Build GRCh38
rs150649086 Chr.3: 49422422-49422422 on Build GRCh38
rs546936770 Chr.3: 49422329-49422329 on Build GRCh38
rs374750709 Chr.3: 49422391-49422391 on Build GRCh38
rs762806582 Chr.3: 49422678-49422678 on Build GRCh38
rs907712325 Chr.3: 49422524-49422524 on Build GRCh38
rs764224916 Chr.3: 49422507-49422507 on Build GRCh38
rs764624214 Chr.3: 49422373-49422373 on Build GRCh38
rs533658788 Chr.3: 49422651-49422651 on Build GRCh38
rs897142645 Chr.3: 49422636-49422636 on Build GRCh38
rs745484929 Chr.3: 49422452-49422452 on Build GRCh38
rs1028631130 Chr.3: 49422689-49422689 on Build GRCh38
rs771681000 Chr.3: 49422331-49422331 on Build GRCh38
rs753743263 Chr.3: 49422508-49422508 on Build GRCh38
rs1026854868 Chr.3: 49422470-49422470 on Build GRCh38
rs1045119115 Chr.3: 49422573-49422573 on Build GRCh38
rs143841175 Chr.3: 49422451-49422451 on Build GRCh38
rs759853191 Chr.3: 49422429-49422429 on Build GRCh38
rs544461335 Chr.3: 49422526-49422526 on Build GRCh38
rs758737016 Chr.3: 49422278-49422278 on Build GRCh38
rs765222298 Chr.3: 49422606-49422606 on Build GRCh38
rs771403409 Chr.3: 49422482-49422482 on Build GRCh38
rs773793813 Chr.3: 49422328-49422328 on Build GRCh38
rs775112005 Chr.3: 49422408-49422408 on Build GRCh38
rs748551244 Chr.3: 49422410-49422410 on Build GRCh38
rs572578341 Chr.3: 49422576-49422576 on Build GRCh38
rs757232514 Chr.3: 49422522-49422522 on Build GRCh38
rs921596268 Chr.3: 49422486-49422486 on Build GRCh38
rs995861547 Chr.3: 49422679-49422679 on Build GRCh38
rs1001144470 Chr.3: 49422591-49422591 on Build GRCh38
rs779766109 Chr.3: 49422546-49422546 on Build GRCh38
rs767785767 Chr.3: 49422500-49422500 on Build GRCh38
rs142950858 Chr.3: 49422380-49422380 on Build GRCh38
rs762434060 Chr.3: 49422343-49422343 on Build GRCh38
rs775374540 Chr.3: 49422504-49422504 on Build GRCh38
rs934826873 Chr.3: 49422323-49422323 on Build GRCh38
rs749854761 Chr.3: 49422479-49422479 on Build GRCh38
rs139941796 Chr.3: 49422416-49422416 on Build GRCh38
rs767448950 Chr.3: 49422430-49422430 on Build GRCh38
rs577999655 Chr.3: 49422705-49422705 on Build GRCh38
rs1041034700 Chr.3: 49422625-49422625 on Build GRCh38
rs911454395 Chr.3: 49422604-49422604 on Build GRCh38
rs753829515 Chr.3: 49422441-49422441 on Build GRCh38
rs776992801 Chr.3: 49422342-49422342 on Build GRCh38
rs369384839 Chr.3: 49422478-49422478 on Build GRCh38
rs750502117 Chr.3: 49422586-49422586 on Build GRCh38
rs930561369 Chr.3: 49422336-49422336 on Build GRCh38
rs774408322 Chr.3: 49422423-49422423 on Build GRCh38
rs386833687 Chr.3: 49422390-49422390 on Build GRCh38
rs761696027 Chr.3: 49422602-49422602 on Build GRCh38
rs748898213 Chr.3: 49422326-49422326 on Build GRCh38
rs780984623 Chr.3: 49422310-49422310 on Build GRCh38
rs779082226 Chr.3: 49422556-49422556 on Build GRCh38
rs771449741 Chr.3: 49422404-49422404 on Build GRCh38
rs757040449 Chr.3: 49422442-49422442 on Build GRCh38
rs778523705 Chr.3: 49422399-49422399 on Build GRCh38
rs770512923 Chr.3: 49422394-49422394 on Build GRCh38
rs544232394 Chr.3: 49422657-49422657 on Build GRCh38
rs914716059 Chr.3: 49422397-49422397 on Build GRCh38
rs386833677 Chr.3: 49422505-49422505 on Build GRCh38
rs779387596 Chr.3: 49422462-49422462 on Build GRCh38
rs758088553 Chr.3: 49422455-49422455 on Build GRCh38
rs773988915 Chr.3: 49422436-49422437 on Build GRCh38
rs776711848 Chr.3: 49422590-49422590 on Build GRCh38
rs756101487 Chr.3: 49422389-49422389 on Build GRCh38
rs371656220 Chr.3: 49422381-49422381 on Build GRCh38
rs930117335 Chr.3: 49422642-49422642 on Build GRCh38
rs562599479 Chr.3: 49422683-49422683 on Build GRCh38
rs568348276 Chr.3: 49422364-49422364 on Build GRCh38
rs1019077308 Chr.3: 49422476-49422476 on Build GRCh38
rs943991980 Chr.3: 49422439-49422439 on Build GRCh38
rs748956297 Chr.3: 49422371-49422371 on Build GRCh38
rs768343793 Chr.3: 49422473-49422473 on Build GRCh38
rs906423906 Chr.3: 49422669-49422669 on Build GRCh38
rs774803281 Chr.3: 49422494-49422494 on Build GRCh38
rs764032357 Chr.3: 49422438-49422438 on Build GRCh38
rs1053088222 Chr.3: 49422592-49422592 on Build GRCh38
rs143389339 Chr.3: 49422262-49422262 on Build GRCh38
rs750654817 Chr.3: 49422267-49422267 on Build GRCh38
rs754874614 Chr.3: 49422285-49422285 on Build GRCh38
rs377647344 Chr.3: 49422477-49422477 on Build GRCh38
rs772378099 Chr.3: 49422575-49422575 on Build GRCh38
rs1012146160 Chr.3: 49422490-49422490 on Build GRCh38
rs892644956 Chr.3: 49422622-49422622 on Build GRCh38
rs749765968 Chr.3: 49422553-49422553 on Build GRCh38
rs386833688 Chr.3: 49422388-49422388 on Build GRCh38
rs1048913285 Chr.3: 49422356-49422356 on Build GRCh38
rs755979959 Chr.3: 49422433-49422433 on Build GRCh38
rs752853937 Chr.3: 49422432-49422432 on Build GRCh38

View Full Product Details