Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49470341-49470877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAG1
Entrez Gene ID
1605
Gene Name
dystroglycan 1
Gene Aliases
156DAG, A3a, AGRNR, DAG, MDDGA9, MDDGC7, MDDGC9
UniGene
Hs.76111
Gene Chromosome Location
Chr.3: 49468703-49535618 on Build GRCh38
Transcripts
NM_001165928.3 NM_001177634.2 NM_001177635.2 NM_001177636.2 NM_001177637.2 NM_001177638.2 NM_001177639.2 NM_001177640.2 NM_001177641.2 NM_001177642.2 NM_001177643.2 NM_001177644.2 NM_004393.5

SNP Detail

SNP ID Chromosome Location
rs889909616 Chr.3: 49470409-49470409 on Build GRCh38
rs959117154 Chr.3: 49470575-49470575 on Build GRCh38
rs994397800 Chr.3: 49470481-49470481 on Build GRCh38
rs917237628 Chr.3: 49470383-49470383 on Build GRCh38
rs1054240226 Chr.3: 49470754-49470754 on Build GRCh38
rs1029771745 Chr.3: 49470513-49470513 on Build GRCh38
rs890790494 Chr.3: 49470528-49470528 on Build GRCh38
rs914927403 Chr.3: 49470583-49470583 on Build GRCh38
rs4855839 Chr.3: 49470414-49470414 on Build GRCh38
rs897123752 Chr.3: 49470478-49470478 on Build GRCh38
rs966433808 Chr.3: 49470845-49470845 on Build GRCh38
rs1042673073 Chr.3: 49470370-49470370 on Build GRCh38
rs897176102 Chr.3: 49470696-49470696 on Build GRCh38
rs1022084813 Chr.3: 49470834-49470834 on Build GRCh38
rs759816945 Chr.3: 49470793-49470793 on Build GRCh38
rs1010672171 Chr.3: 49470830-49470830 on Build GRCh38
rs898226708 Chr.3: 49470832-49470832 on Build GRCh38
rs922535651 Chr.3: 49470625-49470625 on Build GRCh38
rs1016898456 Chr.3: 49470420-49470420 on Build GRCh38
rs950205720 Chr.3: 49470384-49470384 on Build GRCh38
rs971653887 Chr.3: 49470503-49470503 on Build GRCh38
rs183558916 Chr.3: 49470785-49470785 on Build GRCh38
rs1034514975 Chr.3: 49470539-49470539 on Build GRCh38
rs899731601 Chr.3: 49470475-49470475 on Build GRCh38
rs553384753 Chr.3: 49470674-49470674 on Build GRCh38
rs1051020687 Chr.3: 49470792-49470792 on Build GRCh38
rs767715914 Chr.3: 49470856-49470856 on Build GRCh38
rs577533019 Chr.3: 49470624-49470624 on Build GRCh38
rs889583864 Chr.3: 49470664-49470664 on Build GRCh38
rs989187479 Chr.3: 49470578-49470578 on Build GRCh38
rs962214652 Chr.3: 49470615-49470615 on Build GRCh38
rs1015305785 Chr.3: 49470540-49470540 on Build GRCh38
rs944994036 Chr.3: 49470598-49470598 on Build GRCh38
rs977774369 Chr.3: 49470608-49470608 on Build GRCh38
rs1050061250 Chr.3: 49470627-49470627 on Build GRCh38
rs1054308235 Chr.3: 49470459-49470459 on Build GRCh38
rs991741484 Chr.3: 49470672-49470672 on Build GRCh38
rs971418539 Chr.3: 49470688-49470688 on Build GRCh38
rs964158413 Chr.3: 49470405-49470405 on Build GRCh38
rs980716950 Chr.3: 49470508-49470508 on Build GRCh38
rs903063283 Chr.3: 49470416-49470416 on Build GRCh38
rs1025005351 Chr.3: 49470488-49470488 on Build GRCh38
rs973542149 Chr.3: 49470653-49470653 on Build GRCh38
rs994151219 Chr.3: 49470723-49470723 on Build GRCh38
rs937340571 Chr.3: 49470368-49470368 on Build GRCh38
rs933848224 Chr.3: 49470396-49470396 on Build GRCh38
rs1027925231 Chr.3: 49470670-49470670 on Build GRCh38
rs907482061 Chr.3: 49470731-49470731 on Build GRCh38
rs933923045 Chr.3: 49470626-49470626 on Build GRCh38
rs752420656 Chr.3: 49470761-49470761 on Build GRCh38
rs1049564776 Chr.3: 49470406-49470406 on Build GRCh38
rs901021228 Chr.3: 49470392-49470392 on Build GRCh38
rs935849413 Chr.3: 49470730-49470730 on Build GRCh38
rs28364662 Chr.3: 49470818-49470818 on Build GRCh38
rs991872605 Chr.3: 49470381-49470381 on Build GRCh38
rs1024306047 Chr.3: 49470728-49470728 on Build GRCh38
rs924664692 Chr.3: 49470697-49470697 on Build GRCh38
rs538278341 Chr.3: 49470671-49470671 on Build GRCh38
rs1038395426 Chr.3: 49470685-49470685 on Build GRCh38

View Full Product Details