Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49488535-49489044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAG1
Entrez Gene ID
1605
Gene Name
dystroglycan 1
Gene Aliases
156DAG, A3a, AGRNR, DAG, MDDGA9, MDDGC7, MDDGC9
UniGene
Hs.76111
Gene Chromosome Location
Chr.3: 49468703-49535618 on Build GRCh38
Transcripts
NM_001165928.3 NM_001177634.2 NM_001177635.2 NM_001177636.2 NM_001177637.2 NM_001177638.2 NM_001177639.2 NM_001177640.2 NM_001177641.2 NM_001177642.2 NM_001177643.2 NM_001177644.2 NM_004393.5

SNP Detail

SNP ID Chromosome Location
rs925166059 Chr.3: 49488749-49488749 on Build GRCh38
rs985928448 Chr.3: 49488613-49488613 on Build GRCh38
rs1011764240 Chr.3: 49488752-49488752 on Build GRCh38
rs571015514 Chr.3: 49488572-49488572 on Build GRCh38
rs557118214 Chr.3: 49488705-49488705 on Build GRCh38
rs938241888 Chr.3: 49489015-49489015 on Build GRCh38
rs140896147 Chr.3: 49488666-49488666 on Build GRCh38
rs370501826 Chr.3: 49488942-49488942 on Build GRCh38
rs940470653 Chr.3: 49488816-49488816 on Build GRCh38
rs539844207 Chr.3: 49488607-49488607 on Build GRCh38
rs573053146 Chr.3: 49488934-49488934 on Build GRCh38
rs978569251 Chr.3: 49488747-49488747 on Build GRCh38
rs942089251 Chr.3: 49488622-49488622 on Build GRCh38
rs974573105 Chr.3: 49488756-49488756 on Build GRCh38
rs759418323 Chr.3: 49488872-49488872 on Build GRCh38
rs933199731 Chr.3: 49488792-49488792 on Build GRCh38
rs535525853 Chr.3: 49488668-49488668 on Build GRCh38
rs768736034 Chr.3: 49488777-49488777 on Build GRCh38
rs749250772 Chr.3: 49488793-49488793 on Build GRCh38
rs4855860 Chr.3: 49488611-49488611 on Build GRCh38
rs150155082 Chr.3: 49488850-49488850 on Build GRCh38
rs4855859 Chr.3: 49488867-49488867 on Build GRCh38
rs528783400 Chr.3: 49488600-49488600 on Build GRCh38
rs1030262344 Chr.3: 49488839-49488839 on Build GRCh38
rs754000818 Chr.3: 49488608-49488609 on Build GRCh38
rs963231753 Chr.3: 49488862-49488862 on Build GRCh38
rs972590891 Chr.3: 49488863-49488863 on Build GRCh38
rs78452073 Chr.3: 49488571-49488571 on Build GRCh38
rs907538349 Chr.3: 49488797-49488797 on Build GRCh38
rs568992067 Chr.3: 49488732-49488732 on Build GRCh38
rs933000135 Chr.3: 49488601-49488604 on Build GRCh38
rs995794918 Chr.3: 49488619-49488619 on Build GRCh38
rs58775733 Chr.3: 49488588-49488588 on Build GRCh38
rs898889498 Chr.3: 49488829-49488829 on Build GRCh38
rs189857561 Chr.3: 49488562-49488562 on Build GRCh38
rs752475295 Chr.3: 49488607-49488608 on Build GRCh38
rs138596149 Chr.3: 49488972-49488972 on Build GRCh38
rs919282585 Chr.3: 49488662-49488662 on Build GRCh38
rs949387577 Chr.3: 49488871-49488871 on Build GRCh38
rs779176872 Chr.3: 49488663-49488663 on Build GRCh38
rs776621959 Chr.3: 49488778-49488778 on Build GRCh38
rs1037837753 Chr.3: 49488820-49488820 on Build GRCh38
rs78095107 Chr.3: 49488621-49488621 on Build GRCh38
rs767063394 Chr.3: 49488608-49488608 on Build GRCh38
rs74780556 Chr.3: 49488620-49488620 on Build GRCh38
rs898643868 Chr.3: 49488614-49488614 on Build GRCh38
rs974983324 Chr.3: 49489014-49489014 on Build GRCh38
rs200848559 Chr.3: 49488611-49488611 on Build GRCh38
rs923181856 Chr.3: 49488610-49488610 on Build GRCh38
rs372635971 Chr.3: 49488626-49488626 on Build GRCh38
rs999510915 Chr.3: 49488960-49488960 on Build GRCh38
rs865860970 Chr.3: 49488794-49488794 on Build GRCh38
rs957313330 Chr.3: 49488565-49488565 on Build GRCh38
rs909064818 Chr.3: 49488855-49488855 on Build GRCh38
rs986001861 Chr.3: 49488853-49488853 on Build GRCh38
rs868729084 Chr.3: 49488779-49488779 on Build GRCh38
rs997146884 Chr.3: 49488796-49488796 on Build GRCh38
rs368823877 Chr.3: 49488568-49488568 on Build GRCh38
rs747219327 Chr.3: 49488750-49488750 on Build GRCh38
rs1032854382 Chr.3: 49488964-49488964 on Build GRCh38
rs577128212 Chr.3: 49488594-49488594 on Build GRCh38

View Full Product Details