Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49492822-49493371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAG1
Entrez Gene ID
1605
Gene Name
dystroglycan 1
Gene Aliases
156DAG, A3a, AGRNR, DAG, MDDGA9, MDDGC7, MDDGC9
UniGene
Hs.76111
Gene Chromosome Location
Chr.3: 49468703-49535618 on Build GRCh38
Transcripts
NM_001165928.3 NM_001177634.2 NM_001177635.2 NM_001177636.2 NM_001177637.2 NM_001177638.2 NM_001177639.2 NM_001177640.2 NM_001177641.2 NM_001177642.2 NM_001177643.2 NM_001177644.2 NM_004393.5

SNP Detail

SNP ID Chromosome Location
rs368114748 Chr.3: 49493033-49493033 on Build GRCh38
rs932345009 Chr.3: 49493127-49493127 on Build GRCh38
rs72166671 Chr.3: 49493059-49493060 on Build GRCh38
rs543190151 Chr.3: 49493328-49493328 on Build GRCh38
rs927434954 Chr.3: 49493204-49493204 on Build GRCh38
rs548016647 Chr.3: 49492883-49492883 on Build GRCh38
rs77632881 Chr.3: 49493036-49493036 on Build GRCh38
rs998099421 Chr.3: 49492904-49492904 on Build GRCh38
rs1019150474 Chr.3: 49492988-49492988 on Build GRCh38
rs796503174 Chr.3: 49492956-49492956 on Build GRCh38
rs550138034 Chr.3: 49493064-49493064 on Build GRCh38
rs55708660 Chr.3: 49493059-49493059 on Build GRCh38
rs1036897180 Chr.3: 49493325-49493325 on Build GRCh38
rs575724933 Chr.3: 49493312-49493312 on Build GRCh38
rs539297899 Chr.3: 49493123-49493123 on Build GRCh38
rs909822282 Chr.3: 49493236-49493236 on Build GRCh38
rs571585803 Chr.3: 49493069-49493069 on Build GRCh38
rs1025753823 Chr.3: 49492906-49492906 on Build GRCh38
rs766916125 Chr.3: 49492891-49492891 on Build GRCh38
rs905865947 Chr.3: 49492881-49492881 on Build GRCh38
rs538387689 Chr.3: 49492919-49492919 on Build GRCh38
rs572418548 Chr.3: 49493184-49493184 on Build GRCh38
rs895689551 Chr.3: 49493344-49493344 on Build GRCh38
rs777944276 Chr.3: 49493306-49493306 on Build GRCh38
rs764541162 Chr.3: 49493220-49493220 on Build GRCh38
rs913443940 Chr.3: 49493269-49493269 on Build GRCh38
rs779153522 Chr.3: 49493285-49493285 on Build GRCh38
rs767858640 Chr.3: 49493261-49493261 on Build GRCh38
rs376471222 Chr.3: 49492893-49492893 on Build GRCh38
rs776108853 Chr.3: 49492938-49492938 on Build GRCh38
rs7643845 Chr.3: 49493278-49493278 on Build GRCh38
rs966600775 Chr.3: 49493037-49493037 on Build GRCh38
rs9856102 Chr.3: 49493297-49493297 on Build GRCh38
rs79189592 Chr.3: 49493038-49493038 on Build GRCh38
rs969124679 Chr.3: 49493193-49493193 on Build GRCh38
rs983725454 Chr.3: 49493173-49493173 on Build GRCh38
rs554140408 Chr.3: 49493131-49493131 on Build GRCh38
rs759958483 Chr.3: 49493118-49493118 on Build GRCh38
rs1057393689 Chr.3: 49493250-49493250 on Build GRCh38
rs946075198 Chr.3: 49493271-49493271 on Build GRCh38
rs752028409 Chr.3: 49492939-49492939 on Build GRCh38
rs561096963 Chr.3: 49493309-49493309 on Build GRCh38
rs536405536 Chr.3: 49493277-49493277 on Build GRCh38
rs756228823 Chr.3: 49493305-49493305 on Build GRCh38

View Full Product Details