Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49675964-49676467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APEH
Entrez Gene ID
327
Gene Name
acylaminoacyl-peptide hydrolase
Gene Aliases
AARE, ACPH, APH, D3F15S2, D3S48E, DNF15S2, OPH
UniGene
Hs.517969
Gene Chromosome Location
Chr.3: 49673102-49683508 on Build GRCh38
Transcripts
NM_001640.3 XM_005265097.2 XM_005265098.4 XM_011533658.2 XM_011533659.1 XM_011533660.2 XM_011533661.2 XM_011533662.2 XM_011533663.2 XM_017006285.1 XM_017006286.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1180958 c.607-3_607-2insA p.? Chr.3: 49676375-49676376 on Build GRCh38
6602122 c.468G>A p.S156S Chr.3: 49676081-49676081 on Build GRCh38
731255 c.585G>A p.K195K Chr.3: 49676198-49676198 on Build GRCh38
6602124 c.509G>A p.R170H Chr.3: 49676122-49676122 on Build GRCh38
4941697 c.515A>G p.K172R Chr.3: 49676128-49676128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926895962 Chr.3: 49676118-49676118 on Build GRCh38
rs1051443146 Chr.3: 49676024-49676024 on Build GRCh38
rs371501935 Chr.3: 49676226-49676226 on Build GRCh38
rs543774757 Chr.3: 49676283-49676283 on Build GRCh38
rs779408033 Chr.3: 49676228-49676228 on Build GRCh38
rs751567862 Chr.3: 49676384-49676384 on Build GRCh38
rs779911863 Chr.3: 49676042-49676042 on Build GRCh38
rs757440528 Chr.3: 49676115-49676115 on Build GRCh38
rs759250632 Chr.3: 49676251-49676251 on Build GRCh38
rs755673130 Chr.3: 49676195-49676197 on Build GRCh38
rs6778597 Chr.3: 49676320-49676320 on Build GRCh38
rs773068447 Chr.3: 49676163-49676163 on Build GRCh38
rs376075594 Chr.3: 49676225-49676225 on Build GRCh38
rs745989082 Chr.3: 49676029-49676029 on Build GRCh38
rs879725137 Chr.3: 49676447-49676447 on Build GRCh38
rs769246975 Chr.3: 49676317-49676317 on Build GRCh38
rs776839461 Chr.3: 49676217-49676217 on Build GRCh38
rs768340520 Chr.3: 49676057-49676057 on Build GRCh38
rs765493835 Chr.3: 49676121-49676121 on Build GRCh38
rs747993647 Chr.3: 49676161-49676161 on Build GRCh38
rs748743812 Chr.3: 49676167-49676167 on Build GRCh38
rs754749897 Chr.3: 49676140-49676140 on Build GRCh38
rs767226633 Chr.3: 49676122-49676123 on Build GRCh38
rs752059919 Chr.3: 49676010-49676010 on Build GRCh38
rs371914233 Chr.3: 49676247-49676247 on Build GRCh38
rs746814689 Chr.3: 49676047-49676047 on Build GRCh38
rs763116001 Chr.3: 49676359-49676359 on Build GRCh38
rs931042814 Chr.3: 49676354-49676354 on Build GRCh38
rs768176249 Chr.3: 49676015-49676015 on Build GRCh38
rs779974389 Chr.3: 49676129-49676129 on Build GRCh38
rs374439004 Chr.3: 49676238-49676238 on Build GRCh38
rs748974784 Chr.3: 49676263-49676263 on Build GRCh38
rs773373106 Chr.3: 49676356-49676356 on Build GRCh38
rs750647145 Chr.3: 49676223-49676223 on Build GRCh38
rs942606623 Chr.3: 49676310-49676310 on Build GRCh38
rs375865304 Chr.3: 49676352-49676352 on Build GRCh38
rs924752150 Chr.3: 49676244-49676244 on Build GRCh38
rs759716643 Chr.3: 49676395-49676395 on Build GRCh38
rs149887495 Chr.3: 49676127-49676127 on Build GRCh38
rs770364290 Chr.3: 49676174-49676174 on Build GRCh38
rs760148005 Chr.3: 49676014-49676014 on Build GRCh38
rs760438421 Chr.3: 49675996-49675996 on Build GRCh38
rs374073428 Chr.3: 49676268-49676268 on Build GRCh38
rs912468483 Chr.3: 49676302-49676302 on Build GRCh38
rs759198160 Chr.3: 49676033-49676033 on Build GRCh38
rs761116888 Chr.3: 49676178-49676178 on Build GRCh38
rs780876657 Chr.3: 49676158-49676158 on Build GRCh38
rs752175538 Chr.3: 49676138-49676140 on Build GRCh38
rs767293583 Chr.3: 49676404-49676404 on Build GRCh38
rs138376291 Chr.3: 49676446-49676446 on Build GRCh38
rs564910254 Chr.3: 49676306-49676306 on Build GRCh38
rs763159787 Chr.3: 49676132-49676132 on Build GRCh38
rs41290714 Chr.3: 49676433-49676433 on Build GRCh38
rs781237097 Chr.3: 49676239-49676239 on Build GRCh38
rs769546586 Chr.3: 49676162-49676162 on Build GRCh38
rs764849002 Chr.3: 49676009-49676009 on Build GRCh38
rs375854335 Chr.3: 49676361-49676361 on Build GRCh38
rs374901664 Chr.3: 49676175-49676175 on Build GRCh38
rs919595067 Chr.3: 49676339-49676339 on Build GRCh38
rs777745736 Chr.3: 49676435-49676435 on Build GRCh38
rs745433511 Chr.3: 49676332-49676332 on Build GRCh38
rs368656374 Chr.3: 49676246-49676246 on Build GRCh38
rs766681463 Chr.3: 49676025-49676026 on Build GRCh38
rs371544018 Chr.3: 49676341-49676341 on Build GRCh38
rs751153352 Chr.3: 49676032-49676033 on Build GRCh38
rs149050354 Chr.3: 49676218-49676218 on Build GRCh38
rs202163464 Chr.3: 49676180-49676180 on Build GRCh38
rs761046511 Chr.3: 49676102-49676102 on Build GRCh38
rs777720796 Chr.3: 49676261-49676261 on Build GRCh38
rs761320467 Chr.3: 49676005-49676005 on Build GRCh38
rs754051626 Chr.3: 49676113-49676113 on Build GRCh38
rs367976946 Chr.3: 49676336-49676336 on Build GRCh38
rs750490555 Chr.3: 49676122-49676122 on Build GRCh38
rs769385492 Chr.3: 49676084-49676084 on Build GRCh38
rs753094989 Chr.3: 49676020-49676020 on Build GRCh38
rs370423564 Chr.3: 49676032-49676032 on Build GRCh38
rs768336061 Chr.3: 49676212-49676212 on Build GRCh38
rs776211291 Chr.3: 49676004-49676004 on Build GRCh38
rs995682756 Chr.3: 49676062-49676062 on Build GRCh38
rs950643800 Chr.3: 49676377-49676377 on Build GRCh38
rs768339973 Chr.3: 49676000-49676000 on Build GRCh38
rs542596273 Chr.3: 49676027-49676027 on Build GRCh38
rs770438548 Chr.3: 49676271-49676271 on Build GRCh38
rs762598527 Chr.3: 49676086-49676086 on Build GRCh38
rs35610696 Chr.3: 49676344-49676345 on Build GRCh38
rs764392325 Chr.3: 49676105-49676105 on Build GRCh38
rs752697574 Chr.3: 49676430-49676430 on Build GRCh38
rs780720403 Chr.3: 49676213-49676213 on Build GRCh38
rs775869101 Chr.3: 49676095-49676095 on Build GRCh38
rs756505077 Chr.3: 49676025-49676025 on Build GRCh38
rs144465961 Chr.3: 49676087-49676087 on Build GRCh38
rs776289558 Chr.3: 49676080-49676080 on Build GRCh38
rs75779287 Chr.3: 49676355-49676355 on Build GRCh38
rs761390455 Chr.3: 49676081-49676081 on Build GRCh38
rs773703244 Chr.3: 49676329-49676329 on Build GRCh38

View Full Product Details