Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49860630-49861137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMKV
Entrez Gene ID
79012
Gene Name
CaM kinase like vesicle associated
Gene Aliases
1G5, VACAMKL
UniGene
Hs.145156
Gene Chromosome Location
Chr.3: 49857981-49870222 on Build GRCh38
Transcripts
NM_001320147.1 NM_024046.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3595431 c.636C>T p.I212I Chr.3: 49860945-49860945 on Build GRCh38
3595433 c.613G>A p.A205T Chr.3: 49860968-49860968 on Build GRCh38
3595429 c.706C>T p.L236F Chr.3: 49860785-49860785 on Build GRCh38
6165095 c.748C>G p.P250A Chr.3: 49860743-49860743 on Build GRCh38
6891108 c.755G>A p.W252* Chr.3: 49860736-49860736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367929526 Chr.3: 49861031-49861031 on Build GRCh38
rs1024562676 Chr.3: 49860726-49860726 on Build GRCh38
rs966029827 Chr.3: 49860965-49860965 on Build GRCh38
rs760783112 Chr.3: 49861034-49861034 on Build GRCh38
rs549814056 Chr.3: 49861114-49861114 on Build GRCh38
rs371001507 Chr.3: 49860702-49860702 on Build GRCh38
rs371721876 Chr.3: 49860922-49860922 on Build GRCh38
rs978016289 Chr.3: 49860894-49860894 on Build GRCh38
rs753996522 Chr.3: 49860891-49860891 on Build GRCh38
rs187588956 Chr.3: 49860858-49860858 on Build GRCh38
rs769240974 Chr.3: 49861104-49861104 on Build GRCh38
rs1010662195 Chr.3: 49860777-49860777 on Build GRCh38
rs201066068 Chr.3: 49860921-49860921 on Build GRCh38
rs754579583 Chr.3: 49861073-49861073 on Build GRCh38
rs966601822 Chr.3: 49860889-49860889 on Build GRCh38
rs200019805 Chr.3: 49860944-49860944 on Build GRCh38
rs779931517 Chr.3: 49860914-49860914 on Build GRCh38
rs201940404 Chr.3: 49860882-49860882 on Build GRCh38
rs756086165 Chr.3: 49860670-49860670 on Build GRCh38
rs754015057 Chr.3: 49861043-49861043 on Build GRCh38
rs772383085 Chr.3: 49861022-49861022 on Build GRCh38
rs746342901 Chr.3: 49861107-49861107 on Build GRCh38
rs772682236 Chr.3: 49861111-49861111 on Build GRCh38
rs757259460 Chr.3: 49860892-49860892 on Build GRCh38
rs764446739 Chr.3: 49861041-49861041 on Build GRCh38
rs760054653 Chr.3: 49860994-49860994 on Build GRCh38
rs544674672 Chr.3: 49860798-49860798 on Build GRCh38
rs745727823 Chr.3: 49860697-49860697 on Build GRCh38
rs374082013 Chr.3: 49861019-49861019 on Build GRCh38
rs763349594 Chr.3: 49860779-49860779 on Build GRCh38
rs147195969 Chr.3: 49860723-49860723 on Build GRCh38
rs368424095 Chr.3: 49860879-49860879 on Build GRCh38
rs779331047 Chr.3: 49860678-49860679 on Build GRCh38
rs750403491 Chr.3: 49860898-49860898 on Build GRCh38
rs779694976 Chr.3: 49861070-49861070 on Build GRCh38
rs758453823 Chr.3: 49861069-49861069 on Build GRCh38
rs139743520 Chr.3: 49860668-49860668 on Build GRCh38
rs988229656 Chr.3: 49860741-49860741 on Build GRCh38
rs373671685 Chr.3: 49860682-49860682 on Build GRCh38
rs190982523 Chr.3: 49861010-49861010 on Build GRCh38
rs774952824 Chr.3: 49860713-49860713 on Build GRCh38
rs900808066 Chr.3: 49860671-49860671 on Build GRCh38
rs777295134 Chr.3: 49861105-49861105 on Build GRCh38
rs149360477 Chr.3: 49860683-49860683 on Build GRCh38
rs780551684 Chr.3: 49860929-49860929 on Build GRCh38
rs1034873003 Chr.3: 49860861-49860861 on Build GRCh38
rs767428599 Chr.3: 49861065-49861065 on Build GRCh38
rs746611666 Chr.3: 49860919-49860919 on Build GRCh38
rs1042043836 Chr.3: 49860664-49860664 on Build GRCh38
rs761930579 Chr.3: 49861050-49861050 on Build GRCh38
rs201634061 Chr.3: 49860896-49860896 on Build GRCh38
rs945266334 Chr.3: 49860655-49860655 on Build GRCh38
rs780827857 Chr.3: 49861077-49861077 on Build GRCh38
rs767881361 Chr.3: 49860873-49860873 on Build GRCh38
rs144675947 Chr.3: 49860888-49860888 on Build GRCh38
rs62260755 Chr.3: 49860885-49860885 on Build GRCh38
rs764046352 Chr.3: 49860887-49860887 on Build GRCh38
rs768011837 Chr.3: 49860730-49860730 on Build GRCh38
rs200390586 Chr.3: 49861064-49861064 on Build GRCh38
rs897420950 Chr.3: 49860707-49860707 on Build GRCh38
rs777927846 Chr.3: 49860677-49860677 on Build GRCh38
rs776246810 Chr.3: 49860747-49860747 on Build GRCh38
rs774782174 Chr.3: 49860986-49860986 on Build GRCh38
rs747808274 Chr.3: 49861097-49861097 on Build GRCh38
rs751431269 Chr.3: 49861072-49861072 on Build GRCh38
rs746257567 Chr.3: 49860875-49860878 on Build GRCh38
rs747722227 Chr.3: 49860952-49860952 on Build GRCh38
rs758218386 Chr.3: 49860908-49860908 on Build GRCh38
rs2681781 Chr.3: 49860840-49860840 on Build GRCh38
rs769296484 Chr.3: 49860985-49860985 on Build GRCh38
rs933653627 Chr.3: 49860676-49860676 on Build GRCh38
rs772000541 Chr.3: 49860703-49860703 on Build GRCh38

View Full Product Details