Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51986513-51987017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD14A-ACY1
Entrez Gene ID
100526760
Gene Name
ABHD14A-ACY1 readthrough
Gene Aliases
-
UniGene
Hs.334707
Gene Chromosome Location
Chr.3: 51975026-51989202 on Build GRCh38
Transcripts
NM_001316331.1 NM_000666.2 NM_001198895.1 NM_001198896.1 NM_001198897.1 NM_001198898.1

Gene Symbol
ACY1
Entrez Gene ID
95
Gene Name
aminoacylase 1
Gene Aliases
ACY-1, ACY1D, HEL-S-5
UniGene
Hs.334707
Gene Chromosome Location
Chr.3: 51983284-51989202 on Build GRCh38
Transcripts
NM_001316331.1 NM_000666.2 NM_001198895.1 NM_001198896.1 NM_001198897.1 NM_001198898.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5922281 c.854-6C>T p.? Chr.3: 51986982-51986982 on Build GRCh38
3595865 c.797G>A p.G266D Chr.3: 51986605-51986605 on Build GRCh38
4659235 c.852G>T p.W284C Chr.3: 51986660-51986660 on Build GRCh38
6006951 c.544G>A p.D182N Chr.3: 51986622-51986622 on Build GRCh38
6361079 c.870C>A p.S290R Chr.3: 51987004-51987004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762010680 Chr.3: 51986577-51986577 on Build GRCh38
rs766679229 Chr.3: 51986604-51986604 on Build GRCh38
rs948630125 Chr.3: 51986858-51986858 on Build GRCh38
rs529152973 Chr.3: 51986804-51986804 on Build GRCh38
rs911448010 Chr.3: 51986792-51986792 on Build GRCh38
rs758418338 Chr.3: 51986988-51986988 on Build GRCh38
rs763442548 Chr.3: 51986806-51986806 on Build GRCh38
rs918750710 Chr.3: 51986700-51986700 on Build GRCh38
rs755887133 Chr.3: 51986631-51986631 on Build GRCh38
rs751168607 Chr.3: 51986616-51986616 on Build GRCh38
rs34300686 Chr.3: 51986775-51986775 on Build GRCh38
rs887540 Chr.3: 51986614-51986614 on Build GRCh38
rs960107711 Chr.3: 51986810-51986810 on Build GRCh38
rs776923612 Chr.3: 51986695-51986695 on Build GRCh38
rs750131593 Chr.3: 51986628-51986628 on Build GRCh38
rs777838025 Chr.3: 51986989-51986989 on Build GRCh38
rs761366650 Chr.3: 51986944-51986944 on Build GRCh38
rs773516710 Chr.3: 51986797-51986797 on Build GRCh38
rs903577840 Chr.3: 51986545-51986545 on Build GRCh38
rs771678688 Chr.3: 51986881-51986881 on Build GRCh38
rs374697368 Chr.3: 51986607-51986607 on Build GRCh38
rs753626169 Chr.3: 51986971-51986971 on Build GRCh38
rs778785581 Chr.3: 51986984-51986984 on Build GRCh38
rs532348676 Chr.3: 51986572-51986572 on Build GRCh38
rs569059581 Chr.3: 51986823-51986823 on Build GRCh38
rs557856270 Chr.3: 51986973-51986973 on Build GRCh38
rs143611624 Chr.3: 51986994-51986994 on Build GRCh38
rs772913923 Chr.3: 51986990-51986990 on Build GRCh38
rs772330147 Chr.3: 51986578-51986578 on Build GRCh38
rs767155768 Chr.3: 51986956-51986956 on Build GRCh38
rs934369244 Chr.3: 51986975-51986975 on Build GRCh38
rs779886954 Chr.3: 51986967-51986967 on Build GRCh38
rs760823696 Chr.3: 51986675-51986675 on Build GRCh38
rs748110984 Chr.3: 51986987-51986987 on Build GRCh38
rs121912700 Chr.3: 51986993-51986993 on Build GRCh38
rs528505269 Chr.3: 51986706-51986706 on Build GRCh38
rs771114222 Chr.3: 51986991-51986991 on Build GRCh38
rs769587878 Chr.3: 51986552-51986552 on Build GRCh38
rs767119395 Chr.3: 51986916-51986916 on Build GRCh38
rs887541 Chr.3: 51986588-51986588 on Build GRCh38
rs963077407 Chr.3: 51986653-51986653 on Build GRCh38
rs771263961 Chr.3: 51986688-51986688 on Build GRCh38
rs752853151 Chr.3: 51986970-51986970 on Build GRCh38
rs781117334 Chr.3: 51986540-51986540 on Build GRCh38
rs550839605 Chr.3: 51986818-51986818 on Build GRCh38
rs759943695 Chr.3: 51986861-51986861 on Build GRCh38
rs992725621 Chr.3: 51986822-51986822 on Build GRCh38
rs896360355 Chr.3: 51986610-51986610 on Build GRCh38
rs776561327 Chr.3: 51986723-51986723 on Build GRCh38
rs779725628 Chr.3: 51986632-51986632 on Build GRCh38
rs755790575 Chr.3: 51986961-51986961 on Build GRCh38
rs145367731 Chr.3: 51986859-51986859 on Build GRCh38
rs756943526 Chr.3: 51986619-51986619 on Build GRCh38
rs1023532959 Chr.3: 51986618-51986618 on Build GRCh38
rs781031234 Chr.3: 51986622-51986622 on Build GRCh38
rs778941108 Chr.3: 51986644-51986644 on Build GRCh38
rs772174586 Chr.3: 51986650-51986650 on Build GRCh38
rs751082521 Chr.3: 51986941-51986941 on Build GRCh38
rs749245599 Chr.3: 51986636-51986636 on Build GRCh38
rs915858311 Chr.3: 51986853-51986853 on Build GRCh38
rs762194323 Chr.3: 51986724-51986724 on Build GRCh38
rs760992766 Chr.3: 51986597-51986597 on Build GRCh38
rs745705587 Chr.3: 51986546-51986546 on Build GRCh38
rs546642804 Chr.3: 51986717-51986717 on Build GRCh38
rs770313896 Chr.3: 51986996-51986996 on Build GRCh38
rs925809647 Chr.3: 51986876-51986876 on Build GRCh38
rs935069968 Chr.3: 51986841-51986841 on Build GRCh38
rs750037617 Chr.3: 51986957-51986957 on Build GRCh38
rs185086428 Chr.3: 51986615-51986615 on Build GRCh38
rs375378800 Chr.3: 51986576-51986576 on Build GRCh38
rs748268468 Chr.3: 51986649-51986649 on Build GRCh38
rs760071840 Chr.3: 51986699-51986699 on Build GRCh38
rs768535020 Chr.3: 51986639-51986639 on Build GRCh38
rs768830579 Chr.3: 51986689-51986689 on Build GRCh38
rs762709516 Chr.3: 51986637-51986638 on Build GRCh38
rs770702363 Chr.3: 51986652-51986653 on Build GRCh38
rs749204999 Chr.3: 51986567-51986567 on Build GRCh38
rs779999350 Chr.3: 51986562-51986562 on Build GRCh38
rs564349352 Chr.3: 51986666-51986666 on Build GRCh38

View Full Product Details