Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52198000-52198496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALAS1
Entrez Gene ID
211
Gene Name
5'-aminolevulinate synthase 1
Gene Aliases
ALAS, ALAS-H, ALAS3, ALASH, MIG4
UniGene
Hs.476308
Gene Chromosome Location
Chr.3: 52198083-52214327 on Build GRCh38
Transcripts
NM_000688.5 NM_001304443.1 NM_001304444.1 NM_199166.2 XM_011533477.1 XM_011533478.2 XM_017005873.1

SNP Detail

SNP ID Chromosome Location
rs530727954 Chr.3: 52198114-52198114 on Build GRCh38
rs377535243 Chr.3: 52198029-52198029 on Build GRCh38
rs969303026 Chr.3: 52198163-52198163 on Build GRCh38
rs953566621 Chr.3: 52198182-52198182 on Build GRCh38
rs896278018 Chr.3: 52198350-52198350 on Build GRCh38
rs937833158 Chr.3: 52198394-52198394 on Build GRCh38
rs976315951 Chr.3: 52198090-52198090 on Build GRCh38
rs709315 Chr.3: 52198169-52198169 on Build GRCh38
rs1005986684 Chr.3: 52198395-52198395 on Build GRCh38
rs561824801 Chr.3: 52198103-52198103 on Build GRCh38
rs756800144 Chr.3: 52198310-52198310 on Build GRCh38
rs549071088 Chr.3: 52198176-52198176 on Build GRCh38
rs537913781 Chr.3: 52198051-52198052 on Build GRCh38
rs369797960 Chr.3: 52198067-52198067 on Build GRCh38
rs72955286 Chr.3: 52198382-52198382 on Build GRCh38
rs905035347 Chr.3: 52198141-52198141 on Build GRCh38
rs962760891 Chr.3: 52198173-52198173 on Build GRCh38
rs904983313 Chr.3: 52198051-52198051 on Build GRCh38
rs755659893 Chr.3: 52198102-52198102 on Build GRCh38
rs564394089 Chr.3: 52198126-52198126 on Build GRCh38
rs1001939014 Chr.3: 52198057-52198057 on Build GRCh38
rs1031137214 Chr.3: 52198033-52198034 on Build GRCh38
rs1002013211 Chr.3: 52198145-52198145 on Build GRCh38
rs72955285 Chr.3: 52198046-52198046 on Build GRCh38
rs1009301578 Chr.3: 52198432-52198432 on Build GRCh38
rs919235444 Chr.3: 52198345-52198345 on Build GRCh38
rs529309778 Chr.3: 52198175-52198175 on Build GRCh38
rs1026046493 Chr.3: 52198448-52198448 on Build GRCh38
rs1020541533 Chr.3: 52198079-52198079 on Build GRCh38
rs867844115 Chr.3: 52198028-52198028 on Build GRCh38
rs905533791 Chr.3: 52198465-52198465 on Build GRCh38
rs777466947 Chr.3: 52198147-52198147 on Build GRCh38
rs1053609665 Chr.3: 52198420-52198420 on Build GRCh38
rs544123932 Chr.3: 52198118-52198118 on Build GRCh38
rs373873328 Chr.3: 52198164-52198164 on Build GRCh38
rs1044017324 Chr.3: 52198302-52198302 on Build GRCh38
rs1039315258 Chr.3: 52198250-52198276 on Build GRCh38
rs1030554522 Chr.3: 52198178-52198178 on Build GRCh38
rs541962284 Chr.3: 52198096-52198096 on Build GRCh38
rs756435244 Chr.3: 52198122-52198122 on Build GRCh38
rs146474312 Chr.3: 52198150-52198150 on Build GRCh38
rs999571270 Chr.3: 52198104-52198104 on Build GRCh38
rs1046760952 Chr.3: 52198368-52198368 on Build GRCh38
rs143479698 Chr.3: 52198422-52198422 on Build GRCh38
rs191728871 Chr.3: 52198049-52198049 on Build GRCh38
rs569059062 Chr.3: 52198305-52198305 on Build GRCh38
rs923414986 Chr.3: 52198093-52198093 on Build GRCh38
rs887765816 Chr.3: 52198027-52198027 on Build GRCh38
rs542308398 Chr.3: 52198153-52198153 on Build GRCh38
rs753988342 Chr.3: 52198404-52198405 on Build GRCh38
rs967679038 Chr.3: 52198174-52198174 on Build GRCh38
rs949383328 Chr.3: 52198356-52198356 on Build GRCh38
rs957708430 Chr.3: 52198068-52198068 on Build GRCh38
rs934179311 Chr.3: 52198071-52198071 on Build GRCh38
rs986223966 Chr.3: 52198183-52198183 on Build GRCh38
rs112304502 Chr.3: 52198342-52198342 on Build GRCh38
rs1025222924 Chr.3: 52198050-52198050 on Build GRCh38
rs1009270453 Chr.3: 52198167-52198167 on Build GRCh38
rs796949462 Chr.3: 52198095-52198095 on Build GRCh38
rs909310151 Chr.3: 52198297-52198297 on Build GRCh38
rs1036287840 Chr.3: 52198121-52198121 on Build GRCh38
rs942814593 Chr.3: 52198284-52198284 on Build GRCh38
rs1036256752 Chr.3: 52198031-52198031 on Build GRCh38
rs987764641 Chr.3: 52198075-52198075 on Build GRCh38
rs1051294866 Chr.3: 52198218-52198218 on Build GRCh38
rs1007129388 Chr.3: 52198091-52198091 on Build GRCh38
rs918644063 Chr.3: 52198069-52198069 on Build GRCh38
rs745756384 Chr.3: 52198438-52198438 on Build GRCh38
rs1046254329 Chr.3: 52198130-52198130 on Build GRCh38
rs968025680 Chr.3: 52198089-52198089 on Build GRCh38
rs969206962 Chr.3: 52198387-52198387 on Build GRCh38
rs1042084018 Chr.3: 52198444-52198444 on Build GRCh38

View Full Product Details