Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52292942-52293449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYCTK
Entrez Gene ID
132158
Gene Name
glycerate kinase
Gene Aliases
HBEBP2, HBEBP4, HBeAgBP4A
UniGene
Hs.415312
Gene Chromosome Location
Chr.3: 52287558-52295256 on Build GRCh38
Transcripts
NM_001144951.1 NM_145262.3 NR_026699.1 NR_026700.1 NR_026701.1 NR_026702.1 XM_017005730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6570106 c.1466C>G p.S489* Chr.3: 52293020-52293020 on Build GRCh38
1726984 c.1547A>G p.H516R Chr.3: 52293101-52293101 on Build GRCh38
3595914 c.1536C>T p.V512V Chr.3: 52293090-52293090 on Build GRCh38
4118983 c.1569G>A p.R523R Chr.3: 52293123-52293123 on Build GRCh38
1208369 c.1416G>T p.Q472H Chr.3: 52292970-52292970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043876799 Chr.3: 52293354-52293354 on Build GRCh38
rs775764441 Chr.3: 52293423-52293424 on Build GRCh38
rs61740226 Chr.3: 52293120-52293120 on Build GRCh38
rs561918673 Chr.3: 52293374-52293374 on Build GRCh38
rs765754153 Chr.3: 52293018-52293018 on Build GRCh38
rs1030916593 Chr.3: 52293211-52293211 on Build GRCh38
rs763434728 Chr.3: 52293102-52293102 on Build GRCh38
rs749901380 Chr.3: 52293030-52293030 on Build GRCh38
rs1051244917 Chr.3: 52293333-52293333 on Build GRCh38
rs758280911 Chr.3: 52293069-52293069 on Build GRCh38
rs747076949 Chr.3: 52293081-52293081 on Build GRCh38
rs934151331 Chr.3: 52293114-52293114 on Build GRCh38
rs770144281 Chr.3: 52293099-52293099 on Build GRCh38
rs1007065246 Chr.3: 52293140-52293140 on Build GRCh38
rs946526496 Chr.3: 52293421-52293421 on Build GRCh38
rs753363748 Chr.3: 52293122-52293122 on Build GRCh38
rs942428095 Chr.3: 52293353-52293353 on Build GRCh38
rs934273983 Chr.3: 52293330-52293330 on Build GRCh38
rs759124933 Chr.3: 52293130-52293130 on Build GRCh38
rs77578880 Chr.3: 52293290-52293290 on Build GRCh38
rs911399155 Chr.3: 52293350-52293350 on Build GRCh38
rs1039651830 Chr.3: 52293203-52293203 on Build GRCh38
rs749641123 Chr.3: 52293231-52293231 on Build GRCh38
rs117180862 Chr.3: 52293278-52293278 on Build GRCh38
rs773278880 Chr.3: 52292992-52292992 on Build GRCh38
rs747943703 Chr.3: 52293067-52293067 on Build GRCh38
rs774760990 Chr.3: 52293116-52293116 on Build GRCh38
rs369555553 Chr.3: 52292988-52292988 on Build GRCh38
rs933978304 Chr.3: 52293226-52293226 on Build GRCh38
rs749904278 Chr.3: 52292962-52292962 on Build GRCh38
rs121909447 Chr.3: 52293002-52293002 on Build GRCh38
rs764903675 Chr.3: 52293131-52293131 on Build GRCh38
rs80017945 Chr.3: 52292968-52292968 on Build GRCh38
rs749088667 Chr.3: 52292979-52292979 on Build GRCh38
rs777617813 Chr.3: 52293070-52293070 on Build GRCh38
rs780955581 Chr.3: 52293423-52293423 on Build GRCh38
rs981782327 Chr.3: 52293338-52293338 on Build GRCh38
rs2293159 Chr.3: 52293292-52293292 on Build GRCh38
rs541005983 Chr.3: 52293362-52293362 on Build GRCh38
rs749082610 Chr.3: 52293027-52293029 on Build GRCh38
rs1023916136 Chr.3: 52293240-52293240 on Build GRCh38
rs755753426 Chr.3: 52292970-52292970 on Build GRCh38
rs781066450 Chr.3: 52293200-52293200 on Build GRCh38
rs121909448 Chr.3: 52293032-52293032 on Build GRCh38
rs757231683 Chr.3: 52293199-52293199 on Build GRCh38
rs144669248 Chr.3: 52293055-52293055 on Build GRCh38
rs770979346 Chr.3: 52293088-52293088 on Build GRCh38
rs751505344 Chr.3: 52293194-52293194 on Build GRCh38
rs970569662 Chr.3: 52293020-52293020 on Build GRCh38
rs764539112 Chr.3: 52293025-52293025 on Build GRCh38
rs751373971 Chr.3: 52293379-52293379 on Build GRCh38
rs999190657 Chr.3: 52293396-52293396 on Build GRCh38
rs747079334 Chr.3: 52293010-52293010 on Build GRCh38
rs777759189 Chr.3: 52293154-52293154 on Build GRCh38
rs752410530 Chr.3: 52293300-52293300 on Build GRCh38
rs896388654 Chr.3: 52293411-52293411 on Build GRCh38
rs1052122072 Chr.3: 52293400-52293401 on Build GRCh38
rs78388149 Chr.3: 52293022-52293022 on Build GRCh38
rs766025190 Chr.3: 52293034-52293034 on Build GRCh38
rs775955431 Chr.3: 52293100-52293100 on Build GRCh38
rs1051559131 Chr.3: 52293254-52293254 on Build GRCh38
rs752464066 Chr.3: 52293147-52293147 on Build GRCh38
rs371018021 Chr.3: 52292978-52292978 on Build GRCh38
rs771159734 Chr.3: 52293011-52293011 on Build GRCh38
rs186354416 Chr.3: 52293152-52293152 on Build GRCh38
rs745975674 Chr.3: 52293206-52293206 on Build GRCh38
rs770091220 Chr.3: 52293214-52293214 on Build GRCh38
rs147924619 Chr.3: 52292980-52292980 on Build GRCh38
rs778624214 Chr.3: 52293063-52293063 on Build GRCh38
rs121909449 Chr.3: 52293112-52293112 on Build GRCh38
rs560961842 Chr.3: 52293373-52293373 on Build GRCh38
rs774909431 Chr.3: 52293241-52293241 on Build GRCh38
rs146250137 Chr.3: 52293121-52293121 on Build GRCh38
rs774599407 Chr.3: 52293361-52293361 on Build GRCh38
rs971660120 Chr.3: 52293316-52293316 on Build GRCh38
rs991718173 Chr.3: 52293013-52293013 on Build GRCh38
rs372098626 Chr.3: 52293040-52293040 on Build GRCh38
rs763485712 Chr.3: 52293023-52293023 on Build GRCh38
rs772013256 Chr.3: 52292987-52292987 on Build GRCh38
rs903640972 Chr.3: 52293375-52293375 on Build GRCh38
rs746028398 Chr.3: 52293093-52293093 on Build GRCh38
rs918720114 Chr.3: 52293054-52293054 on Build GRCh38
rs1013086924 Chr.3: 52293299-52293299 on Build GRCh38
rs988300145 Chr.3: 52293380-52293380 on Build GRCh38
rs574808932 Chr.3: 52293409-52293409 on Build GRCh38
rs751918729 Chr.3: 52293224-52293224 on Build GRCh38
rs778822139 Chr.3: 52292984-52292984 on Build GRCh38
rs146102973 Chr.3: 52293091-52293091 on Build GRCh38
rs759970139 Chr.3: 52293016-52293016 on Build GRCh38
rs547441620 Chr.3: 52293027-52293027 on Build GRCh38
rs767047161 Chr.3: 52293014-52293014 on Build GRCh38
rs941592166 Chr.3: 52293037-52293037 on Build GRCh38
rs748144159 Chr.3: 52292986-52292986 on Build GRCh38
rs776183482 Chr.3: 52293203-52293204 on Build GRCh38
rs768970076 Chr.3: 52293235-52293235 on Build GRCh38
rs781463016 Chr.3: 52293092-52293092 on Build GRCh38
rs763738933 Chr.3: 52293375-52293375 on Build GRCh38
rs865778756 Chr.3: 52293227-52293227 on Build GRCh38
rs572056020 Chr.3: 52293319-52293319 on Build GRCh38
rs370143058 Chr.3: 52293115-52293115 on Build GRCh38
rs762173198 Chr.3: 52293264-52293264 on Build GRCh38
rs757101092 Chr.3: 52293400-52293400 on Build GRCh38
rs779596586 Chr.3: 52292977-52292977 on Build GRCh38

View Full Product Details