Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109740816-109741224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTM3
Entrez Gene ID
2947
Gene Name
glutathione S-transferase mu 3
Gene Aliases
GST5, GSTB, GSTM3-3, GTM3
UniGene
Hs.2006
Gene Chromosome Location
Chr.1: 109733932-109741038 on Build GRCh38
Transcripts
NM_000849.4 NR_024537.1

SNP Detail

SNP ID Chromosome Location
rs954054829 Chr.1: 109741051-109741051 on Build GRCh38
rs988553881 Chr.1: 109741166-109741166 on Build GRCh38
rs942576402 Chr.1: 109740930-109740930 on Build GRCh38
rs984135294 Chr.1: 109741068-109741068 on Build GRCh38
rs896524314 Chr.1: 109741036-109741036 on Build GRCh38
rs190592833 Chr.1: 109741196-109741196 on Build GRCh38
rs1051059271 Chr.1: 109740913-109740913 on Build GRCh38
rs894855280 Chr.1: 109740897-109740897 on Build GRCh38
rs973362025 Chr.1: 109741154-109741154 on Build GRCh38
rs1028393228 Chr.1: 109741042-109741042 on Build GRCh38
rs1001095643 Chr.1: 109741117-109741117 on Build GRCh38
rs552574041 Chr.1: 109740859-109740859 on Build GRCh38
rs961652450 Chr.1: 109741104-109741104 on Build GRCh38
rs754898472 Chr.1: 109740847-109740847 on Build GRCh38
rs546557108 Chr.1: 109741200-109741200 on Build GRCh38
rs879508838 Chr.1: 109741179-109741179 on Build GRCh38
rs994259210 Chr.1: 109740979-109740979 on Build GRCh38
rs1023914745 Chr.1: 109741066-109741066 on Build GRCh38
rs560861074 Chr.1: 109740840-109740840 on Build GRCh38
rs1043656596 Chr.1: 109740961-109740961 on Build GRCh38
rs752552504 Chr.1: 109741058-109741058 on Build GRCh38
rs933649005 Chr.1: 109740891-109740891 on Build GRCh38
rs548285750 Chr.1: 109740976-109740976 on Build GRCh38
rs901664565 Chr.1: 109741017-109741017 on Build GRCh38
rs767513346 Chr.1: 109741008-109741008 on Build GRCh38
rs1054795626 Chr.1: 109740881-109740881 on Build GRCh38
rs1038621765 Chr.1: 109740935-109740935 on Build GRCh38
rs141867647 Chr.1: 109740896-109740896 on Build GRCh38
rs4970777 Chr.1: 109741163-109741163 on Build GRCh38
rs55706939 Chr.1: 109741198-109741198 on Build GRCh38
rs1032942616 Chr.1: 109741160-109741160 on Build GRCh38
rs912906497 Chr.1: 109740887-109740887 on Build GRCh38
rs911110082 Chr.1: 109740923-109740923 on Build GRCh38
rs905430860 Chr.1: 109741096-109741096 on Build GRCh38
rs531738640 Chr.1: 109740905-109740905 on Build GRCh38
rs1047146611 Chr.1: 109740997-109740997 on Build GRCh38
rs1014003412 Chr.1: 109741047-109741047 on Build GRCh38
rs532379951 Chr.1: 109740858-109740858 on Build GRCh38
rs1018784891 Chr.1: 109741016-109741016 on Build GRCh38

View Full Product Details