Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 56623508-56624020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM208A
Entrez Gene ID
23272
Gene Name
family with sequence similarity 208 member A
Gene Aliases
C3orf63, RAP140, se89-1
UniGene
Hs.116877
Gene Chromosome Location
Chr.3: 56620132-56683242 on Build GRCh38
Transcripts
NM_001112736.1 NM_015224.3 XM_005264999.1 XM_006713077.2 XM_006713078.2 XM_011533552.2 XM_011533553.2 XM_017006030.1 XM_017006031.1 XM_017006032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1047505 c.4303C>A p.L1435I Chr.3: 56623564-56623564 on Build GRCh38
3824422 c.4342C>G p.L1448V Chr.3: 56623525-56623525 on Build GRCh38
3824421 c.3214C>G p.L1072V Chr.3: 56623525-56623525 on Build GRCh38
5865105 c.3217G>A p.D1073N Chr.3: 56623522-56623522 on Build GRCh38
1424763 c.3186C>T p.N1062N Chr.3: 56623553-56623553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998252721 Chr.3: 56623755-56623755 on Build GRCh38
rs374479800 Chr.3: 56623610-56623613 on Build GRCh38
rs976938290 Chr.3: 56623889-56623889 on Build GRCh38
rs150226106 Chr.3: 56623948-56623948 on Build GRCh38
rs965032658 Chr.3: 56623884-56623884 on Build GRCh38
rs375725979 Chr.3: 56623917-56623917 on Build GRCh38
rs767856680 Chr.3: 56623551-56623551 on Build GRCh38
rs561775495 Chr.3: 56623856-56623856 on Build GRCh38
rs759157601 Chr.3: 56623603-56623603 on Build GRCh38
rs752687213 Chr.3: 56623656-56623656 on Build GRCh38
rs563758660 Chr.3: 56623923-56623923 on Build GRCh38
rs902465963 Chr.3: 56623990-56623990 on Build GRCh38
rs918849219 Chr.3: 56623907-56623907 on Build GRCh38
rs1014270463 Chr.3: 56623984-56623984 on Build GRCh38
rs895668853 Chr.3: 56623658-56623658 on Build GRCh38
rs917066022 Chr.3: 56623971-56623971 on Build GRCh38
rs957844280 Chr.3: 56623730-56623730 on Build GRCh38
rs879433730 Chr.3: 56623907-56623908 on Build GRCh38
rs937111681 Chr.3: 56623649-56623649 on Build GRCh38
rs769588554 Chr.3: 56623805-56623806 on Build GRCh38
rs559971311 Chr.3: 56623653-56623653 on Build GRCh38
rs183432229 Chr.3: 56623987-56623987 on Build GRCh38
rs758466141 Chr.3: 56623701-56623701 on Build GRCh38
rs937062233 Chr.3: 56623962-56623964 on Build GRCh38
rs994070168 Chr.3: 56623662-56623662 on Build GRCh38
rs141000613 Chr.3: 56623871-56623871 on Build GRCh38
rs777503289 Chr.3: 56623772-56623772 on Build GRCh38
rs895788886 Chr.3: 56623981-56623981 on Build GRCh38
rs565787244 Chr.3: 56623883-56623887 on Build GRCh38
rs190447121 Chr.3: 56623700-56623700 on Build GRCh38
rs910910316 Chr.3: 56623863-56623863 on Build GRCh38
rs1013488388 Chr.3: 56623655-56623655 on Build GRCh38
rs200344458 Chr.3: 56623584-56623584 on Build GRCh38
rs762236481 Chr.3: 56623908-56623908 on Build GRCh38
rs917884597 Chr.3: 56623609-56623618 on Build GRCh38
rs768896673 Chr.3: 56623582-56623582 on Build GRCh38
rs780396137 Chr.3: 56623545-56623553 on Build GRCh38
rs1055909306 Chr.3: 56623969-56623969 on Build GRCh38
rs1055835061 Chr.3: 56623632-56623632 on Build GRCh38
rs753555079 Chr.3: 56623553-56623553 on Build GRCh38
rs546522995 Chr.3: 56623951-56623951 on Build GRCh38
rs372449943 Chr.3: 56623609-56623609 on Build GRCh38
rs751380219 Chr.3: 56623853-56623853 on Build GRCh38
rs1022977014 Chr.3: 56623888-56623889 on Build GRCh38
rs957108678 Chr.3: 56623855-56623855 on Build GRCh38
rs573519750 Chr.3: 56623654-56623654 on Build GRCh38
rs992591945 Chr.3: 56623967-56623967 on Build GRCh38
rs1018265667 Chr.3: 56623719-56623719 on Build GRCh38
rs764776253 Chr.3: 56623608-56623608 on Build GRCh38
rs1029755536 Chr.3: 56623796-56623796 on Build GRCh38
rs546548226 Chr.3: 56623612-56623612 on Build GRCh38
rs1047440624 Chr.3: 56623689-56623689 on Build GRCh38
rs966910617 Chr.3: 56623691-56623691 on Build GRCh38
rs777057409 Chr.3: 56623605-56623606 on Build GRCh38
rs747607787 Chr.3: 56623548-56623550 on Build GRCh38

View Full Product Details