Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57227431-57227928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APPL1
Entrez Gene ID
26060
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1
Gene Aliases
APPL, DIP13alpha, MODY14
UniGene
Hs.476415
Gene Chromosome Location
Chr.3: 57227737-57273471 on Build GRCh38
Transcripts
NM_012096.2 XM_011533583.2 XM_005265526.4 XM_011534204.2 XM_011534205.2 XM_017007421.1

Gene Symbol
HESX1
Entrez Gene ID
8820
Gene Name
HESX homeobox 1
Gene Aliases
ANF, CPHD5, RPX
UniGene
Hs.171980
Gene Chromosome Location
Chr.3: 57197838-57227643 on Build GRCh38
Transcripts
NM_012096.2 XM_011533583.2 XM_005265526.4 XM_011534204.2 XM_011534205.2 XM_017007421.1

SNP Detail

SNP ID Chromosome Location
rs866978701 Chr.3: 57227772-57227772 on Build GRCh38
rs981859656 Chr.3: 57227719-57227719 on Build GRCh38
rs976475926 Chr.3: 57227610-57227610 on Build GRCh38
rs770562918 Chr.3: 57227849-57227849 on Build GRCh38
rs893890640 Chr.3: 57227713-57227713 on Build GRCh38
rs974041000 Chr.3: 57227794-57227794 on Build GRCh38
rs998814599 Chr.3: 57227635-57227635 on Build GRCh38
rs554193687 Chr.3: 57227740-57227740 on Build GRCh38
rs765290388 Chr.3: 57227867-57227867 on Build GRCh38
rs578118925 Chr.3: 57227731-57227731 on Build GRCh38
rs1007369551 Chr.3: 57227790-57227790 on Build GRCh38
rs912768161 Chr.3: 57227825-57227825 on Build GRCh38
rs747387990 Chr.3: 57227581-57227581 on Build GRCh38
rs1054013712 Chr.3: 57227851-57227851 on Build GRCh38
rs935826598 Chr.3: 57227544-57227544 on Build GRCh38
rs1055721764 Chr.3: 57227552-57227552 on Build GRCh38
rs902537018 Chr.3: 57227671-57227671 on Build GRCh38
rs957222430 Chr.3: 57227638-57227638 on Build GRCh38
rs375189154 Chr.3: 57227870-57227870 on Build GRCh38
rs7643644 Chr.3: 57227866-57227866 on Build GRCh38
rs940254004 Chr.3: 57227574-57227574 on Build GRCh38
rs958505254 Chr.3: 57227845-57227845 on Build GRCh38
rs971416486 Chr.3: 57227718-57227718 on Build GRCh38
rs917050720 Chr.3: 57227761-57227761 on Build GRCh38
rs996999793 Chr.3: 57227604-57227604 on Build GRCh38
rs113307246 Chr.3: 57227733-57227733 on Build GRCh38
rs950070372 Chr.3: 57227563-57227563 on Build GRCh38
rs757579727 Chr.3: 57227588-57227588 on Build GRCh38
rs944199832 Chr.3: 57227828-57227828 on Build GRCh38
rs747037943 Chr.3: 57227545-57227545 on Build GRCh38
rs934363902 Chr.3: 57227847-57227847 on Build GRCh38
rs958177744 Chr.3: 57227470-57227470 on Build GRCh38
rs1013626534 Chr.3: 57227558-57227558 on Build GRCh38
rs918378587 Chr.3: 57227523-57227523 on Build GRCh38
rs943723030 Chr.3: 57227482-57227482 on Build GRCh38
rs1025864607 Chr.3: 57227738-57227738 on Build GRCh38
rs935374333 Chr.3: 57227781-57227781 on Build GRCh38
rs769044973 Chr.3: 57227835-57227835 on Build GRCh38
rs576610110 Chr.3: 57227856-57227856 on Build GRCh38
rs991365436 Chr.3: 57227759-57227759 on Build GRCh38
rs1022835366 Chr.3: 57227890-57227890 on Build GRCh38
rs879640477 Chr.3: 57227762-57227762 on Build GRCh38
rs1012709200 Chr.3: 57227771-57227771 on Build GRCh38
rs888297085 Chr.3: 57227538-57227538 on Build GRCh38
rs903069928 Chr.3: 57227779-57227779 on Build GRCh38
rs557919192 Chr.3: 57227637-57227637 on Build GRCh38
rs547377828 Chr.3: 57227506-57227506 on Build GRCh38
rs186782833 Chr.3: 57227627-57227627 on Build GRCh38
rs372788109 Chr.3: 57227630-57227630 on Build GRCh38
rs571175742 Chr.3: 57227892-57227892 on Build GRCh38
rs932818624 Chr.3: 57227737-57227737 on Build GRCh38
rs774690109 Chr.3: 57227844-57227844 on Build GRCh38
rs1040146567 Chr.3: 57227830-57227830 on Build GRCh38
rs999585311 Chr.3: 57227569-57227569 on Build GRCh38
rs991664989 Chr.3: 57227854-57227854 on Build GRCh38
rs538392029 Chr.3: 57227633-57227633 on Build GRCh38
rs13066738 Chr.3: 57227589-57227589 on Build GRCh38
rs562898267 Chr.3: 57227773-57227773 on Build GRCh38
rs949994262 Chr.3: 57227654-57227654 on Build GRCh38
rs991256753 Chr.3: 57227640-57227640 on Build GRCh38
rs113472303 Chr.3: 57227564-57227564 on Build GRCh38
rs868169706 Chr.3: 57227453-57227453 on Build GRCh38
rs573947766 Chr.3: 57227744-57227744 on Build GRCh38
rs749769502 Chr.3: 57227834-57227834 on Build GRCh38
rs569386895 Chr.3: 57227629-57227629 on Build GRCh38
rs918230450 Chr.3: 57227626-57227626 on Build GRCh38
rs971631570 Chr.3: 57227777-57227777 on Build GRCh38
rs987121540 Chr.3: 57227473-57227473 on Build GRCh38
rs999449622 Chr.3: 57227684-57227684 on Build GRCh38
rs1032360705 Chr.3: 57227573-57227573 on Build GRCh38
rs951603726 Chr.3: 57227827-57227827 on Build GRCh38
rs1007095765 Chr.3: 57227726-57227726 on Build GRCh38
rs908603389 Chr.3: 57227782-57227782 on Build GRCh38
rs565124633 Chr.3: 57227465-57227465 on Build GRCh38
rs1053709253 Chr.3: 57227783-57227783 on Build GRCh38
rs994624455 Chr.3: 57227461-57227461 on Build GRCh38
rs983821830 Chr.3: 57227831-57227831 on Build GRCh38
rs966008173 Chr.3: 57227606-57227606 on Build GRCh38
rs1032391888 Chr.3: 57227700-57227700 on Build GRCh38
rs1025901871 Chr.3: 57227813-57227813 on Build GRCh38
rs958208801 Chr.3: 57227575-57227575 on Build GRCh38
rs901349054 Chr.3: 57227798-57227798 on Build GRCh38
rs542640717 Chr.3: 57227756-57227756 on Build GRCh38
rs908576182 Chr.3: 57227729-57227729 on Build GRCh38
rs1018571726 Chr.3: 57227631-57227631 on Build GRCh38
rs920192537 Chr.3: 57227796-57227796 on Build GRCh38
rs998328674 Chr.3: 57227803-57227803 on Build GRCh38
rs1046416230 Chr.3: 57227656-57227656 on Build GRCh38
rs746341864 Chr.3: 57227787-57227787 on Build GRCh38
rs977279844 Chr.3: 57227878-57227878 on Build GRCh38
rs752732231 Chr.3: 57227888-57227888 on Build GRCh38
rs13066730 Chr.3: 57227583-57227583 on Build GRCh38
rs1018337446 Chr.3: 57227797-57227797 on Build GRCh38
rs371205045 Chr.3: 57227596-57227596 on Build GRCh38
rs760307551 Chr.3: 57227848-57227848 on Build GRCh38
rs965683268 Chr.3: 57227732-57227732 on Build GRCh38
rs1025387332 Chr.3: 57227560-57227560 on Build GRCh38
rs1048280004 Chr.3: 57227532-57227532 on Build GRCh38
rs777556419 Chr.3: 57227529-57227529 on Build GRCh38
rs909672315 Chr.3: 57227748-57227748 on Build GRCh38
rs776638200 Chr.3: 57227597-57227597 on Build GRCh38
rs887361681 Chr.3: 57227571-57227572 on Build GRCh38
rs527539950 Chr.3: 57227472-57227472 on Build GRCh38
rs916017193 Chr.3: 57227710-57227710 on Build GRCh38
rs1042034695 Chr.3: 57227765-57227765 on Build GRCh38
rs957263732 Chr.3: 57227766-57227766 on Build GRCh38
rs895767157 Chr.3: 57227639-57227639 on Build GRCh38
rs895683570 Chr.3: 57227556-57227556 on Build GRCh38
rs911430328 Chr.3: 57227474-57227474 on Build GRCh38

View Full Product Details