Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57626405-57626953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND6A
Entrez Gene ID
201627
Gene Name
DENN domain containing 6A
Gene Aliases
AFI1A, FAM116A
UniGene
Hs.91085
Gene Chromosome Location
Chr.3: 57625454-57693118 on Build GRCh38
Transcripts
NM_152678.2 XM_006713019.3 XM_006713020.2 XM_017005864.1 NM_001322176.1

Gene Symbol
PDE12
Entrez Gene ID
201626
Gene Name
phosphodiesterase 12
Gene Aliases
2'-PDE, 2-PDE
UniGene
Hs.572993
Gene Chromosome Location
Chr.3: 57556247-57656480 on Build GRCh38
Transcripts
NM_152678.2 XM_006713019.3 XM_006713020.2 XM_017005864.1 NM_001322176.1

SNP Detail

SNP ID Chromosome Location
rs943465557 Chr.3: 57626744-57626744 on Build GRCh38
rs929320761 Chr.3: 57626710-57626710 on Build GRCh38
rs202120160 Chr.3: 57626820-57626820 on Build GRCh38
rs1009680208 Chr.3: 57626827-57626827 on Build GRCh38
rs1049143318 Chr.3: 57626731-57626731 on Build GRCh38
rs896265887 Chr.3: 57626771-57626771 on Build GRCh38
rs200144269 Chr.3: 57626831-57626831 on Build GRCh38
rs1143354 Chr.3: 57626485-57626485 on Build GRCh38
rs939560464 Chr.3: 57626657-57626657 on Build GRCh38
rs972502585 Chr.3: 57626592-57626592 on Build GRCh38
rs1020601993 Chr.3: 57626488-57626488 on Build GRCh38
rs532287574 Chr.3: 57626570-57626570 on Build GRCh38
rs968401146 Chr.3: 57626490-57626490 on Build GRCh38
rs567921070 Chr.3: 57626499-57626499 on Build GRCh38
rs903516965 Chr.3: 57626823-57626823 on Build GRCh38
rs980899593 Chr.3: 57626628-57626628 on Build GRCh38
rs193003650 Chr.3: 57626602-57626602 on Build GRCh38
rs1008087520 Chr.3: 57626920-57626920 on Build GRCh38
rs76957180 Chr.3: 57626875-57626875 on Build GRCh38
rs559180114 Chr.3: 57626805-57626805 on Build GRCh38
rs773258119 Chr.3: 57626790-57626790 on Build GRCh38
rs6762914 Chr.3: 57626896-57626896 on Build GRCh38
rs1042481221 Chr.3: 57626808-57626808 on Build GRCh38
rs878954242 Chr.3: 57626730-57626730 on Build GRCh38
rs887897999 Chr.3: 57626741-57626741 on Build GRCh38
rs938154709 Chr.3: 57626670-57626670 on Build GRCh38
rs953820708 Chr.3: 57626542-57626542 on Build GRCh38
rs1028200250 Chr.3: 57626527-57626527 on Build GRCh38
rs899728453 Chr.3: 57626463-57626463 on Build GRCh38
rs1001194481 Chr.3: 57626846-57626846 on Build GRCh38
rs1049262350 Chr.3: 57626633-57626633 on Build GRCh38
rs868593406 Chr.3: 57626666-57626666 on Build GRCh38
rs986223376 Chr.3: 57626569-57626569 on Build GRCh38
rs576870283 Chr.3: 57626634-57626634 on Build GRCh38
rs539174504 Chr.3: 57626821-57626822 on Build GRCh38
rs919251113 Chr.3: 57626706-57626706 on Build GRCh38
rs905364891 Chr.3: 57626444-57626444 on Build GRCh38
rs930927773 Chr.3: 57626613-57626613 on Build GRCh38
rs905376257 Chr.3: 57626658-57626658 on Build GRCh38
rs575909837 Chr.3: 57626438-57626438 on Build GRCh38
rs375395456 Chr.3: 57626475-57626475 on Build GRCh38
rs1029307193 Chr.3: 57626536-57626536 on Build GRCh38
rs1140775 Chr.3: 57626446-57626446 on Build GRCh38
rs1001910925 Chr.3: 57626462-57626462 on Build GRCh38
rs1056161705 Chr.3: 57626766-57626766 on Build GRCh38
rs1140776 Chr.3: 57626441-57626441 on Build GRCh38
rs561269249 Chr.3: 57626906-57626906 on Build GRCh38
rs896170898 Chr.3: 57626476-57626476 on Build GRCh38
rs560833732 Chr.3: 57626646-57626646 on Build GRCh38
rs987896295 Chr.3: 57626581-57626581 on Build GRCh38
rs6770988 Chr.3: 57626573-57626573 on Build GRCh38
rs1034696673 Chr.3: 57626464-57626464 on Build GRCh38

View Full Product Details