Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57627846-57628358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND6A
Entrez Gene ID
201627
Gene Name
DENN domain containing 6A
Gene Aliases
AFI1A, FAM116A
UniGene
Hs.91085
Gene Chromosome Location
Chr.3: 57625454-57693118 on Build GRCh38
Transcripts
NM_152678.2 XM_006713019.3 XM_006713020.2 XM_017005864.1 NM_001322176.1

Gene Symbol
PDE12
Entrez Gene ID
201626
Gene Name
phosphodiesterase 12
Gene Aliases
2'-PDE, 2-PDE
UniGene
Hs.572993
Gene Chromosome Location
Chr.3: 57556247-57656480 on Build GRCh38
Transcripts
NM_152678.2 XM_006713019.3 XM_006713020.2 XM_017005864.1 NM_001322176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1047706 c.1696-1G>A p.? Chr.3: 57628346-57628346 on Build GRCh38
4988340 c.1764T>A p.D588E Chr.3: 57628277-57628277 on Build GRCh38
6848977 c.1757A>G p.H586R Chr.3: 57628284-57628284 on Build GRCh38
6642521 c.1791C>T p.D597D Chr.3: 57628250-57628250 on Build GRCh38
5465944 c.1812A>C p.K604N Chr.3: 57628229-57628229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773874451 Chr.3: 57628328-57628328 on Build GRCh38
rs752407190 Chr.3: 57628232-57628232 on Build GRCh38
rs1033682989 Chr.3: 57627968-57627968 on Build GRCh38
rs1041376949 Chr.3: 57628129-57628129 on Build GRCh38
rs112847444 Chr.3: 57628292-57628292 on Build GRCh38
rs145204368 Chr.3: 57628332-57628332 on Build GRCh38
rs371159441 Chr.3: 57628175-57628175 on Build GRCh38
rs746213985 Chr.3: 57628321-57628322 on Build GRCh38
rs886590605 Chr.3: 57628107-57628107 on Build GRCh38
rs761202880 Chr.3: 57628329-57628329 on Build GRCh38
rs985220033 Chr.3: 57628281-57628281 on Build GRCh38
rs959327470 Chr.3: 57627983-57627983 on Build GRCh38
rs751065476 Chr.3: 57628246-57628246 on Build GRCh38
rs1025197045 Chr.3: 57628005-57628005 on Build GRCh38
rs772003329 Chr.3: 57628311-57628311 on Build GRCh38
rs535164850 Chr.3: 57628319-57628322 on Build GRCh38
rs749583282 Chr.3: 57628274-57628274 on Build GRCh38
rs992378756 Chr.3: 57627995-57627995 on Build GRCh38
rs781222985 Chr.3: 57628261-57628261 on Build GRCh38
rs570399791 Chr.3: 57628191-57628191 on Build GRCh38
rs77006676 Chr.3: 57628088-57628089 on Build GRCh38
rs757189097 Chr.3: 57628254-57628254 on Build GRCh38
rs967299910 Chr.3: 57628011-57628011 on Build GRCh38
rs200664403 Chr.3: 57628179-57628179 on Build GRCh38
rs755916953 Chr.3: 57628266-57628266 on Build GRCh38
rs367748277 Chr.3: 57628206-57628206 on Build GRCh38
rs772430222 Chr.3: 57628129-57628130 on Build GRCh38
rs758056981 Chr.3: 57628238-57628238 on Build GRCh38
rs929871391 Chr.3: 57628161-57628161 on Build GRCh38
rs548400247 Chr.3: 57628095-57628095 on Build GRCh38
rs751663781 Chr.3: 57628277-57628277 on Build GRCh38
rs897093128 Chr.3: 57628144-57628144 on Build GRCh38
rs148745406 Chr.3: 57628226-57628226 on Build GRCh38
rs566527979 Chr.3: 57628119-57628119 on Build GRCh38
rs561862559 Chr.3: 57628146-57628146 on Build GRCh38
rs985168304 Chr.3: 57628085-57628085 on Build GRCh38
rs769541408 Chr.3: 57628185-57628185 on Build GRCh38
rs746921865 Chr.3: 57628163-57628163 on Build GRCh38
rs181409692 Chr.3: 57627899-57627899 on Build GRCh38
rs943749941 Chr.3: 57628333-57628333 on Build GRCh38
rs779984094 Chr.3: 57628087-57628088 on Build GRCh38
rs144976169 Chr.3: 57628072-57628072 on Build GRCh38
rs374181329 Chr.3: 57628204-57628204 on Build GRCh38
rs779758729 Chr.3: 57628267-57628267 on Build GRCh38
rs978312467 Chr.3: 57628029-57628029 on Build GRCh38
rs943654999 Chr.3: 57628120-57628120 on Build GRCh38
rs925224172 Chr.3: 57627873-57627873 on Build GRCh38
rs748231125 Chr.3: 57628302-57628302 on Build GRCh38
rs758582965 Chr.3: 57627905-57627905 on Build GRCh38
rs1006256629 Chr.3: 57627955-57627955 on Build GRCh38
rs150849318 Chr.3: 57628227-57628227 on Build GRCh38
rs763154394 Chr.3: 57628193-57628193 on Build GRCh38
rs925434602 Chr.3: 57628030-57628030 on Build GRCh38
rs1047504809 Chr.3: 57628097-57628097 on Build GRCh38
rs777213921 Chr.3: 57628240-57628240 on Build GRCh38
rs774346883 Chr.3: 57628291-57628291 on Build GRCh38
rs528312209 Chr.3: 57628113-57628113 on Build GRCh38
rs989283546 Chr.3: 57628208-57628208 on Build GRCh38
rs768049711 Chr.3: 57628211-57628211 on Build GRCh38
rs780904989 Chr.3: 57628174-57628174 on Build GRCh38
rs764217541 Chr.3: 57628253-57628253 on Build GRCh38
rs370442425 Chr.3: 57628290-57628290 on Build GRCh38
rs200286921 Chr.3: 57628170-57628170 on Build GRCh38
rs201027906 Chr.3: 57628088-57628088 on Build GRCh38
rs996400717 Chr.3: 57628147-57628147 on Build GRCh38
rs185775349 Chr.3: 57628263-57628263 on Build GRCh38
rs370696444 Chr.3: 57628089-57628090 on Build GRCh38
rs966525539 Chr.3: 57628299-57628299 on Build GRCh38
rs545848612 Chr.3: 57628063-57628063 on Build GRCh38

View Full Product Details