Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58293804-58294308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD6
Entrez Gene ID
57406
Gene Name
abhydrolase domain containing 6
Gene Aliases
-
UniGene
Hs.476454
Gene Chromosome Location
Chr.3: 58237502-58294736 on Build GRCh38
Transcripts
NM_001320126.1 NM_020676.6 XM_005265334.3 XM_005265335.3 XM_017006930.1

SNP Detail

SNP ID Chromosome Location
rs556978204 Chr.3: 58293863-58293863 on Build GRCh38
rs963769745 Chr.3: 58294002-58294002 on Build GRCh38
rs933680845 Chr.3: 58294259-58294259 on Build GRCh38
rs990933621 Chr.3: 58294005-58294005 on Build GRCh38
rs191560218 Chr.3: 58294199-58294199 on Build GRCh38
rs576959384 Chr.3: 58293931-58293931 on Build GRCh38
rs1020129193 Chr.3: 58293948-58293948 on Build GRCh38
rs751847942 Chr.3: 58293866-58293866 on Build GRCh38
rs923894489 Chr.3: 58294108-58294108 on Build GRCh38
rs949511591 Chr.3: 58294047-58294047 on Build GRCh38
rs995831851 Chr.3: 58293890-58293890 on Build GRCh38
rs114913182 Chr.3: 58294034-58294034 on Build GRCh38
rs780394839 Chr.3: 58294072-58294072 on Build GRCh38
rs1030479016 Chr.3: 58293913-58293913 on Build GRCh38
rs942413086 Chr.3: 58294189-58294189 on Build GRCh38
rs781177697 Chr.3: 58293940-58293940 on Build GRCh38
rs915096590 Chr.3: 58294170-58294170 on Build GRCh38
rs35963058 Chr.3: 58294242-58294243 on Build GRCh38
rs9815089 Chr.3: 58294191-58294191 on Build GRCh38
rs1053460453 Chr.3: 58293859-58293859 on Build GRCh38
rs935205055 Chr.3: 58293825-58293825 on Build GRCh38
rs1039751774 Chr.3: 58293869-58293869 on Build GRCh38
rs2176083 Chr.3: 58294044-58294044 on Build GRCh38
rs931628928 Chr.3: 58294267-58294267 on Build GRCh38
rs75637466 Chr.3: 58294260-58294260 on Build GRCh38
rs893511705 Chr.3: 58293864-58293864 on Build GRCh38
rs1037907610 Chr.3: 58294248-58294248 on Build GRCh38
rs543673473 Chr.3: 58293860-58293860 on Build GRCh38
rs997752347 Chr.3: 58293912-58293912 on Build GRCh38
rs147044751 Chr.3: 58294138-58294138 on Build GRCh38
rs981992546 Chr.3: 58294102-58294102 on Build GRCh38
rs1006642737 Chr.3: 58293868-58293868 on Build GRCh38
rs757294057 Chr.3: 58293891-58293891 on Build GRCh38
rs575666167 Chr.3: 58293955-58293955 on Build GRCh38
rs890233749 Chr.3: 58293938-58293938 on Build GRCh38
rs900176598 Chr.3: 58293877-58293877 on Build GRCh38
rs959348385 Chr.3: 58294017-58294017 on Build GRCh38
rs978676588 Chr.3: 58293977-58293977 on Build GRCh38
rs11706920 Chr.3: 58293945-58293945 on Build GRCh38
rs935256028 Chr.3: 58294111-58294111 on Build GRCh38
rs969005812 Chr.3: 58293949-58293949 on Build GRCh38
rs545639765 Chr.3: 58293939-58293939 on Build GRCh38
rs1051456231 Chr.3: 58294268-58294268 on Build GRCh38
rs886653884 Chr.3: 58294278-58294278 on Build GRCh38
rs908040048 Chr.3: 58294142-58294142 on Build GRCh38

View Full Product Details