Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58526374-58526878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOX2
Entrez Gene ID
8309
Gene Name
acyl-CoA oxidase 2
Gene Aliases
BCOX, BRCACOX, BRCOX, THCCox
UniGene
Hs.444959
Gene Chromosome Location
Chr.3: 58505136-58537202 on Build GRCh38
Transcripts
NM_003500.3 XM_005265505.1 XM_006713340.3 XM_017007302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592368 c.1156-2A>G p.? Chr.3: 58526658-58526658 on Build GRCh38
5555437 c.1175G>A p.G392D Chr.3: 58526637-58526637 on Build GRCh38
3774113 c.1164A>G p.A388A Chr.3: 58526648-58526648 on Build GRCh38
3767526 c.1325T>C p.V442A Chr.3: 58526487-58526487 on Build GRCh38
4584581 c.1319A>T p.N440I Chr.3: 58526493-58526493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037003008 Chr.3: 58526708-58526708 on Build GRCh38
rs753332672 Chr.3: 58526533-58526533 on Build GRCh38
rs778158815 Chr.3: 58526572-58526572 on Build GRCh38
rs371116080 Chr.3: 58526573-58526573 on Build GRCh38
rs370154949 Chr.3: 58526673-58526673 on Build GRCh38
rs754620970 Chr.3: 58526480-58526480 on Build GRCh38
rs371382536 Chr.3: 58526407-58526407 on Build GRCh38
rs530318063 Chr.3: 58526587-58526587 on Build GRCh38
rs755002165 Chr.3: 58526471-58526471 on Build GRCh38
rs776721338 Chr.3: 58526495-58526495 on Build GRCh38
rs771872968 Chr.3: 58526513-58526513 on Build GRCh38
rs1055266737 Chr.3: 58526756-58526756 on Build GRCh38
rs769873539 Chr.3: 58526680-58526680 on Build GRCh38
rs751881483 Chr.3: 58526646-58526646 on Build GRCh38
rs748462053 Chr.3: 58526672-58526672 on Build GRCh38
rs969411345 Chr.3: 58526575-58526575 on Build GRCh38
rs752376288 Chr.3: 58526564-58526564 on Build GRCh38
rs760393579 Chr.3: 58526605-58526605 on Build GRCh38
rs946508061 Chr.3: 58526413-58526413 on Build GRCh38
rs1047385881 Chr.3: 58526840-58526840 on Build GRCh38
rs11553928 Chr.3: 58526698-58526698 on Build GRCh38
rs112570370 Chr.3: 58526483-58526483 on Build GRCh38
rs979677931 Chr.3: 58526841-58526841 on Build GRCh38
rs753313081 Chr.3: 58526474-58526474 on Build GRCh38
rs927910002 Chr.3: 58526639-58526639 on Build GRCh38
rs777082170 Chr.3: 58526661-58526661 on Build GRCh38
rs781551808 Chr.3: 58526583-58526583 on Build GRCh38
rs764449251 Chr.3: 58526652-58526667 on Build GRCh38
rs754647879 Chr.3: 58526803-58526803 on Build GRCh38
rs1127745 Chr.3: 58526510-58526510 on Build GRCh38
rs769932022 Chr.3: 58526504-58526504 on Build GRCh38
rs945530299 Chr.3: 58526458-58526458 on Build GRCh38
rs556651880 Chr.3: 58526769-58526769 on Build GRCh38
rs541741407 Chr.3: 58526650-58526650 on Build GRCh38
rs570151281 Chr.3: 58526606-58526606 on Build GRCh38
rs766243451 Chr.3: 58526522-58526522 on Build GRCh38
rs1044275658 Chr.3: 58526465-58526465 on Build GRCh38
rs11553929 Chr.3: 58526612-58526612 on Build GRCh38
rs1052275840 Chr.3: 58526396-58526396 on Build GRCh38
rs781590589 Chr.3: 58526654-58526654 on Build GRCh38
rs549224860 Chr.3: 58526808-58526808 on Build GRCh38
rs925595264 Chr.3: 58526467-58526467 on Build GRCh38
rs373957272 Chr.3: 58526690-58526690 on Build GRCh38
rs745958162 Chr.3: 58526503-58526503 on Build GRCh38
rs752284595 Chr.3: 58526644-58526644 on Build GRCh38
rs995406221 Chr.3: 58526749-58526749 on Build GRCh38
rs896125064 Chr.3: 58526571-58526571 on Build GRCh38
rs771371772 Chr.3: 58526492-58526492 on Build GRCh38
rs374565538 Chr.3: 58526659-58526659 on Build GRCh38
rs1002253705 Chr.3: 58526783-58526783 on Build GRCh38
rs578245933 Chr.3: 58526745-58526745 on Build GRCh38
rs768786183 Chr.3: 58526601-58526601 on Build GRCh38
rs1043848798 Chr.3: 58526428-58526428 on Build GRCh38
rs778048051 Chr.3: 58526486-58526486 on Build GRCh38
rs921062368 Chr.3: 58526846-58526846 on Build GRCh38
rs934076890 Chr.3: 58526404-58526412 on Build GRCh38
rs760613442 Chr.3: 58526520-58526520 on Build GRCh38
rs13097249 Chr.3: 58526558-58526558 on Build GRCh38
rs574320007 Chr.3: 58526693-58526693 on Build GRCh38
rs1015106314 Chr.3: 58526529-58526529 on Build GRCh38
rs761676073 Chr.3: 58526809-58526809 on Build GRCh38
rs761596161 Chr.3: 58526417-58526417 on Build GRCh38
rs760833034 Chr.3: 58526485-58526485 on Build GRCh38
rs144118647 Chr.3: 58526657-58526657 on Build GRCh38
rs1009531271 Chr.3: 58526524-58526524 on Build GRCh38
rs746437214 Chr.3: 58526586-58526586 on Build GRCh38
rs773956508 Chr.3: 58526711-58526711 on Build GRCh38
rs773003921 Chr.3: 58526603-58526603 on Build GRCh38
rs754399964 Chr.3: 58526652-58526653 on Build GRCh38
rs554563773 Chr.3: 58526677-58526677 on Build GRCh38
rs1035029142 Chr.3: 58526802-58526802 on Build GRCh38
rs1021285443 Chr.3: 58526822-58526822 on Build GRCh38
rs775393171 Chr.3: 58526426-58526427 on Build GRCh38
rs569971615 Chr.3: 58526645-58526645 on Build GRCh38
rs905278915 Chr.3: 58526761-58526761 on Build GRCh38
rs758163018 Chr.3: 58526568-58526568 on Build GRCh38
rs367613877 Chr.3: 58526667-58526667 on Build GRCh38
rs778713535 Chr.3: 58526481-58526481 on Build GRCh38
rs772915511 Chr.3: 58526519-58526519 on Build GRCh38
rs111832572 Chr.3: 58526454-58526454 on Build GRCh38
rs898022848 Chr.3: 58526714-58526714 on Build GRCh38
rs756048777 Chr.3: 58526455-58526455 on Build GRCh38
rs1050727552 Chr.3: 58526683-58526683 on Build GRCh38
rs200815295 Chr.3: 58526626-58526626 on Build GRCh38
rs543195747 Chr.3: 58526730-58526730 on Build GRCh38
rs921010127 Chr.3: 58526668-58526668 on Build GRCh38
rs768859114 Chr.3: 58526664-58526664 on Build GRCh38
rs747031412 Chr.3: 58526651-58526651 on Build GRCh38
rs1027778234 Chr.3: 58526753-58526753 on Build GRCh38
rs750356793 Chr.3: 58526442-58526442 on Build GRCh38
rs747517862 Chr.3: 58526488-58526488 on Build GRCh38
rs1127743 Chr.3: 58526648-58526648 on Build GRCh38
rs967949191 Chr.3: 58526830-58526830 on Build GRCh38
rs142358437 Chr.3: 58526596-58526596 on Build GRCh38
rs759114027 Chr.3: 58526557-58526557 on Build GRCh38
rs767277395 Chr.3: 58526432-58526432 on Build GRCh38
rs201605181 Chr.3: 58526640-58526640 on Build GRCh38
rs775080003 Chr.3: 58526684-58526684 on Build GRCh38

View Full Product Details