Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58805985-58806534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
-
UniGene
Hs.368434
Gene Chromosome Location
Chr.3: 58716415-59050078 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533460.2 XM_011533461.1 XM_011533463.2 XM_017005848.1 XM_017005849.1 XM_017005850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7078826 c.1861C>T p.R621W Chr.3: 58806396-58806396 on Build GRCh38
4893841 c.1836G>A p.M612I Chr.3: 58806421-58806421 on Build GRCh38
5731909 c.1447C>G p.Q483E Chr.3: 58806432-58806432 on Build GRCh38
5731910 c.1825C>G p.Q609E Chr.3: 58806432-58806432 on Build GRCh38
4119695 c.1439G>A p.R480H Chr.3: 58806440-58806440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918128985 Chr.3: 58806256-58806256 on Build GRCh38
rs6804653 Chr.3: 58806360-58806360 on Build GRCh38
rs1037479655 Chr.3: 58806294-58806294 on Build GRCh38
rs996219175 Chr.3: 58806326-58806326 on Build GRCh38
rs1025468336 Chr.3: 58806334-58806334 on Build GRCh38
rs770403540 Chr.3: 58806388-58806388 on Build GRCh38
rs374594874 Chr.3: 58806441-58806441 on Build GRCh38
rs113733928 Chr.3: 58806354-58806355 on Build GRCh38
rs199625612 Chr.3: 58806446-58806446 on Build GRCh38
rs766560118 Chr.3: 58806440-58806440 on Build GRCh38
rs765117804 Chr.3: 58806468-58806468 on Build GRCh38
rs545942905 Chr.3: 58806509-58806509 on Build GRCh38
rs937984174 Chr.3: 58806232-58806232 on Build GRCh38
rs757127996 Chr.3: 58806471-58806471 on Build GRCh38
rs750492496 Chr.3: 58806438-58806438 on Build GRCh38
rs1047921818 Chr.3: 58806280-58806280 on Build GRCh38
rs986827823 Chr.3: 58806064-58806064 on Build GRCh38
rs1016650205 Chr.3: 58806210-58806210 on Build GRCh38
rs990241222 Chr.3: 58806141-58806141 on Build GRCh38
rs539173742 Chr.3: 58806278-58806278 on Build GRCh38
rs758169884 Chr.3: 58806372-58806372 on Build GRCh38
rs749689211 Chr.3: 58806398-58806398 on Build GRCh38
rs796259475 Chr.3: 58806355-58806356 on Build GRCh38
rs938262582 Chr.3: 58806033-58806033 on Build GRCh38
rs762230777 Chr.3: 58806418-58806418 on Build GRCh38
rs182583245 Chr.3: 58806049-58806049 on Build GRCh38
rs966757245 Chr.3: 58806213-58806213 on Build GRCh38
rs938926097 Chr.3: 58806170-58806170 on Build GRCh38
rs547746727 Chr.3: 58806137-58806137 on Build GRCh38
rs768140092 Chr.3: 58806430-58806430 on Build GRCh38
rs572463523 Chr.3: 58806341-58806341 on Build GRCh38
rs570143564 Chr.3: 58806250-58806250 on Build GRCh38
rs973069232 Chr.3: 58806032-58806032 on Build GRCh38
rs894811231 Chr.3: 58806194-58806194 on Build GRCh38
rs750578168 Chr.3: 58806352-58806352 on Build GRCh38
rs1010149199 Chr.3: 58806361-58806361 on Build GRCh38
rs558788712 Chr.3: 58806335-58806335 on Build GRCh38
rs942798408 Chr.3: 58806092-58806092 on Build GRCh38
rs781652866 Chr.3: 58806171-58806171 on Build GRCh38
rs775320019 Chr.3: 58806396-58806396 on Build GRCh38
rs1006015060 Chr.3: 58806107-58806107 on Build GRCh38
rs756329489 Chr.3: 58806354-58806354 on Build GRCh38
rs990641232 Chr.3: 58806416-58806416 on Build GRCh38
rs576967579 Chr.3: 58806501-58806501 on Build GRCh38
rs746298888 Chr.3: 58806237-58806237 on Build GRCh38
rs372630966 Chr.3: 58806395-58806395 on Build GRCh38
rs559351939 Chr.3: 58806104-58806104 on Build GRCh38
rs886150005 Chr.3: 58806100-58806100 on Build GRCh38
rs757313119 Chr.3: 58806344-58806344 on Build GRCh38
rs774864586 Chr.3: 58806407-58806407 on Build GRCh38
rs186650048 Chr.3: 58806378-58806378 on Build GRCh38
rs954666056 Chr.3: 58806186-58806186 on Build GRCh38
rs757254166 Chr.3: 58806461-58806461 on Build GRCh38
rs59197884 Chr.3: 58806359-58806360 on Build GRCh38
rs767508386 Chr.3: 58806351-58806351 on Build GRCh38
rs781327588 Chr.3: 58806472-58806472 on Build GRCh38
rs1014142411 Chr.3: 58806071-58806071 on Build GRCh38
rs763338980 Chr.3: 58806399-58806399 on Build GRCh38
rs928790809 Chr.3: 58806492-58806492 on Build GRCh38
rs776276876 Chr.3: 58806391-58806391 on Build GRCh38
rs1030642547 Chr.3: 58806169-58806169 on Build GRCh38
rs925287063 Chr.3: 58806227-58806227 on Build GRCh38
rs1012443637 Chr.3: 58806204-58806204 on Build GRCh38
rs886642429 Chr.3: 58806286-58806286 on Build GRCh38
rs397942323 Chr.3: 58806365-58806366 on Build GRCh38
rs979513585 Chr.3: 58806215-58806215 on Build GRCh38
rs757391652 Chr.3: 58806063-58806063 on Build GRCh38
rs549998931 Chr.3: 58806189-58806189 on Build GRCh38
rs567637819 Chr.3: 58806145-58806145 on Build GRCh38
rs907438364 Chr.3: 58806167-58806167 on Build GRCh38
rs898326709 Chr.3: 58806273-58806273 on Build GRCh38
rs528083258 Chr.3: 58806127-58806127 on Build GRCh38
rs900267704 Chr.3: 58806315-58806315 on Build GRCh38
rs528751838 Chr.3: 58806024-58806025 on Build GRCh38
rs756237848 Chr.3: 58806439-58806439 on Build GRCh38
rs777430958 Chr.3: 58806443-58806443 on Build GRCh38
rs911207425 Chr.3: 58806081-58806081 on Build GRCh38
rs35756556 Chr.3: 58806279-58806280 on Build GRCh38
rs1026492219 Chr.3: 58806077-58806077 on Build GRCh38
rs768998544 Chr.3: 58806499-58806499 on Build GRCh38
rs866230996 Chr.3: 58806277-58806277 on Build GRCh38
rs762301067 Chr.3: 58806505-58806505 on Build GRCh38
rs747918717 Chr.3: 58806366-58806366 on Build GRCh38
rs537239037 Chr.3: 58806355-58806355 on Build GRCh38
rs748908695 Chr.3: 58806490-58806490 on Build GRCh38
rs535187841 Chr.3: 58806355-58806356 on Build GRCh38
rs554881003 Chr.3: 58806128-58806128 on Build GRCh38
rs536523520 Chr.3: 58806154-58806154 on Build GRCh38
rs138626633 Chr.3: 58806476-58806476 on Build GRCh38
rs1055145588 Chr.3: 58806240-58806240 on Build GRCh38
rs201804120 Chr.3: 58806448-58806448 on Build GRCh38
rs557166153 Chr.3: 58806484-58806484 on Build GRCh38
rs764390074 Chr.3: 58806400-58806400 on Build GRCh38
rs1042381289 Chr.3: 58806158-58806158 on Build GRCh38

View Full Product Details