Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58863808-58864309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
-
UniGene
Hs.368434
Gene Chromosome Location
Chr.3: 58716415-59050078 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533460.2 XM_011533461.1 XM_011533463.2 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Gene Symbol
C3orf67-AS1
Entrez Gene ID
101929238
Gene Name
C3orf67 antisense RNA 1
Gene Aliases
-
UniGene
Hs.683953
Gene Chromosome Location
Chr.3: 58824471-59019093 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533460.2 XM_011533461.1 XM_011533463.2 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1617978 c.948A>G p.L316L Chr.3: 58863828-58863828 on Build GRCh38
4812070 c.955G>A p.G319S Chr.3: 58863821-58863821 on Build GRCh38
4786889 c.958G>A p.D320N Chr.3: 58863818-58863818 on Build GRCh38
1485441 c.899C>T p.P300L Chr.3: 58863877-58863877 on Build GRCh38
3916559 c.935G>A p.S312N Chr.3: 58863841-58863841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020256793 Chr.3: 58864256-58864256 on Build GRCh38
rs79166898 Chr.3: 58863960-58863960 on Build GRCh38
rs760252763 Chr.3: 58863873-58863873 on Build GRCh38
rs527725574 Chr.3: 58864118-58864118 on Build GRCh38
rs143431166 Chr.3: 58864177-58864177 on Build GRCh38
rs547732086 Chr.3: 58864211-58864211 on Build GRCh38
rs939140562 Chr.3: 58863996-58863996 on Build GRCh38
rs182844803 Chr.3: 58863899-58863899 on Build GRCh38
rs969143242 Chr.3: 58864263-58864263 on Build GRCh38
rs532801200 Chr.3: 58863997-58863997 on Build GRCh38
rs532267349 Chr.3: 58864245-58864245 on Build GRCh38
rs781443002 Chr.3: 58863923-58863923 on Build GRCh38
rs756972411 Chr.3: 58864117-58864117 on Build GRCh38
rs1047412228 Chr.3: 58863942-58863942 on Build GRCh38
rs146056417 Chr.3: 58864043-58864043 on Build GRCh38
rs765796909 Chr.3: 58863900-58863900 on Build GRCh38
rs772866007 Chr.3: 58863866-58863866 on Build GRCh38
rs139975511 Chr.3: 58864088-58864088 on Build GRCh38
rs929628109 Chr.3: 58864194-58864194 on Build GRCh38
rs1018388174 Chr.3: 58864071-58864071 on Build GRCh38
rs754799795 Chr.3: 58863929-58863929 on Build GRCh38
rs923640595 Chr.3: 58864120-58864120 on Build GRCh38
rs774197666 Chr.3: 58863855-58863855 on Build GRCh38
rs748587127 Chr.3: 58864059-58864059 on Build GRCh38
rs968965426 Chr.3: 58864058-58864058 on Build GRCh38
rs777246674 Chr.3: 58864021-58864021 on Build GRCh38
rs769173716 Chr.3: 58864020-58864020 on Build GRCh38
rs533577832 Chr.3: 58864239-58864239 on Build GRCh38
rs1056034335 Chr.3: 58864122-58864122 on Build GRCh38
rs898319232 Chr.3: 58864202-58864202 on Build GRCh38
rs1038179085 Chr.3: 58864019-58864019 on Build GRCh38
rs994519500 Chr.3: 58864203-58864203 on Build GRCh38
rs751728049 Chr.3: 58863917-58863917 on Build GRCh38
rs780111933 Chr.3: 58863966-58863966 on Build GRCh38
rs148867431 Chr.3: 58864197-58864197 on Build GRCh38
rs533640101 Chr.3: 58863922-58863922 on Build GRCh38
rs145576422 Chr.3: 58864179-58864179 on Build GRCh38
rs939004608 Chr.3: 58864121-58864121 on Build GRCh38
rs142608286 Chr.3: 58864237-58864237 on Build GRCh38
rs769367068 Chr.3: 58864147-58864147 on Build GRCh38
rs895007301 Chr.3: 58864002-58864002 on Build GRCh38
rs978962988 Chr.3: 58864273-58864273 on Build GRCh38
rs954866732 Chr.3: 58864026-58864026 on Build GRCh38
rs879290578 Chr.3: 58864204-58864204 on Build GRCh38
rs770488908 Chr.3: 58864108-58864108 on Build GRCh38
rs895466651 Chr.3: 58863926-58863926 on Build GRCh38
rs1008243080 Chr.3: 58864208-58864208 on Build GRCh38
rs1022246408 Chr.3: 58864246-58864246 on Build GRCh38
rs535748666 Chr.3: 58864209-58864209 on Build GRCh38
rs373795177 Chr.3: 58864247-58864247 on Build GRCh38
rs749053541 Chr.3: 58863928-58863928 on Build GRCh38
rs1005884752 Chr.3: 58863983-58863983 on Build GRCh38
rs571764983 Chr.3: 58863912-58863912 on Build GRCh38
rs1030449095 Chr.3: 58864010-58864010 on Build GRCh38
rs1018085622 Chr.3: 58863990-58863990 on Build GRCh38
rs35778488 Chr.3: 58863864-58863864 on Build GRCh38
rs367576176 Chr.3: 58863910-58863910 on Build GRCh38
rs560489095 Chr.3: 58864107-58864107 on Build GRCh38
rs187521479 Chr.3: 58864205-58864205 on Build GRCh38
rs371602629 Chr.3: 58863913-58863913 on Build GRCh38
rs747656672 Chr.3: 58863861-58863861 on Build GRCh38
rs191328120 Chr.3: 58864248-58864248 on Build GRCh38
rs963891944 Chr.3: 58863993-58863993 on Build GRCh38
rs1012513033 Chr.3: 58864018-58864018 on Build GRCh38
rs770226276 Chr.3: 58864044-58864044 on Build GRCh38
rs552686621 Chr.3: 58864119-58864119 on Build GRCh38
rs964130294 Chr.3: 58864102-58864102 on Build GRCh38
rs112867020 Chr.3: 58863888-58863888 on Build GRCh38
rs13084369 Chr.3: 58864169-58864169 on Build GRCh38
rs558272980 Chr.3: 58863955-58863955 on Build GRCh38
rs766205138 Chr.3: 58863885-58863885 on Build GRCh38
rs778864224 Chr.3: 58863935-58863935 on Build GRCh38
rs772597207 Chr.3: 58863943-58863943 on Build GRCh38
rs770963109 Chr.3: 58863937-58863937 on Build GRCh38
rs144235333 Chr.3: 58864027-58864027 on Build GRCh38
rs141700320 Chr.3: 58863843-58863843 on Build GRCh38
rs115596810 Chr.3: 58864249-58864249 on Build GRCh38

View Full Product Details