Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58869883-58870405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
-
UniGene
Hs.368434
Gene Chromosome Location
Chr.3: 58716415-59050078 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533460.2 XM_011533461.1 XM_011533463.2 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Gene Symbol
C3orf67-AS1
Entrez Gene ID
101929238
Gene Name
C3orf67 antisense RNA 1
Gene Aliases
-
UniGene
Hs.683953
Gene Chromosome Location
Chr.3: 58824471-59019093 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533460.2 XM_011533461.1 XM_011533463.2 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4947882 c.348C>A p.F116L Chr.3: 58870302-58870302 on Build GRCh38
7166442 c.443C>G p.S148C Chr.3: 58870207-58870207 on Build GRCh38
3408844 c.373C>T p.R125W Chr.3: 58870277-58870277 on Build GRCh38
145178 c.422T>A p.V141D Chr.3: 58870228-58870228 on Build GRCh38
350914 c.465G>T p.K155N Chr.3: 58870185-58870185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753941195 Chr.3: 58870304-58870304 on Build GRCh38
rs763700532 Chr.3: 58870161-58870161 on Build GRCh38
rs575181998 Chr.3: 58870054-58870065 on Build GRCh38
rs928978109 Chr.3: 58870281-58870281 on Build GRCh38
rs532543663 Chr.3: 58870121-58870122 on Build GRCh38
rs893620850 Chr.3: 58870345-58870345 on Build GRCh38
rs768015877 Chr.3: 58870286-58870286 on Build GRCh38
rs755231177 Chr.3: 58870201-58870201 on Build GRCh38
rs574610243 Chr.3: 58870065-58870066 on Build GRCh38
rs770640349 Chr.3: 58870226-58870226 on Build GRCh38
rs775430600 Chr.3: 58870334-58870334 on Build GRCh38
rs888702297 Chr.3: 58869924-58869924 on Build GRCh38
rs376016930 Chr.3: 58870296-58870296 on Build GRCh38
rs554277868 Chr.3: 58869946-58869946 on Build GRCh38
rs749439727 Chr.3: 58870199-58870199 on Build GRCh38
rs796445188 Chr.3: 58869936-58869938 on Build GRCh38
rs77036152 Chr.3: 58870245-58870245 on Build GRCh38
rs535932529 Chr.3: 58869941-58869941 on Build GRCh38
rs974905211 Chr.3: 58870130-58870130 on Build GRCh38
rs769699697 Chr.3: 58870332-58870332 on Build GRCh38
rs761357782 Chr.3: 58870348-58870348 on Build GRCh38
rs527467174 Chr.3: 58869945-58869945 on Build GRCh38
rs140406193 Chr.3: 58869932-58869932 on Build GRCh38
rs567736284 Chr.3: 58870367-58870367 on Build GRCh38
rs745797883 Chr.3: 58870232-58870232 on Build GRCh38
rs370424677 Chr.3: 58870324-58870324 on Build GRCh38
rs776561565 Chr.3: 58870229-58870229 on Build GRCh38
rs780278997 Chr.3: 58870181-58870181 on Build GRCh38
rs982104580 Chr.3: 58870107-58870107 on Build GRCh38
rs756677660 Chr.3: 58870277-58870277 on Build GRCh38
rs748757191 Chr.3: 58870335-58870335 on Build GRCh38
rs985036071 Chr.3: 58870062-58870062 on Build GRCh38
rs764727543 Chr.3: 58870124-58870124 on Build GRCh38
rs773897268 Chr.3: 58870249-58870249 on Build GRCh38
rs772240975 Chr.3: 58870054-58870057 on Build GRCh38
rs895660512 Chr.3: 58870338-58870338 on Build GRCh38
rs752267864 Chr.3: 58870131-58870131 on Build GRCh38
rs764244681 Chr.3: 58870046-58870046 on Build GRCh38
rs370873815 Chr.3: 58870053-58870054 on Build GRCh38
rs768056423 Chr.3: 58870339-58870339 on Build GRCh38
rs943622099 Chr.3: 58870083-58870084 on Build GRCh38
rs1042706145 Chr.3: 58870247-58870247 on Build GRCh38
rs909505568 Chr.3: 58870072-58870072 on Build GRCh38
rs966698006 Chr.3: 58870090-58870090 on Build GRCh38
rs758985653 Chr.3: 58870122-58870122 on Build GRCh38
rs984751794 Chr.3: 58870372-58870372 on Build GRCh38
rs767266330 Chr.3: 58870352-58870352 on Build GRCh38
rs767778190 Chr.3: 58870272-58870272 on Build GRCh38
rs918819351 Chr.3: 58870262-58870262 on Build GRCh38
rs866896455 Chr.3: 58870190-58870190 on Build GRCh38
rs745705785 Chr.3: 58870325-58870325 on Build GRCh38
rs890185354 Chr.3: 58870006-58870006 on Build GRCh38
rs565152406 Chr.3: 58870018-58870018 on Build GRCh38
rs13324082 Chr.3: 58870176-58870176 on Build GRCh38
rs189150652 Chr.3: 58869968-58869968 on Build GRCh38
rs1030513372 Chr.3: 58869987-58869987 on Build GRCh38
rs377254999 Chr.3: 58870342-58870342 on Build GRCh38
rs748336375 Chr.3: 58870313-58870313 on Build GRCh38
rs540075463 Chr.3: 58870016-58870016 on Build GRCh38
rs779151426 Chr.3: 58870309-58870309 on Build GRCh38
rs1010764172 Chr.3: 58870036-58870036 on Build GRCh38
rs577072335 Chr.3: 58870102-58870102 on Build GRCh38
rs746783906 Chr.3: 58870223-58870223 on Build GRCh38
rs755048372 Chr.3: 58870305-58870305 on Build GRCh38
rs774956308 Chr.3: 58870244-58870244 on Build GRCh38
rs762180383 Chr.3: 58870267-58870267 on Build GRCh38
rs192814643 Chr.3: 58870321-58870321 on Build GRCh38
rs113389381 Chr.3: 58870365-58870365 on Build GRCh38
rs760746360 Chr.3: 58870133-58870133 on Build GRCh38
rs762578614 Chr.3: 58870152-58870152 on Build GRCh38
rs957989918 Chr.3: 58870032-58870032 on Build GRCh38
rs750886020 Chr.3: 58870276-58870276 on Build GRCh38
rs10670247 Chr.3: 58870082-58870083 on Build GRCh38
rs751920132 Chr.3: 58870171-58870171 on Build GRCh38
rs779423324 Chr.3: 58870057-58870058 on Build GRCh38
rs757593286 Chr.3: 58870175-58870175 on Build GRCh38
rs373535464 Chr.3: 58870166-58870166 on Build GRCh38
rs527860574 Chr.3: 58870379-58870379 on Build GRCh38
rs780677941 Chr.3: 58870248-58870248 on Build GRCh38
rs773678218 Chr.3: 58870354-58870354 on Build GRCh38
rs1007815729 Chr.3: 58870009-58870009 on Build GRCh38
rs891481268 Chr.3: 58869910-58869910 on Build GRCh38
rs779047644 Chr.3: 58870209-58870209 on Build GRCh38
rs769778992 Chr.3: 58870234-58870234 on Build GRCh38

View Full Product Details