Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 63952273-63952773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7
Entrez Gene ID
6314
Gene Name
ataxin 7
Gene Aliases
ADCAII, OPCA3, SCA7
UniGene
Hs.476595
Gene Chromosome Location
Chr.3: 63864557-64003462 on Build GRCh38
Transcripts
NM_000333.3 NM_001177387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6215682 c.413T>C p.F138S Chr.3: 63952397-63952397 on Build GRCh38
7447420 c.416G>C p.C139S Chr.3: 63952400-63952400 on Build GRCh38
5011139 c.419C>A p.P140Q Chr.3: 63952403-63952403 on Build GRCh38
420320 c.463C>T p.Q155* Chr.3: 63952447-63952447 on Build GRCh38
5046623 c.430G>A p.D144N Chr.3: 63952414-63952414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039853668 Chr.3: 63952563-63952563 on Build GRCh38
rs900027380 Chr.3: 63952578-63952578 on Build GRCh38
rs750516198 Chr.3: 63952576-63952576 on Build GRCh38
rs761760983 Chr.3: 63952437-63952437 on Build GRCh38
rs774268576 Chr.3: 63952428-63952428 on Build GRCh38
rs758765055 Chr.3: 63952505-63952505 on Build GRCh38
rs771002676 Chr.3: 63952512-63952512 on Build GRCh38
rs12485317 Chr.3: 63952316-63952316 on Build GRCh38
rs369999332 Chr.3: 63952421-63952421 on Build GRCh38
rs533114875 Chr.3: 63952590-63952590 on Build GRCh38
rs754561262 Chr.3: 63952481-63952481 on Build GRCh38
rs753850071 Chr.3: 63952391-63952391 on Build GRCh38
rs373439539 Chr.3: 63952509-63952509 on Build GRCh38
rs767560604 Chr.3: 63952444-63952444 on Build GRCh38
rs540528502 Chr.3: 63952591-63952591 on Build GRCh38
rs759418780 Chr.3: 63952438-63952438 on Build GRCh38
rs772707223 Chr.3: 63952313-63952313 on Build GRCh38
rs781103155 Chr.3: 63952517-63952517 on Build GRCh38
rs746010945 Chr.3: 63952533-63952533 on Build GRCh38
rs747023528 Chr.3: 63952359-63952359 on Build GRCh38
rs750395144 Chr.3: 63952448-63952448 on Build GRCh38
rs886733517 Chr.3: 63952632-63952632 on Build GRCh38
rs192106966 Chr.3: 63952461-63952461 on Build GRCh38
rs894820478 Chr.3: 63952685-63952685 on Build GRCh38
rs781627319 Chr.3: 63952559-63952559 on Build GRCh38
rs910550541 Chr.3: 63952532-63952532 on Build GRCh38
rs755063963 Chr.3: 63952517-63952518 on Build GRCh38
rs776864967 Chr.3: 63952409-63952409 on Build GRCh38
rs1003839169 Chr.3: 63952644-63952644 on Build GRCh38
rs906281860 Chr.3: 63952615-63952615 on Build GRCh38
rs1013105476 Chr.3: 63952661-63952661 on Build GRCh38
rs1005240950 Chr.3: 63952710-63952710 on Build GRCh38
rs375586094 Chr.3: 63952387-63952387 on Build GRCh38
rs1052479912 Chr.3: 63952538-63952538 on Build GRCh38
rs959834580 Chr.3: 63952324-63952324 on Build GRCh38
rs372383621 Chr.3: 63952328-63952328 on Build GRCh38
rs989823637 Chr.3: 63952585-63952585 on Build GRCh38
rs557737563 Chr.3: 63952549-63952549 on Build GRCh38
rs139423433 Chr.3: 63952654-63952654 on Build GRCh38
rs760178137 Chr.3: 63952455-63952455 on Build GRCh38
rs1035355607 Chr.3: 63952645-63952645 on Build GRCh38
rs751889955 Chr.3: 63952501-63952502 on Build GRCh38
rs1042281366 Chr.3: 63952594-63952594 on Build GRCh38
rs752446278 Chr.3: 63952497-63952497 on Build GRCh38
rs747536865 Chr.3: 63952510-63952510 on Build GRCh38
rs545773039 Chr.3: 63952473-63952473 on Build GRCh38
rs757485946 Chr.3: 63952540-63952540 on Build GRCh38
rs753052802 Chr.3: 63952493-63952493 on Build GRCh38
rs778682428 Chr.3: 63952492-63952492 on Build GRCh38
rs748269106 Chr.3: 63952663-63952663 on Build GRCh38
rs1054765777 Chr.3: 63952684-63952684 on Build GRCh38

View Full Product Details