Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 63982815-63983336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7
Entrez Gene ID
6314
Gene Name
ataxin 7
Gene Aliases
ADCAII, OPCA3, SCA7
UniGene
Hs.476595
Gene Chromosome Location
Chr.3: 63864557-64003462 on Build GRCh38
Transcripts
NM_000333.3 NM_001128149.2 NM_001177387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6199429 c.1057G>A p.D353N Chr.3: 63982983-63982983 on Build GRCh38
4458270 c.1078C>T p.R360W Chr.3: 63983004-63983004 on Build GRCh38
6483366 c.1073G>A p.C358Y Chr.3: 63982999-63982999 on Build GRCh38
6483365 c.1073G>A p.C358Y Chr.3: 63982999-63982999 on Build GRCh38
3916580 c.1062C>T p.T354T Chr.3: 63982988-63982988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761032732 Chr.3: 63982852-63982852 on Build GRCh38
rs760770524 Chr.3: 63982959-63982959 on Build GRCh38
rs932863925 Chr.3: 63983197-63983197 on Build GRCh38
rs755397411 Chr.3: 63983051-63983051 on Build GRCh38
rs552143838 Chr.3: 63983206-63983206 on Build GRCh38
rs756467582 Chr.3: 63982900-63982900 on Build GRCh38
rs905790376 Chr.3: 63983299-63983299 on Build GRCh38
rs911568898 Chr.3: 63983121-63983121 on Build GRCh38
rs754960667 Chr.3: 63983185-63983185 on Build GRCh38
rs779467999 Chr.3: 63982925-63982925 on Build GRCh38
rs765639234 Chr.3: 63983285-63983285 on Build GRCh38
rs985200091 Chr.3: 63982879-63982880 on Build GRCh38
rs953784630 Chr.3: 63982866-63982866 on Build GRCh38
rs562640418 Chr.3: 63983075-63983075 on Build GRCh38
rs973219203 Chr.3: 63982843-63982843 on Build GRCh38
rs778268055 Chr.3: 63983250-63983250 on Build GRCh38
rs926033095 Chr.3: 63983205-63983205 on Build GRCh38
rs983129365 Chr.3: 63982863-63982863 on Build GRCh38
rs983077121 Chr.3: 63983114-63983114 on Build GRCh38
rs895861931 Chr.3: 63983251-63983251 on Build GRCh38
rs779766020 Chr.3: 63983028-63983028 on Build GRCh38
rs577045411 Chr.3: 63982946-63982946 on Build GRCh38
rs1014109042 Chr.3: 63983261-63983261 on Build GRCh38
rs1044321814 Chr.3: 63983297-63983297 on Build GRCh38
rs562189517 Chr.3: 63983274-63983274 on Build GRCh38
rs544060033 Chr.3: 63983060-63983060 on Build GRCh38
rs750091665 Chr.3: 63982893-63982893 on Build GRCh38
rs374380476 Chr.3: 63983057-63983057 on Build GRCh38
rs754080640 Chr.3: 63983047-63983047 on Build GRCh38
rs780976365 Chr.3: 63982889-63982889 on Build GRCh38
rs761382914 Chr.3: 63983004-63983004 on Build GRCh38
rs969943765 Chr.3: 63983048-63983048 on Build GRCh38
rs769089955 Chr.3: 63982921-63982921 on Build GRCh38
rs143979784 Chr.3: 63983081-63983082 on Build GRCh38
rs749646697 Chr.3: 63982901-63982902 on Build GRCh38
rs140380661 Chr.3: 63983276-63983277 on Build GRCh38
rs772601095 Chr.3: 63983073-63983073 on Build GRCh38
rs372047981 Chr.3: 63982868-63982868 on Build GRCh38
rs893939281 Chr.3: 63983081-63983081 on Build GRCh38
rs755671706 Chr.3: 63983024-63983024 on Build GRCh38
rs920238194 Chr.3: 63982851-63982851 on Build GRCh38
rs749976928 Chr.3: 63983015-63983015 on Build GRCh38
rs748217496 Chr.3: 63982945-63982945 on Build GRCh38
rs773456165 Chr.3: 63982951-63982951 on Build GRCh38
rs375282353 Chr.3: 63983143-63983143 on Build GRCh38
rs544552831 Chr.3: 63983003-63983003 on Build GRCh38
rs72876981 Chr.3: 63983093-63983093 on Build GRCh38
rs186693548 Chr.3: 63982915-63982915 on Build GRCh38
rs900034897 Chr.3: 63982908-63982908 on Build GRCh38
rs368165026 Chr.3: 63982961-63982961 on Build GRCh38
rs944274916 Chr.3: 63982891-63982891 on Build GRCh38
rs932725143 Chr.3: 63982909-63982909 on Build GRCh38
rs1017104738 Chr.3: 63983260-63983260 on Build GRCh38
rs201714944 Chr.3: 63982922-63982922 on Build GRCh38
rs191602653 Chr.3: 63983228-63983228 on Build GRCh38
rs1029339145 Chr.3: 63982849-63982849 on Build GRCh38
rs950540467 Chr.3: 63983103-63983103 on Build GRCh38
rs554647153 Chr.3: 63983172-63983172 on Build GRCh38
rs994985234 Chr.3: 63983068-63983068 on Build GRCh38
rs1053013471 Chr.3: 63983212-63983212 on Build GRCh38
rs998421474 Chr.3: 63983161-63983161 on Build GRCh38
rs777390825 Chr.3: 63982932-63982932 on Build GRCh38
rs929017556 Chr.3: 63982855-63982855 on Build GRCh38
rs895696360 Chr.3: 63982977-63982977 on Build GRCh38
rs371856622 Chr.3: 63982982-63982982 on Build GRCh38
rs182954278 Chr.3: 63983307-63983307 on Build GRCh38
rs748594662 Chr.3: 63983204-63983204 on Build GRCh38
rs535054469 Chr.3: 63983044-63983044 on Build GRCh38
rs751604463 Chr.3: 63983000-63983000 on Build GRCh38
rs767174221 Chr.3: 63983005-63983005 on Build GRCh38
rs79954701 Chr.3: 63983187-63983187 on Build GRCh38
rs73834164 Chr.3: 63982902-63982902 on Build GRCh38
rs889489390 Chr.3: 63983213-63983213 on Build GRCh38
rs994657662 Chr.3: 63983300-63983300 on Build GRCh38

View Full Product Details